 |
Glassware
Designed for Organic Chemistry
System
45 is an integrated system of flasks, glassware components, and
patented PTFE adapters that incorporate the best in design, safety,
and modern materials. System 45 is compatible with existing glassware,
and can be phased-in for immediate benefits with minimal replacement
expense.
Features:
- Wide GL 32 or GL 45 screw thread flask openings
- Greaseless, gas-tight, 100% freeze-proof seals
- Chemically inert, glass-to-PTFE connections throughout
- Quick-disconnect tubing fittings
|