|
Feature |
Aldrich FT-NMR Library with Spectral Viewer |
Aldrich/ACD Pro FT-NMR Library |
| Access to Aldrich Library of
1HNMR Spectra |
Y |
Y |
| Access to Aldrich Library of
13CNMR Spectra |
Y |
Y |
| Installable on user's hard drive |
Y |
Y |
| User-created database on hard drive |
|
Y |
| Physical data window |
Y |
Y |
| Spectrum window |
Y |
Y |
|
Spectrum processing: |
Y |
Y |
|
Peak integration: Automatic |
Y |
Y |
|
Peak integration: Manual |
Y |
Y |
|
Peak labeling:
Automatic |
Y |
Y |
|
Peak labeling: Manual |
|
Y |
|
Distance measuring |
|
Y |
|
Zoom |
Y |
Y |
|
Search
according to: |
Y |
Y |
| |
Aldrich catalog number |
Y |
Y |
| |
CAS number |
Y |
Y |
| |
Chemical name |
Y |
Y |
| |
Chemical category |
Y |
|
| |
Molecular formula |
Y |
Y |
| |
Drawn structure or
sub-structure |
|
Y |
| |
Query peaks |
|
Y |
| |
Solvents |
|
Y |
| |
Portion of spectrum |
|
Y |
| |
MW, boiling point,
melting point, flash point, refractive index, or density
|
Y |
Y |
| |
Multiple fields simultaneously |
Y |
|
| Print spectra, structure and technical data |
Y |
Y |
| Customize printouts |
|
Y |
|
Export spectra to a file: |
Y |
Y |
| |
Bitmap |
Y |
Y |
| |
Metafile |
Y |
Y |
| |
JPEG |
Y |
|
| |
Paintbrush |
|
Y |
| |
TIFF |
|
Y |
| |
GIF |
|
Y |
| Web links for additional technical data |
Y |
|
| Creation of search lists which can be further modified |
|
Y |
| Duplicate, exchange, merge, subtract search lists of compounds |
|
Y |
| Save and load search lists of compounds |
|
Y |
| Import processed spectra for spectral searching |
|
Y |
| Network versions available |
Y |
Y |