Order Center
Equipment, Supplies & Books
Corning Products
Product Lines
Products by Application
Product Highlights
Precision Seal
Aldrich AtmosBag
Buchi Evaporators
Kodak BioMax Film
Microflex
MolyMod
SensIR ATR Library
Septa
SubaSeal
Whatman Membrane Selection
Books
Gases Equipment
Glassware Catalog
Key Resources
Labware Literature
New Products
 Kodak BioMax Film & Accessories
Labware
Kodak Autoradiography and Chemiluminescence Film plus Accessories
Kodak Biomax Film
Sigma-Aldrich has supplied Kodak Biomax Autoradiography and Chemiluminescence film products for years and continues today to distribute them worldwide. Check our product selection guide below. We support these products with high stock levels, sophisticated logistics, easy-to-use order systems and fast delivery to provide scientists easy access to the best products for their science. Contact us and we will be pleased to help solve any supply issues or concerns you may have.
These Kodak films are designed specifically for Autoradiography and Chemiluminescence purposes and Kodak T-Grain technology will intercept more radiation, providing enhanced sensitivity, speed and less noticeable grain in the film. With development of intensifying screens, including the innovative Transcreen, Kodak further improved the sensitivity, image quality and resolution that laboratory scientists need.
Kodak Autoradiography & Chemiluminescence Films
Description and use Packaging Size (in.) Size (cm) Kodak # Sigma-Aldrich #
BioMax Light - Chemiluminescence film
A double emulsion (double-sided) film that provides maximum clarity and sensitivity for detection chemiluminescence. Produces highest signal-to-noise ratio of any Kodak film for detecting chemiluminescence and suitable for use with all chemiluminescence kits.
Light-1: Sheets of film with no interleaved paper 5 x 7 13 x 18 868 9358 Z370398-50EA
7 x 9.5 18 x 24 819 4540 Z373508-50EA
8 x 10 20.3 x 25.4 178 8207 Z370371-50EA
9.5 x 11.8 24 x 30 153 8966 Z373516-50EA
14 x 17 35 x 43 862 5170 Z373494-50EA
Light-2: Each sheet wrapped in light-tight envelope 5 x 7 13 x 18 191 7012 Z370428-50EA
8 x 10 20.3 x 25.4 876 1520 Z370401-50EA
BioMax MR - Maximum Resolution
A single emulsion film (single-sided), provides maximum resolution and clearest background. Makes readings and digital scanning autoradiograms from sequencing gels easy and accurate.The most sensitive film for direct 14C, 35S and 33P autoradiography. Higher resolution than BioMax XAR and MS for 32P and 125I direct and screen exposures, but with reduced sensitivity.
MR-1: Sheets of film with no interleaved paper 5 x 7 13 x 18 894 1114 Z350397-50EA
7 x 9.5 18 x 24 873 6936 Z353949-50EA
7 x 17 18 x 43 875 6447 Z353957-50EA
8 x 10 20.3 x 25.4 870 1302 Z350370-50EA
9.5 x 11.8 24 x 30 891 2560 Z350427-50EA
11.8 x 15.8 30 x 40 892 9655 Z358478-50EA
14 x 17 35 x 43 871 5187 Z350419-50EA
MR-2: Each sheet wrapped in light-tight envelope 5 x 7 13 x 18 857 2786 Z350427-50EA
8 x 10 20.3 x 25.4 895 2855 Z350400-50EA
14 x 17 35 x 43 856 7232 Z350419-50EA
BioMax MS - Maximum Sensitivity
A double emulsion film (double-sided) on clear base. Extremely sensitive to the light emitted from BioMax intensifying screens. When used with intensifying screens, provides maximum sensitivity for 32P, 125I and other isotopes in Northern, Southern and Dot blot applications. Ideal for low intensity samples or when shorter exposure times needed.
MS-1: Sheets of film with no interleaved paper 5 x 7 13 x 18 111 1681 Z363022-50EA
7 x 9.5 18 x 24 822 2648 Z363030-50EA
8 x 10 20.3 x 25.4 829 4985 Z363006-50EA
9.5 x 11.8 24 x 30 832 6886 Z363049-50EA
11.8 x 15.8 30 x 40 822 0774 Z363057-50EA
14 x 17 35 x 43 143 5726 Z363014-50EA
MS-2: Each sheet wrapped in light-tight envelope 5 x 7 13 x 18 177 7853 Z363103-50EA
8 x 10 20.3 x 25.4 837 7616 Z363073-50EA
14 x 17 35 x 43 176 4505 Z363081-50EA
BioMax XAR - General Purpose
A double emulsion film (double-sided) on clear base. Very versatile, suitable for use with low and high energy isotopes, or chemiluminescence. Sensitivity with high energy isotopes can be increased by using intensifying screens
XAR: Sheets of film interleaved with sheets of paper 5 x 7 13 x 18 165 1496 F5388-50EA
8 x 10 20.3 x 25.4 165 1454 F5513-50EA
7 x 9.5 18 x 24 853 2655 Z358487-50EA
9.5 x 11.8 24 x 30 150 1451 Z358495-50EA
11.8 x 15.8 30 x 40 150 6955 Z358584-50EA
14 x 17 35 x 43 165 1512 F5263-50EA
XAR-2: Each sheet wrapped in light-tight envelope 5 x 7 13 x 18 166 0760 F5888-50EA
8 x 10 20.3 x 25.4 165 1579 F5763-50EA
14 x 17 35 x 43 165 1678 F5014-50EA
XAR-roll: 25m roll of film 20 x 2500 153 1680 F5138-1EA
X-OMAT LS - Economic, blue tint
Suitable for all isotopes and chemiluminescence, but less sensitive, requiring longer exposure times
Sheets of film interleaved with sheets of paper 5 x 7 13 x 18 161 1342 F1274-50EA
8 x 10 20.3 x 25.4 864 6770 F1149-50EA
14 x 17 35 x 43 892 6024 F0899-50EA
Kodak Autoradiography Accessories
For Film size
Description Size (in.) Size (cm) Kodak # Sigma-Aldrich #
BioMax Exposure cassette : Lightweight, durable, vinyl covered aluminum cassette with locking clip. When used at ultracold temperatures, allow to come to room temperature before opening. 8 x 10 20.3 x 25.4 820 9140 C4604-1EA
14 x 17 35 x 43 819 7600 C4854-1EA
BioMax MS Intensifying screen: Phospor emits green light, closely matched to the spectral sensitivity of BioMax MS film, to provide highest sensitivity for 32P and 125I autoradiography 7 x 9.5 18 x 24 875 7270 Z363189-1EA
8 x 10 20.3 x 25.4 851 8706 Z363154-1EA
9.5 x 11.8 24 x 30 896 7754 Z363197-1EA
11.8 x 15.8 30 x 40 877 0331 Z363200-1EA
14 x 17 35 x 43 898 1706 Z363162-1EA
BioMax MS Exposure cassette/Intensifying screen set:
Contains cassette and screen described above
8 x 10 20.3 x 25.4 875 0499 C4979-1EA
14 x 17 35 x 43 857 8825 C4854-1EA
BioMax Transcreen:
Provides improved resolution and maximum sensitivity. Optimised for use with BioMax MS film. Transcreen system places screen between blot and film, capturing low intensity radiation that does not reach other intensifying screens
LE: Low energy,for 3H, 14C, 33P, 35S, 45Ca, 59FE 8 x 10 20.3 x 25.4 162 2034 Z374253-1EA
14 x 17 35 x 43 866 0789 Z374261-1EA
HE: High energy, for 32P and 125I 8 x 10 20.3 x 25.4 856 3959 Z374318-1EA
14 x 17 35 x 43 881 1457 Z374326-1EA
BioMax MS Exposure Cassette/Transcreen set:
Contains cassette and Transcreen described above
LE: Low energy (as above) 8 x 10 20.3 x 25.4 103 0667 C5479-1EA
HE: High energy (as above) 8 x 10 20.3 x 25.4 835 6800 C5229-1EA
Intensifying screen cleaner 106 4930 Z357097-16FLOZ
Film hangers: For tank development Autoradiography film.Stainless steel frame with clips to hold film taut. 5 x 7 13 x 18 150 2723 F6013-1EA
8 x 10 20.3 x 25.4 150 2764 F6138-1EA
14 x 17 35 x 43 150 2822 F6263-1EA
Kodak Processing chemicals for Autoradiography films
Description Kodak # Sigma-Aldrich #
For manual processing GBX developer/replenisher Liquid concentrate to make up indicated volume 190 0943 P7042-1GAP7042-5GA
GBX fixer/replenisher Liquid concentrate to make up indicated volume 190 1875 P7167-1GAP7167-5GA
For automated processing systems RP X-OMAT developer/starter Packet sufficient to start 8 US gal developer 133 2642 Z354112-1PAK
RP X-OMAT developer/replenisher Packet sufficient to make up indicated volumes 124 9259
895 6799
Z354090-2x5GA
Z719692-4x4L
RP X-OMAT LO fixer/replenisher Packet sufficient to make up indicated volumes 805 5102 Z354104-2X5GA
Developer system cleaner Liquid concentrate, makes 6 US gal 101 3176 Z354120-1GA
Fixer/wash system cleaner Packet sufficient to make up indicated volumes 139 5110 Z354139-2.5GA