| Use the blotting membrane comparison table below to select the most appropriate Whatman membrane for your application. Click on the transfer membrane type to view list of available products. |
| TRANSFER MEMBRANES |
|
Protran® |
Optitran® |
Nytran® N |
Nytran® SuPerCharge |
Westran® S |
Westran® Clear Signal |
| FEATURE: |
Universal nitrocellulose blotting membrane. Superior for chemiluminescence detection using horseradish peroxidase. |
Nitrocellulose membrane with reinforcing fleece. Suitable for repeated probing. |
Nylon membrane for nucleic acid applications. Easier to block than Nytran SuPerCharge. |
Nylon membrane for nucleic acid applications. Highest binding capacity available. |
PVDF membrane for protein blotting, designed for protein sequencing. Better binding low MW proteins. |
PVDF membrane for protein blotting. Specifically designed for Western blotting. |
| MEMBRANE TYPE: |
Nitrocellulose, 100% pure |
Nitrocellulose, reinforced |
Nylon, moderately positively charged |
Nylon, highly positively charged |
PVDF |
PVDF |
| APPLICATIONS: |
Western, Southern, Northern blotting |
Western, Southern, Northern blotting |
Southern, Northern blotting |
Southern, Northern blotting |
Western blotting, Sequencing |
Western blotting |
| BINDING: |
75-110 µg/cm2
|
75-90 µg/cm2
|
>400 µg/cm2
|
>600 µg/cm2
|
>50-100 µg/cm2
|
>50-100 µg/cm2
|
| PORE SIZES: |
0.45 µm 0.2 µm 0.1 µm |
0.45 µm 0.2 µm - |
0.45 µm 0.2 µm - |
0.45 µm - - |
- 0.2 µm - |
0.45 µm - - |
| TRANSFER METHODS |
| Semi-dry Blotting |
++ |
++ |
++ |
++ |
++ |
++ |
| Tank Blotting |
++ |
++ |
++ |
++ |
++ |
++ |
| Vacuum Blotting |
++ |
++ |
++ |
++ |
+ |
+ |
| Capillary Blotting |
++ |
++ |
++ |
++ |
+ |
+ |
| Alkaline Method |
not recommended |
not recommended |
++ |
++ |
not recommended |
not recommended |
| IMMOBILIZATION |
| UV-crosslinking, DNA, RNA |
++ |
++ |
++ |
++ |
- |
- |
| Baking (80 °C), DNA, RNA |
++ |
++ |
+ |
+ |
- |
- |
| Drying, DNA, RNA |
- |
- |
+ |
+ |
- |
- |
| Drying, Protein |
++ |
++ |
- |
- |
++ |
++ |
| DETECTION METHODS |
| Colorimetric |
++ |
++ |
+ |
+ |
++ |
++ |
| Chemiluminescent |
++ |
++ |
++ |
++ |
++ |
++ |
| Isotopic |
++ |
++ |
++ |
++ |
++ |
++ |
| Fluorescent |
++ |
- |
- |
- |
- |
- |
| REPROBING |
limited |
++ |
++ |
++ |
++ |
++ |
++ Recommended + Satisfactory |
|
| Membranes with smaller pore sizes are generally recommended for the transfer of smaller molecules. A 0.2 µm pore size ensures high retention of proteins below 20 kD. For molecules below 7 kD, a 0.1 µm pore size is recommended. |
Product Lists:
|