|
The Aldrich®/SensIR® ATR-IR Library (Attenuated Total
Reflectance) contains high-resolution FT-IR spectra for use with Thermo
Electron SpecID (Z40,748-8) spectral search software.
- Suitable for doing searches using spectral subtraction.
- Contains nearly 18,000 pure compounds of the most common chemicals found in the Aldrich Handbook
- Spectra archived using 2 cm-1 data resolution and 16-bit ordinate precision
- Searchable by ATR and standard FT-IR spectra*
- Frequency range: 650-4,000 cm-1
|
|
|
Z54,751-4 |
 Infrared radiation enters the diamond's prism, which creates an
evanescent wave that extends beyond the surface of the diamond into the
sample, producing a spectrum. ATR offers a very short pathlength,
making it ideal for highly absorbent materials. The Diamond ATR
provides the highest quality spectra because it can withstand scratches from
solids, fracture from pressure, and damage from the cleaning process. *Transition spectra are fundamentally different than ATR spectra, and
current software conversions have made them interchangable. The best
method is to evaluate the reference data in the same method that the sample
was analyzed.
Multi-unit library discounts are available.
To inquire, please phone 800-231-8327 or email aldrich@sial.com.
Click here for Spectral Viewer Info.
Aldrich is a registered trademark of Sigma-Aldrich Biotechnology LP
SensIR is a registered trademark of SensIR Technologies
SpecID is a trademark of Thermo Electron Corporation
Spectral Viewer is a trademark of Sigma-Aldrich Biotechnology LP
|