Order Center
Europe Home
Austria
Belgium (Nederlands)
Belgium (Francais)
Czech Republic
Denmark
Finland
France
Germany
Greece
Hungary
Ireland
Israel
Italy
Nederland
Norway
Poland
Portugal
Russian Federation
Slovakia
South Africa
Spain
Sweden
Switzerland (Schweiz)
Switzerland (Suisse)
UK
Events
 FREQUENTLY ASKED QUESTIONS
EXPORT SALES AND SERVICE
Here are the answers on the most frequently questions
Should you have any further questions please contact us : deuexport@europe.sial.com
Why does Sigma ship frozen / refrigerated products under room temperature?
We always distinguish between shipping and stocking temperatures as in most of the cases they differ anyway. Sigma Aldrich has a lot of experience in production, handling and shipping of reagents of all kind. We dispatch our products all over the world in different climatic zones. We determinant the conditions how to ship each product as we can rely on our experiences. We guarantee quality in all times of the year. Whenever possible we ship products under room temperature in order to avoid extra charges for you for wet or dry ice shipments. All products which have to be shipped with dry or wet ice are marked in our catalogue. We also deliver products to you on wet or dry ice even if these products do not require this special transportation method under the condition that you will pay for it.
Independently from shipping instructions we set up the required stocking conditions for each item. These conditions are marked in the catalogue as well as on the label. The delivery time can be relatively short but the stocking period may be several years. Due to this reason the recommended stocking temperatures are far lower than the actual shipping temperatures.
back to top
How can I find the right Purity Grade/Quality?
Fluka and Riedel-de Häen products are classified in a system of 3 purity grades: puriss, purum and techn., for details please check on the page 07 in the FL/RDH Catalogue2005/2006.( Other quality-related data can be found on page 08 )
back to top
Where to find MSDS ( or Certificate of Analysis )?
To request a MSDS ( Certificate of Analysis ) please login and go to our Advanced Search / Related Information. If you are not registered yet, please register now.
back to top
Can I use Sigma-Aldrich products for food production or Human use?
As mentioned in our catalogues under General Information please note that our products are intended primarily for laboratory research purposes and, unless otherwise stated on product labels or catalogue, are not to be used for any other purposes, including but not limited to, in vitro diagnostic purposes, in foods, drugs, medical devices or cosmetics for humans or animals or for commercial purposes.
back to top
What are the exact freight charges / weight / dimensions for the products I requested?
The freight charges mentioned in our Proforma Invoices are always approx. as it is not possible to advice the weight / dimensions after packing in advance. Therefore the price for freight might slightly differ from time to time.
back to top
When can a product be shipped by DHL or Fedex?
Only NONHAZARD products can be shipped by DHL. If a product is hazard you can find on our webside ( go to "Advance Search" under "Safety") or in our catalogue. A UN number will be mentioned if a product is hazard. Please note that products which require to be shipped in Dry Ice can also not be shipped by DHL, as Dry Ice is classified as hazard. In this case we can check if it is possible to deliver the product with Wet Ice instead.
back to top
What does the abbreviation ECACC mean?
The European Collection of Cell Cultures (ECACC) and Sigma–Aldrich combine forces to deliver the finest in cell culture products and services. For further information please click here.
For Export ECACC cell lines are not yet available through Sigma–Aldrich, but may be ordered directly through the ECACC website. Cell lines are for research use only. Please forward your request for commercial use to ecacc@hpa.org.uk.
back to top
What is Amberlite?
Amberlite is a resin for ion exchange which can either be sold as a pre-conditioned ( which can be carried out for an extra cost for our customers ) or it can be bought and then has to be washed for preparation by the customer.For further information please click here.
back to top
Which products can be offered to train animals for drug search?
All of the listed products are not addictive for animals: KR348, P0561, P1310, P1334, P7184, P7309, P7681, P7806, P3929, P4304, P7188, P2423, P2548, B0286, N9772, N9897 or C6551.