Order Center
Europe Home
Austria
Belgium (Nederlands)
Belgium (Francais)
Czech Republic
Denmark
Finland
France
Germany
Greece
Hungary
Ireland
About Us
Consult Analytical Specialist
Customer Service
New Lab Start-Up
Online Services
Ordering
Israel
Italy
Nederland
Norway
Poland
Portugal
Russian Federation
Slovakia
South Africa
Spain
Sweden
Switzerland (Schweiz)
Switzerland (Suisse)
UK
Events
 Ascentis Express Fused-Core™ HPLC Columns
 Free Trial
Promotion Code W15
Ireland
31st December 2008
Ireland
image for fused core hplc column
Ascentis Express columns provide a breakthrough in HPLC column performance.
Based on Fused-Core™ particle technology, Ascentis Express provides the benefits of high speed and high efficiencies of sub-2 µm particles at much lower backpressures. Due to the high efficiencies at low backpressures, Ascentis Express can benefit both conventional HPLC users as well as UPLC™ or other ultra pressure system users.
Ascentis Express Extreme Performance benefits include:
  • Double the efficiencies of conventional 3 µm particles
  • Equal efficiencies of sub-2 µm columns at half of the backpressure
  • Rugged design capable of high pressure operation
Sign up to receive the Fused-Core Report! A quarterly report on the lastest applications, publications, and new products related to fused-core particle technology.
Ordering information for Ascentis Express HPLC columns
Risk Free Trial on Ascentis Express Colums, quote Promotion Code W15 to avail of this offer
Technical Resources
Publication Title Type Size (kb) No. Pages
Ascentis Express Brochure (view / request) Brochure 1,075 8
High Resolution Liquid Chromatography with Commercial HPLC Systems and Ascentis® Express HPLC Columns: Increasing Resolution by Column Coupling App Note 182 353 2
Ascentis® Express: A Fused-Core™ Particle HPLC Column for High Speed and High Efficiency Separations with Low Back Pressures App Note 181 91 2
Guidelines for Optimizing Performance with Ascentis® Express HPLC Columns Data Sheet 31 1
Supelco Introduces Ascentis® Express HPLC Columns for High Speed and High Efficiency Separations (Feb. 2007) Press Release 116 2
Trademarks
Ascentis - Sigma-Aldrich Co.
Fused-Core - Advanced Materials Technology, Inc.
UPLC - Waters Corp.