Analytical Seminar Series 2008
Supelco are holding a series of seminars and events during 2008. These events support new technology developments and application areas of interest. Click here to view the list of events and register for our seminar series.
Ascentis® Express HILIC HPLC Columns
Supelco has expanded the Ascentis Express product line, a family of innovative high-purity liquid chromatography (HPLC) columns designed to provide the analysis speed and resolving power of sub-2-micron columns with a 50% reduction in backpressure. Ascentis Express HILIC can perform high-speed and high-resolution analyses on highly polar compounds.
HILIC chromatography is complementary to reversed-phase separations due to increased retention for polar compounds. The elution order is typically the opposite of that for C18 with the non-polar compounds eluting early and the highly polar compounds having greater retention and selectivity.
Key Benefits of Ascentis Express l Double the efficiency of 3 micron columns l Same efficiency of sub 2 micron columns at half of the backpressure l Rugged design capable of ultra high pressure operationKey.
Benefits of HILIC
Retention of highly polar analytes like metabolites
lncreased MS sensitivity
Orthogonal selectivity to C18
For product information simply click here.
For ordering information simply click here.
Hydranal;The Market Leader in Reagents for Karl Fischer
Sigma-Aldrich reagents for moisture determination by Karl Fischer titration are sold under the registered trademark HydranalTM. HydranalTM is a result of our dedicated investigations into improving the safety, accuracy and ease-of-use of the Karl Fischer technique. HydranalTM reagents fulfill all the requirements of practical laboratory analyasis and enable analysts to choose the optimum reagent for their specific application and instrumentation.
Join Our Hydranal Seminar Tour this year! Register early to avoid disappointment - no registration Fee
Click Here for Details
Solvents
Sigma-Aldrich's high-purity CHROMASOLV® Solvent family delivers superior purity, stability and lot-to-lot consistency making them ideal for a variety of analytical and preparative separations utilizing LC-MS, HPLC, GPC and Spectrophometric analysis.
CHROMASOLV® solvents come in a variety of grades designed to offer customers an optimal combination of performanceand value. In order to illustrate differences between material grades, specifications for the three most common solventsemployed in reversed-phase HPLC, acetonitrile, methanol and water, are presented below. In general, specification differences
between solvent grades stem from the quantity and stringency of QC testing. It should be noted that for the HPLCgradient tests, direct comparison is often difficult due to differences in solvent programs, column type / dimensions, and detector wavelength.
Click here for our solvent application guide
Or click here for more information on all solvents
Custom Solvents/Standards are also available please email UKAnalytical specifying the components (CAS no), concentrations, solvent and volume. By clicking here and filling in the form.
Gas Purification - 15 % off SGT Gas Purifiers
No matter what the source, trace levels of hydrocarbons, moisture and oxygen in your carrier gas can adversely affect your analyses and your system. SGT Super Clean Gas Purifiers eliminate these contaminants and allow you to focus on the chromatography.
Supelco offers the unique SGT Click-On Inline Super Clean Gas Purifiers and Super Clean Gas Filters with Fast-Connect technology. The purifiers remove gas contaminants producing better than 6.0 gas quality (99.9999% purity). When used as part of a comprehensive gas management system, these state-of-the-art purifiers eliminate costly instrument downtime.
For more information on these products and how to order click here
To qualify for this offer please quote X10 on your order. Offer valid until 15th July 2008.
For any further information on these or any other products please email our Technical Services Team on EurTechServ@sial.com or call 0800 27 25 72
|