 |
Anti-mTOR (mammalian target of Rapamycin): Marker for translation initiation activation |
|
mTOR (mammalian target of Rapamycin), also named FKBP12 rapamycin-associated protein
(FRAP/RAFT/RAPT/SEP), is a serine/threonine protein kinase that is an evolutionary conserved
member of the phosphoinositol kinase-related kinase (PIKK) family that also includes DNA-PK, ATM,
ATR and several other proteins [1-5]. mTOR is involved in the regulation of cell growth through
initiation of gene translation in response to nutrients such as amino acids (mainly leucine), growth
factors, insulin and mitogens. mTOR initiates translation by activating the ribosomal p70S6k protein
kinase (S6K1) and by inhibiting the eIF4E inhibitor 4E-BP1. mTOR is thought to be involved in numerous
additional cellular functions including actin organization, membrane trafficking, secretion, protein
degradation, protein kinase C signaling, ribosome biogenesis and tRNA synthesis [1].
Rapamycin (R 0395) complexes
with the immunophilin FK-506 binding protein FKBP12 peptide prolyl cis/trans isomerase. Interaction
of this FKBP12-rapamycin complex with mTOR inhibits its function. It has been suggested that mTOR
may sense cellular ATP levels and suppress protein synthesis when ATP levels decrease [6]. mTOR is
phosphorylated at serine 2448 via the phosphatidylinositol 3-kinase (PI3K)/Akt pathway and is
autophosphorylated at serine 2481 [7,8].
Anti-mTOR (FRAP) (T 2949) is
produced using a peptide corresponding to amino acids 2433-2450 of human mTOR. The corresponding
sequence is identical in mouse and differs by one amino acid in rat. The antibody specifically
recognizes human, mouse and rat mTOR (289 kDa) and is suitable for immunoblotting, immunoprecipitation
and immunofluorescence.
|
References:
- Schmelzle, T. and Hall, M.N., Cell, 103, 253-262 (2000).
- Brown, E.J., et al., Nature, 369, 756-758 (1994).
- Sabatini, D.M, et al., Cell, 78, 35-43 (1994).
- Chiu, M.I and Berlin, V., Proc. Natl. Acad. Sci. USA, 91, 12574-12578 (1994).
- Sabers, C.J., et al., J. Biol. Chem., 270, 815-822 (1995).
- Dennis, P.B., et al., Science, 294, 1102-1105 (2001).
- Nave, B.T., et al., Biochem. J., 344, 427-431 (1999).
- Peterson, R.T., et al., J. Biol. Chem., 275, 7416-7423 (2000).
back to top |