|
Basic Antibody Structure
Antibody Classes
Normal Immunoglobulin Ranges
Basic Antibody Structure
Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma.
Table (Human)
and Table (Mouse) summarize general
properties of immunoglobulin classes. The Ig molecule in monomeric form is a glycoprotein
with a molecular weight of approximately 150 kDa that is shaped more or less like a Y.
Basic structure of the Ig monomer
[Figure]
consists of two
identical halves connected by two disulfide bonds. Each half is made up of a heavy chain
of approximately 50 kDa and a light chain of approximately 25 kDa, joined together by a
disulfide bond near the carboxyl terminus of the light chain. The heavy chain is divided
into an Fc portion, which is at the carboxyl terminal (the base of the Y), and a Fab portion,
which is at the amino terminal (the arm of the Y). Carbohydrate chains are attached to the
Fc portion of the molecule. The Fc portion of the Ig molecule is composed only of
heavy chains. Fc regions of IgG and IgM can bind to receptors on the surface of
immunomodulatory cells such as macrophages and stimulate the release of cytokines
that regulate the immune response. The Fc region contains protein sequences common to
all Igs as well as determinants unique to the individual classes. These regions are referred
to as the constant regions because they do not vary significantly among different Ig
molecules within the same class. The Fab portion of the Ig molecule contains both heavy and
light chains joined together by a single disulfide bond. One heavy and one light chain pair
combine to form the antigen binding site of the antibody. Each Ig monomer is capable of
binding two antigen molecules.
back to top
Antibody Classes
The main immunoglobulin (Ig) classes
[Figure] are IgA, IgD, IgE, IgG,
and IgM
(see Table (Human) and
Table (Mouse)).
Birds also produce
IgY in egg yolks. Class identity is determined by class-specific sequences in the Fc region
of the heavy chain which are designated by Greek letters corresponding to the Ig letter
designation: alpha-IgA, delta-IgD, epsilon-IgE, gamma-IgG, mu-IgM. Ig light chains occur
as two types — kappa or lambda. These are usually designated by the Greek letters kappa
and lambda. The ratio of kappa to lambda found in the Ig population varies by species
(see Table).
back to top
Normal Immunoglobulin Ranges
See Table.