|
PKH Fluorescent Cell Linker Kits provide fluorescent labeling of live
cells over an extended period of time, with no apparent toxic effects. PKH linker kits are
effective for a variety of cell types and exhibit no significant leaking, or transfer from cell
to cell. They provide stable, clear, intense, accurate and reproducible fluorescent labeling of
cells. Patented cell linker technology incorporates aliphatic reporter molecules into the cell
membrane lipid bilayer by selective partitioning.
Three fluorescent linkers are available. PKH2 and PKH67 are green fluorochromes with excitation
(490 nm) and emission (504 nm) similar to fluorescein, while PKH26, a red fluorochrome, has excitation
(551 nm) and emission (567 nm) characteristics compatible with rhodamine or phycoerythrin detection systems.
PKH26 may also be excited by the 488 nm emission of an argon-ion laser. The linkers are physiologically
stable and show little to no toxic side-effects on cell systems. Labeled cells retain both biological and
proliferative activity, and are ideal for cell tracking and cell-cell interaction studies.
Due to the non-specific labeling mechanism of the cell linkers, a wide variety of cell types have been
labeled successfully. The linkers have been applied to both animal and plant cells as well as other membrane
containing particles. The pattern of staining is dependent upon the cell type being labeled and the membrane
of the cells. Although most applications center around general labeling (GL) methods involving
membrane incorporation of the probes, the linkers may also be used for selective phagocytic cell labeling
(PCL). Appearance of labeled cells may vary from bright "immunofluorescence" labeling to a punctate or patchy
appearance. Since the labeling is not a saturation reaction, but rather a function of both dye and cell
concentration, it is essential that the amount of dye available for incorporation be limited. Overlabeling
of the cells will result in loss of membrane integrity and cell recovery.
Sigma developed the Cell Census Plus System by taking advantage of the PKH dye properties. It is a
proliferation assay using flow cytometry that permits measurement of the proliferative activity of
phenotypically distinct subsets within a mixed cell population. The Cell Census Plus System measures
cell proliferation without radioactive materials.
Features:
- Fluorescent labeling of live cells
- Stable for up to 100 days on live cells in vivo
- Intense and easy-to-use
- Non-cytotoxic - no effect on biological or proliferative activity
- Versatile - labels various eukaryotic cells and cell lines, bacteria, and parasites
- Uniform and reproducible
- No significant dye leakage or transfer from cell to cell
- Different dyes permit multi-color analysis
- Compatible with other fluorescent labels
- Technology that works, with hundreds of literature references to prove it
|