| Proteins and Peptides |
| Product # |
Product Name |
Application |
Add to Cart |
| B5170 |
Breast Tumor Antigen from human cell culture, ≥5,000 units/mL, buffered aqueous solution |
Tumor antigen for breast cancer. |
|
| B5295 |
Breast Tumor Antigen from human fluids, ≥10,000 units/mL, buffered aqueous solution |
Tumor antigen for breast cancer. |
|
| T3535 |
Calcitonin human ≥97% (HPLC), powder |
Hypocalcemic hormone produced by the parafollicular C cells of the thyroid or by the ultimobranchial bodies of nonmammalian vertebrates. Decreases blood calcium and phosphate due to inhibition of resorption by osteoblasts and osteocytes. |
|
| C4835 |
Carcinoembryonic Antigen from Human Fluids ≥95% (SDS-PAGE), buffered aqueous solution |
Produced by some cancers by the developing fetus. |
|
| C0150 |
Cathepsin B from human placenta lyophilized powder, ≥5 units/mg protein |
Cathepsin B has been found to cleave procaspase 1 and procaspase 11 and to induce apoptosis in digitonin-permeabilized cells. Translocation of cathepsin B from the cytoplasm to the nucleus contributes to bile salt induced apoptosis of rat hepatocytes. |
|
| C8571 |
Cathepsin B from Human Liver buffered aqueous solution, 1,000-4,000 units/mg protein (E1%/280) |
Cathepsin B has been found to cleave procaspase 1 and procaspase 11 and to induce apoptosis in digitonin-permeabilized cells. Translocation of cathepsin B from the cytoplasm to the nucleus contributes to bile salt induced apoptosis of rat hepatocytes. |
|
| C6286 |
Cathepsin B from bovine spleen lyophilized powder, ≥10 units/mg protein |
Cathepsin B has been found to cleave procaspase 1 and procaspase 11 and to induce apoptosis in digitonin-permeabilized cells. Translocation of cathepsin B from the cytoplasm to the nucleus contributes to bile salt induced apoptosis of rat hepatocytes. |
|
| C8696 |
Cathepsin D from Human Liver lyophilized powder, ≥300 units/mg protein (E1%/280) |
Cathepsin D is an endosomal-lysosomal aspartic protease implicated in breast cancer metastasis and Alzheimer’s disease. Lysosomal release of cathepsin D has been found to preceed cytochrome c release and loss of membrane potential. |
|
| C3138 |
Cathepsin D from bovine spleen lyophilized powder, ≥5 units/mg protein |
Cathepsin D is an endosomal-lysosomal aspartic protease implicated in breast cancer metastasis and Alzheimer’s disease. Lysosomal release of cathepsin D has been found to preceed cytochrome c release and loss of membrane potential. |
|
| C1109 |
CYFRA 21-1 human |
Tumor marker useful for detection of several cancer types. |
|
| N4773 |
Neuron-specific enolase from Human Brain ≥90% (SDS-PAGE), buffered aqueous solution |
Neuron-specific enolase (NSE) is expressed in all neuronal cell types, and its expression marks the acquisition of synaptic function. Following acute neuronal injury, NSE levels are increased in neuronal cell bodies. |
|
| G8535 |
Gastrointestinal Tumor Antigen from Human Cell Culture ≥10,000 units/mL, buffered aqueous solution |
Tumor marker for gastrointestinal and pancreatic cancers. |
|
| G8660 |
Gastrointestinal Tumor Antigen from Human Fluids ≥50,000 units/mL, buffered aqueous solution |
Tumor marker for gastrointestinal and pancreatic cancers. |
|
| O6008 |
Ovarian tumor antigen from Human Fluids ≥50,000 units/mL, buffered aqueous solution |
Tumor marker for ovarian cancer. |
|
| P3235 |
Prostate Specific Antigen from human semen ≥99% (SDS-PAGE), buffered aqueous solution |
Prostate Specific Antigen (PSA), a glycoprotein of the glandular kallikrein family, is a serine protease with chymotrypsin-like enzymatic activity. Circulating PSA exists in free and protease inhibitor-bound forms. Studying the ratios of free to inhibitor |
|
| P3338 |
Prostate Specific Antigen from human semen ≥95% (SDS-PAGE), buffered aqueous solution, contains 0.05% sodium azide |
Prostate Specific Antigen (PSA), a glycoprotein of the glandular kallikrein family, is a serine protease with chymotrypsin-like enzymatic activity. Circulating PSA exists in free and protease inhibitor-bound forms. |
|
| P3360 |
Prostate Specific Antigen–α1-Antichymotrypsin Complex human >99% (SDS-PAGE), buffered aqueous solution |
PSA from human seminal fluid; antichymotrypsin from human plasma. |
|
| P2985 |
Prostate Specific Antigen–α2-Macroglobulin Complex human buffered aqueous solution |
PSA from human seminal fluid; α2-macroglobulin from human plasma. |
|
| T9306 |
Tag-72 antigen from Human Fluids ≥10,000 units/mL, buffered aqueous solution |
Tumor-associated mucin; a cancer marker expressed by a majority of human carcinomas. |
|
|
|
| Product # |
Product Name |
Application |
Add to Cart |
| A4469 |
Monoclonal Anti-ACTR/AIB1 Antibody produced in mouse clone AX 15, ascites fluid |
Ascites fluid ACTR, a nuclear receptor coactivator, is also referred to as amplified in breast cancer 1 (AIB1), thyroid hormone receptor activator molecule-1 (TRAM-1), and receptor-associated coactivator 3 (RAC3 and SRC3). Immunogen: G |
|
| A2951 |
Monoclonal Anti-Alkaline Phosphatase, Human Placental Antibody produced in mouse clone 8B6, ascites fluid |
Ascites fluid Immunogen: A human epidermoid carcinoma cell line expressing placental alkaline phosphatase. Species reactivity: human |
|
| B7684 |
Monoclonal Anti-Breast Cancer Resistance Protein Antibody produced in mouse 250 μg/mL, clone BXP-34, tissue culture supernatant, aqueous solution |
Tissue culture supernatant Immunogen: Mitoxanthrone-resistant, BCRP overexpressing cell line MCF7 MR. Species reactivity: human |
|
| B7059 |
Monoclonal Anti-Breast Cancer Resistance Protein Antibody produced in mouse 250 μg/mL, clone BXP-21, tissue culture supernatant, aqueous solution |
Tissue culture supernatant Immunogen: Fusion protein containing human BCRP (amino acids 271-396) and maltose-binding protein. Species reactivity: human |
|
| B5558 |
Anti-BRCA2 Antibody produced in rabbit fractionated antiserum, buffered aqueous solution |
Immunogen: Synthetic peptide corresponding to amino acids 1323-1346 of the human BRCA2 protein coupled to KLH. Species reactivity: human |
|
| C2355 |
Anti-Calcitonin Antibody produced in rabbit whole antiserum, lyophilized powder |
Immunogen: Highly purified peptide corresponding to human calcitonin. Species reactivity: human, rat |
|
| C0715 |
Monoclonal Anti-Cathepsin D Antibody produced in mouse clone CTD-19, ascites fluid |
Ascites fluid Immunogen: Human liver cathepsin D. Species reactivity: human |
|
| C2955 |
Monoclonal Anti-CD74 Antibody produced in mouse clone LN-2, ascites fluid, Immunohistology Grade |
Ascites fluid Immunogen: Nuclei from a SU-DHL-4 diffuse histiocytic human lymphoma cell line. Species reactivity: human |
|
| C7659 |
Monoclonal Anti-Chorionic gonadotropin (β-subunit) (βHCG) Antibody produced in mouse clone PC-2, diluted ascites fluid |
Diluted Ascites fluid Immunogen: HCG. Species reactivity: human |
|
| C7534 |
Monoclonal Anti-Chorionic Gonadotropin (β-subunit) (βHCG) Antibody produced in mouse clone PE-4, ascites fluid |
Ascites fluid Immunogen: HCG. Species reactivity: human |
|
| C6930 |
Monoclonal Anti-Cytokeratin Peptide 19 Antibody produced in mouse clone A53-B/A2, tissue culture supernatant |
Tissue culture supernatant Immunogen: Human mammary carcinoma cell line MCF-7. Also cited as clone no. KS 19.1, the antibody reacts with the rod domain of cytokeratin peptide 19, a cytoskeletal protein |
|
| F5012 |
Anti-Ferritin, Human Antibody produced in rabbit whole antiserum, liquid |
Immunogen: Ferritin isolated from normal human spleen. Species reactivity: human |
|
| A8452 |
Monoclonal Anti-α-Fetoprotein (AFP) Antibody produced in mouse ascites fluid, clone C3 |
Ascites fluid Immunogen: Human α-fetoprotein. Species reactivity: human, dog, pig |
|
| L8289 |
Monoclonal Anti-LRP/MVP Antibody produced in mouse ~1 mg/mL, clone 1032, purified immunoglobulin, buffered aqueous solution |
Purified immunoglobulin Immunogen: Protein immunoprecipitated from the effluent of a phosphocellulose column loaded with the extract of human breast cancer MCF-7 cells. Species reactivity: human, rat |
|
| M5649 |
Anti-α2-Macroglobulin Antibody produced in goat whole antiserum |
Immunogen: Purified α2-macroglobulin. Species reactivity: human |
|
| M1893 |
Anti-α2-Macroglobulin Antibody produced in rabbit IgG fraction of antiserum, lyophilized powder |
Immunogen: Purified human α2-macroglobulin. Species reactivity: human |
|
| M7067 |
Monoclonal Anti-Major Vault Protein Antibody produced in mouse 250 μg/mL, clone LRP-56, tissue culture supernatant |
Tissue culture supernatant Immunogen: Drug resistant tumor cell line SW-1573/2R120. Species reactivity: human |
|
| M7192 |
Monoclonal Anti-Major Vault Protein Antibody produced in mouse 250 μg/mL, clone MVP-37, tissue culture supernatant |
Tissue culture supernatant Immunogen: Full length cloned human MVP/LRP. Species reactivity: human |
|
| M8523 |
Anti-β2-Microglobulin Antibody produced in rabbit whole antiserum |
Immunogen: β2-microglobulin. Species reactivity: human |
|
| M7398 |
Monoclonal Anti-β2-Microglobulin Antibody produced in mouse clone BM-63, ascites fluid |
Ascites fluid Immunogen: Human β2-microglobulin and a lysate of a lymphoblastoid cell line. Species reactivity: human |
|
| M5293 |
Monoclonal Anti-Mucin Gastric Antibody produced in mouse clone 45M1, ascites fluid |
Ascites fluid Immunogen: Mucin from human ovarian cyst fluid Species reactivity: human, monkey, mouse, rat, feline, rabbit, pig, hedgehog, chicken |
|
| N0649 |
Anti-Neuronal Specific Enolase Antibody produced in rabbit whole antiserum, buffered aqueous solution |
Immunogen: ?/? isoenzyme of brain NSE from human. Species reactivity: human |
|
| N217 |
Anti-Nitric Oxide Synthase, Universal Antibody produced in rabbit affinity isolated antibody, buffered aqueous solution |
Affinity isolated antibody Immunogen: Synthetic peptide derived from amino acids 1113-1122 of mouse iNOS and nNOS. The immunogen sequence is highly conserved among the different NOS isoforms. Species reactivity: mouse, rat, bovine |
|
| P6834 |
Monoclonal Anti-Proliferating Cell Protein Ki-67 Antibody produced in mouse clone PP-67, ascites fluid |
Ascites fluid Immunogen: HeLa cell nuclear preparation. Species reactivity: human |
|
| P5664 |
Anti-Prostatic Acid Phosphatase Antibody produced in rabbit whole antiserum |
Immunogen: Human prostatic acid phosphatase from prostatic fluid. Species reactivity: human |
|
| P5687 |
Monoclonal Anti-Prostatic Acid Phosphatase, Human Antibody produced in mouse clone PAP-12, ascites fluid |
Ascites fluid Immunogen: Human prostatic acid phosphatase. Species reactivity: human |
|
| P9808 |
Monoclonal Anti-Prostatic Acid Phosphatase, Human Antibody produced in mouse clone PAP-29, ascites fluid |
Ascites fluid Immunogen: Human prostatic acid phosphatase. Species reactivity: human |
|
| T5405 |
Monoclonal Anti-Thyroglobulin Antibody produced in mouse clone RBU/01, ascites fluid |
Ascites fluid Immunogen: Purified human thyroglobulin. Species reactivity: human, bovine, pig, goat, guinea pig |
|
| Back to top |