Sigma-Aldrich offers a range of medium supplements and reagents to help you optimize productivity in your CHO systems.
Designed with the biopharmaceutical industry in mind, these animal-component free products are manufactured in large scale under cGMP. |
 |
| CHO Feed Bioreactor Supplement |
| Product Code |
Product Name |
Size |
| C 1615 |
CHO Feed Bioreactor Supplement |
1L |
Animal-component free feed concentrate formulated to optimize CHO cell growth and recombinant protein production in fed-batch bioreactor cultures.
The formulation includes amino acids, vitamins, recombinant human insulin, plant hydrolysates, trace elements and other organic compounds.
| Bioreactor pH Adjustment Solution |
| Product Code |
Product Name |
Size |
| B 1185 |
Bioreactor pH Adjustment Solution |
1L |
Concentrated carbonate/bicarbonate solution designed specifically to adjust pH in bioreactor applications. Using this buffered solution to adjust the pH in bioreactors minimizes detrimental localized pH effects that can occur when using NaOH and reduces the osmolality impact when compared to using NaOH.
| Product Code |
Product Name |
Size |
| F 7050 |
Fatty Acid Supplement |
1L |
Animal-component free. Proprietary aqueous mixture of fatty acids that have been found to be optimal for cell growth and protein expression. Recommended for use in CHO cultures at a concentration of 0.25 ml/L to 0.5 ml/L of medium.
| Product Code |
Product Name |
Size |
| G 9785 |
GSEM Supplement, 50X |
1L |
The glutamine synthetase (GS) system is used to express recombinant proteins from mammalian cells. Glutamine-dependent cells transfected with a glutamine synthetase gene can be selectively grown in a suitable glutamine-deficient medium. GSEM Supplement is designed to improve cell growth and productivity in GS systems. Recommended for use in CHO cultures at a concentration of 20 ml/L of medium.
| HyPep 4601 Protein Hydrolysate from Wheat Gluten |
| Product Code |
Product Name |
Size |
| H 6784 |
HyPep 4601 Protein Hydrolysate from Wheat Gluten |
1L |
Animal-component free protein hydrolysate that is a rich source of L-glutamine. Recommended for CHO medium and feed supplementation at 1 to 5 g/L of medium.
HyPep 4601 is a trademark of Quest International.
Further Information on our selection of CHO supplements.
|