Invitrogen/Gibco Tissue Culture Media Cross Reference Chart for Sigma Aldrich Tissue culture products. Find direct replacements for Gibco cell culture products.
To place an immediate order or view package sizes and prices click on the Sigma product number.
Liquid products may be directly compared in the table. Any differences in products are noted in the righthand column.
Invitrogen/Gibco Product # |
Sigma Product # |
Product Description |
Product Differences |
| 10010023 | P3813 | Phosphate Buffered Saline, pH 7.4 | Sigma is powder, Invitrogen/Gibco is liquid |
| 10010031 | P3813 | Phosphate Buffered Saline, pH 7.4 | Sigma is powder, Invitrogen/Gibco is liquid |
| 10010049 | P3813 | Phosphate Buffered Saline, pH 7.4 | Sigma is powder, Invitrogen/Gibco is liquid |
| 10082139 | F4135 | Fetal Bovine Serum, Heat Inactivated | Fetal Bovine Serum, Heat Inactivated |
| 10082147 | F4135 | Fetal Bovine Serum, Heat Inactivated | Exact match. No differences. |
| 10082170 | F4135 | Fetal Bovine Serum, Heat Inactivated | Exact match. No differences. |
| 10099133 | 12003C | Fetal Bovine Serum, Australian Origin | Exact match. No differences. |
| 10099141 | 12003C | Fetal Bovine Serum, Australian Origin | Exact match. No differences. |
| 10099158 | 12003C | Fetal Bovine Serum, Australian Origin | Exact match. No differences. |
| 10100139 | F4135 | Fetal Bovine Serum, Heat Inactivated | Sigma is US origin, Invitrogen/Gibco is Australia origin |
| 10100147 | F4135 | Fetal Bovine Serum, Heat Inactivated | Sigma is US origin, Invitrogen/Gibco is Australia origin |
| 10100154 | F4135 | Fetal Bovine Serum, Heat Inactivated | Sigma is US origin, Invitrogen/Gibco is Australia origin |
| 10131027 | G8168 | G418 disulfate solution | Exact match. No differences. |
| 10131035 | G8168 | G418 disulfate solution | Exact match. No differences. |
| 10177012 | C3416 | Carbenicillin disodium salt | Exact match. No differences. |
| 10213015 | C7039 | Cefotaxime sodium salt | Exact match. No differences. |
| 10313021 | D6546 | Dulbecco's Modified Eagle's Medium - high glucose | Exact match. No differences. |
| 10370021 | M5650 |
Minimum Essential Medium Eagle | Exact match. No differences. |
| 10370039 | M5650 |
Minimum Essential Medium Eagle | Exact match. No differences. |
| 10372019 | M8537 |
MCDB 131 Medium | Sigma is powder, Invitrogen/Gibco is liquid |
| 10378016 | G1146 |
L-Glutamine-Penicillin-Streptomycin solution | Exact match. No differences. |
| 10437010 | F0926 |
Fetal Bovine Serum, USDA approved | Exact match. No differences. |
| 10437028 | F0926 |
Fetal Bovine Serum, USDA approved | Exact match. No differences. |
| 10437036 | F0926 |
Fetal Bovine Serum, USDA approved | Exact match. No differences. |
| 10438018 | 12306C |
Fetal Bovine Serum, Heat Inactivated | Exact match. No differences. |
| 10438026 | 12306C |
Fetal Bovine Serum, Heat Inactivated | Exact match. No differences. |
| 10438034 | 12306C |
Fetal Bovine Serum, Heat Inactivated | Exact match. No differences. |
| 10564011 | D6171 |
Dulbecco's Modified Eagle's Medium - high glucose | Sigma is without L-glutamine, Invitrogen/Gibco contains dipeptide Glutamax |
| 10565018 | D8437 |
Dulbecco's Modified Eagle's Medium/Nutrient Mixture F-12 Ham | Sigma contains L-glutamine, Invitrogen/Gibco contains dipeptide Glutamax |
| 10566016 | D0819 |
Dulbecco's Modified Eagle's Medium AQmedia | Exact Match, No differences. |
| 10569010 | D6429 |
Dulbecco's Modified Eagle's Medium - high glucose | Sigma contains L-glutamine, Invitrogen/Gibco contains dipeptide Glutamax |
| 10575090 | P9327 |
Potassium chloride solution | Exact match. No differences. |
| 10576015 | L8902 |
Phytohemagglutinin PHA-M from Phaseolus vulgaris (red kidney bean) |
Exact match. No differences. |
| 10639011 | S0192 |
Stemline Hematopoietic Stem Cell Expansion Medium |
Recommended alternative. |
| 10687010 | H0654 |
Hygromycin B solution | Exact Match, No difference. |
| 10743011 | C1490 |
EX-CELL® CD CHO-3 Media | Recommended alternative. |
| 10743029 | C1490 |
EX-CELL® CD CHO-3 Media | Recommended alternative. |
| 10744019 | S0196 |
Stemline® Keratinocyte II Medium | Recommended alternative. |
| 10907012 | G1777 |
Glial Cell Line-derived Neurotrophic Factor human | Exact match. No differences. |
| 10908010 | B3795 |
Brain-derived Neurotrophic Factor human | Exact match. No differences. |
| 11018041 | A9418 |
Albumin from bovine serum | Exact match. No differences. |
| 11039021 | D6434 |
Dulbecco's Modified Eagle's Medium/Nutrient Mixture F-12 Ham |
Sigma is without L-glutamine, Invitrogen/Gibco contains L-glutamine |
| 11039047 | D6434 |
Dulbecco's Modified Eagle's Medium/Nutrient Mixture F-12 Ham |
Sigma is without L-glutamine, Invitrogen/Gibco contains L-glutamine |
| 11043023 | M4530 |
Medium 199 | Sigma contains phenol red, Invitrogen/Gibco is without phenol red |
| 11067030 | H0137 |
HT Media Supplement (50x) Hybri-Max. | Sigma is 50X, lyophilized, Invitrogen/Gibco is 100X, liquid |
| 11090073 | M2279 |
Minimum Essential Medium Eagle | Exact match. No differences. |
| 11090081 | M2279 |
Minimum Essential Medium Eagle | Exact match. No differences. |
| 11090099 | M2279 |
Minimum Essential Medium Eagle | Exact match. No differences. |
| 11095072 | M4655 |
Minimum Essential Medium Eagle | Exact match. No differences. |
| 11095080 | M4655 |
Minimum Essential Medium Eagle | Exact match. No differences. |
| 11095098 | M4655 |
Minimum Essential Medium Eagle | Exact match. No differences. |
| 11101011 | C6164 |
Cell Freezing Medium-DMSO (1W) | Recommended alternative. |
| 11107018 | T1283 |
holo-Transferrin bovine | Exact match. No differences. |
| 11108016 | T1428 |
apo-Transferrin | Exact match. No differences. |
| 11120052 | M6895 |
MEM Vitamin Solution (100x) | Exact match. No differences. |
| 11130051 | M5550 |
MEM Amino Acids Solution (50W) | Exact match. No differences. |
| 11140050 | M7145 |
MEM Non-essential Amino Acid Solution (100x) | Exact match. No differences. |
| 11140076 | M7145 |
MEM Non-essential Amino Acid Solution (100x) | Exact match. No differences. |
| 11150059 | M4530 |
Medium 199 | Exact match. No differences. |
| 11220035 | W1625 |
Waymouth MB 752/1 Medium | Sigma is powder, Invitrogen/Gibco is liquid |
| 11279023 | 14660C |
EX-CELL® Sp2/0 Serum-Free Medium | Recommended lternative. |
| 11300027 | G9771 |
Grace's Insect Medium | Exact match. No differences. |
| 11320033 | D8062 |
Dulbecco's Modified Eagle's Medium/Nutrient Mixture F-12 Ham | Exact match. No differences. |
| 11330032 | D8437 |
Dulbecco's Modified Eagle's Medium/Nutrient Mixture F-12 Ham | Exact match. No differences. |
| 11330057 | D8437 |
Dulbecco's Modified Eagle's Medium/Nutrient Mixture F-12 Ham | Exact match. No differences. |
| 11344041 | H4034 |
HEPES | Exact match. No differences. |
| 11360070 | S8636 |
Sodium pyruvate solution | Exact match. No differences. |
| 11380037 | M8028 |
Minimum Essential Medium Eagle | Exact match. No differences. |
| 11405081 | I7760 | IPL-41 Insect Medium | Exact match. No differences. |
| 11415064 | L1518 |
L-15 Medium (Leibovitz) | Exact match. No differences. |
| 11430030 | M0275 |
Minimum Essential Medium Eagle | Exact match. No differences. |
| 11530037 | C0422 |
CMRL-1066 | Sigma is powder, Invitrogen/Gibco is liquid |
| 11550043 | N6908 |
Nutrient Mixture F-10 Ham | Exact match. No differences. |
| 11575032 | M4780 |
Minimum Essential Medium Eagle | Exact match. No differences. |
| 11593019 | A0797 |
Ampicillin sodium salt | Exact match. No differences. |
| 11605094 | G8142 |
Grace's Insect Medium | Exact match. No differences. |
| 11605102 | G8142 |
Grace's Insect Medium | Exact match. No differences. |
| 11667037 | G8142 |
Grace's Insect Medium | Sigma is 1X, Invitrogen/Gibco is 2X |
| 11681020 | M3678 |
MDCK Protein-free Medium | Recommended alternative. |
| 11700077 | M0769 |
Minimum Essential Medium Eagle | Exact match. No differences. |
| 11710035 | G5154 |
Glasgow Minimum Essential Medium | Sigma is without L-glutamine, Invitrogen/Gibco contains L-glutamine |
| 11720034 | S0146 |
Schneider's Insect Medium | Exact match. No differences. |
| 11765047 | N6658 |
Nutrient Mixture F-12 Ham | Exact match. No differences. |
| 11765054 | N6658 |
Nutrient Mixture F-12 Ham | Exact match. No differences. |
| 11765062 | N6658 |
Nutrient Mixture F-12 Ham | Exact match. No differences. |
| 11765070 | N6658 |
Nutrient Mixture F-12 Ham | Exact match. No differences. |
| 11800042 | L9010 |
Lactalbumin Enzymatic Hydrolysate | Exact match. No differences. |
| 11805017 | A9415 |
Actinomycin D | Exact match. No differences. |
| 11811023 | A1720 |
G418 disulfate salt | Non-branded form of Geneticin |
| 11811031 | A1720 |
G418 disulfate salt | Non-branded form of Geneticin |
| 11811098 | A1720 |
G418 disulfate salt | Non-branded form of Geneticin |
| 11814019 | M3536 |
Mycophenolic acid from Penicillium brevi-compactum | Exact match. No differences. |
| 11815024 | K1637 |
Kanamycin monosulfate from Streptomyces kanamyceticus |
Exact match. No differences. |
| 11815032 | K1637 |
Kanamycin monosulfate from Streptomyces kanamyceticus | Exact match. No differences. |
| 11825015 | M0650 |
Medium 199 | Exact match. No differences. |
| 11835030 | R8758 |
RPMI 1640 Medium | Sigma contains phenol red, Invitrogen/Gibco is without phenol red |
| 11835055 |
R8758 |
RPMI 1640 Medium |
Sigma contains phenol red, Invitrogen/Gibco is without phenol red |
| 11860038 |
S9137 |
Streptomycin sulfate salt |
Exact match. No differences. |
| 11875085 |
R8758 |
RPMI 1640 Medium |
Exact match. No differences. |
| 11875093 |
R8758 |
RPMI 1640 Medium |
Exact match. No differences. |
| 11875101 |
R8758 |
RPMI 1640 Medium |
Exact match. No differences. |
| 11875119 |
R8758 |
RPMI 1640 Medium |
Exact match. No differences. |
| 11875127 |
R8758 |
RPMI 1640 Medium |
Exact match. No differences. |
| 11875135 |
R8758 |
RPMI 1640 Medium |
Exact match. No differences. |
| 11885076 |
D6046 |
Dulbecco's Modified Eagle's Medium - low glucose |
Exact match. No differences. |
| 11885084 |
D6046 |
Dulbecco's Modified Eagle's Medium - low glucose |
Exact match. No differences. |
| 11885092 |
D6046 |
Dulbecco's Modified Eagle's Medium - low glucose |
Exact match. No differences. |
| 11900024 |
M0644 |
Minimum Essential Medium Eagle |
Exact match. No differences. |
| 11900073 |
M0644 |
Minimum Essential Medium Eagle |
Exact match. No differences. |
| 11905031 |
L0288 |
Lipid Mixture 1, Chemically Defined |
Exact match. No differences. |
| 11935046 |
M4655 |
Minimum Essential Medium Eagle |
Sigma is 1X, Invitrogen/Gibco is 2X |
| 11960044 |
D5671 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 11960051 |
D5671 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 11960069 |
D5671 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 11960077 |
D5671 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 11965084 |
D5796 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 11965092 |
D5796 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 11965118 |
D5796 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 11965126 |
D5796 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 11995040 |
D6429 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 11995065 |
D6429 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 11995073 |
D6429 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 11995081 |
D6429 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 12000022 |
M0894 |
Minimum Essential Medium Eagle |
Exact match. No differences. |
| 12000063 |
M0894 |
Minimum Essential Medium Eagle |
Exact match. No differences. |
| 12040077 |
H4409 |
Hybridoma Medium, Animal Component-free |
Recommended alternative. |
| 12045076 |
14610C |
EX-CELL® 610-HSF Serum-Free Medium |
Recommended alternative. |
| 12045084 |
14610C |
EX-CELL® 610-HSF Serum-Free Medium |
Recommended alternative. |
| 12052098 |
C5467 |
EX-CELL® ACF CHO Medium |
Recommended alternative. |
| 12052114 |
C5467 |
EX-CELL® ACF CHO Medium |
Recommended alternative. |
| 12100046 |
D5648 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 12100061 |
D5648 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 12200036 |
I2510 |
Iscove's Modified Dulbecco's Medium |
Exact match. No differences. |
| 12200069 |
I2510 |
Iscove's Modified Dulbecco's Medium |
Exact match. No differences. |
| 12318010 |
F1051 |
Fetal Bovine Serum, Canadian Origin |
Exact match. No differences. |
| 12318028 |
F1051 |
Fetal Bovine Serum, Canadian Origin |
Exact match. No differences. |
| 12318036 |
F1051 |
Fetal Bovine Serum, Canadian Origin |
Exact match. No differences. |
| 12319018 |
12306C |
Fetal Bovine Serum, Heat Inactivated |
Sigma is US origin, Invitrogen/Gibco is Canadian origin |
| 12319026 |
12306C |
Fetal Bovine Serum, Heat Inactivated |
Sigma is US origin, Invitrogen/Gibco is Canadian origin |
| 12319034 |
12306C |
Fetal Bovine Serum, Heat Inactivated |
Sigma is US origin, Invitrogen/Gibco is Canadian origin |
| 12338018 |
14571C |
EX-CELL® 293 Medium |
Recommended alternative. |
| 12330031 |
M8403 |
McCoy's 5A Medium |
Sigma is without L-glutamine, Invitrogen/Gibco contains L-glutamine |
| 12340030 |
M7528 |
Medium 199 |
Sigma is without L-glutamine, Invitrogen/Gibco contains L-glutamine |
| 12360038 |
M7278 |
Minimum Essential Medium Eagle |
Exact match. No differences. |
| 12390035 |
N2147 |
Nutrient Mixture F-10 Ham |
Exact match. No differences. |
| 12400024 |
D8900 |
Dulbecco's Modified Eagle's Medium/Nutrient Mixture F-12 Ham |
Exact match. No differences. |
| 12430047 |
D6171 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 12430054 |
D6171 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 12430062 |
D6171 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 12440046 |
I3390 |
Iscove's Modified Dulbecco's Medium |
Sigma is without L-glutamine, Invitrogen/Gibco contains L-glutamine |
| 12440053 |
I3390 |
Iscove's Modified Dulbecco's Medium |
Sigma is without L-glutamine, Invitrogen/Gibco contains L-glutamine |
| 12440061 |
I3390 |
Iscove's Modified Dulbecco's Medium |
Sigma is without L-glutamine, Invitrogen/Gibco contains L-glutamine |
| 12500062 |
D0547 |
Dulbecco's Modified Eagle's Medium/Nutrient Mixture F-12 Ham |
Exact match. No differences. |
| 12500096 |
D0547 |
Dulbecco's Modified Eagle's Medium/Nutrient Mixture F-12 Ham |
Exact match. No differences. |
| 12551032 |
W4128 |
Williams' Medium E |
Exact match. No differences. |
| 12561049 |
M4526 |
Minimum Essential Medium Eagle |
Sigma is without L-glutamine, Invitrogen/Gibco contains L-glutamine |
| 12561056 |
M4526 |
Minimum Essential Medium Eagle |
Sigma is without L-glutamine, Invitrogen/Gibco contains L-glutamine |
| 12571048 |
M8042 |
Minimum Essential Medium Eagle |
Sigma is without L-glutamine, Invitrogen/Gibco contains L-glutamine |
| 12571063 |
M8042 |
Minimum Essential Medium Eagle |
Sigma is without L-glutamine, Invitrogen/Gibco contains L-glutamine |
| 12571071 |
M8042 |
Minimum Essential Medium Eagle |
Sigma is without L-glutamine, Invitrogen/Gibco contains L-glutamine |
| 12591038 |
B6644 |
BGJb Medium |
Sigma is powder, Invitrogen/Gibco is liquid |
| 12800017 |
D7777 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 12800082 |
D7777 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 13007018 |
I0516 |
Insulin solution from bovine pancreas |
Sigma is liquid, Invitrogen/Gibco is powder |
| 13008016 |
T5391 |
apo-Transferrin |
Exact match. No differences. |
| 13028014 |
P1476 |
Pituitary Extract bovine |
Recommended alternative. |
| 13241013 |
F5542 |
Fibroblast Growth Factor-Acidic human |
Exact match. No differences. |
| 13244033 |
P3201 |
Platelet-Derived Growth Factor-BB human |
Exact match. No differences. |
| 13245063 |
I3769 |
Insulin-like Growth Factor-I human |
Exact match. No differences. |
| 13247051 |
E9644 |
Epidermal Growth Factor human |
Exact match. No differences. |
| 13256029 |
F0291 |
Fibroblast Growth Factor-Basic human |
Exact match. No differences. |
| 13257019 |
N6009 |
Nerve Growth Factor-2.5S from murine submaxillary gland |
Exact match. No differences. |
| 13290010 |
N0513 |
Nerve Growth Factor-7S from murine submaxillary gland |
Exact match. No differences. |
| 14025076 |
H8264 |
Hanks' Balanced Salt Solutions |
Exact match. No differences. |
| 14025092 |
H8264 |
Hanks' Balanced Salt Solutions |
Exact match. No differences. |
| 14040117 |
D8662 |
Dulbecco's Phosphate Buffered Saline |
Exact match. No differences. |
| 14040133 |
D8662 |
Dulbecco's Phosphate Buffered Saline |
Exact match. No differences. |
| 14040141 |
D8662 |
Dulbecco's Phosphate Buffered Saline |
Exact match. No differences. |
| 14040182 |
D8662 |
Dulbecco's Phosphate Buffered Saline |
Exact match. No differences. |
| 14040216 |
D8662 |
Dulbecco's Phosphate Buffered Saline |
Exact match. No differences. |
| 14065056 |
H1641 |
Hanks' Balanced Salt Solutions |
Sigma contains phenol red, Invitrogen/Gibco is without phenol red |
| 14080055 |
D1283 |
Dulbecco's Phosphate Buffered Saline |
Exact match. No differences. |
| 14170112 |
H9394 |
Hanks' Balanced Salt Solutions |
Exact match. No differences. |
| 14170120 |
H9394 |
Hanks' Balanced Salt Solutions |
Exact match. No differences. |
| 14170161 |
H9394 |
Hanks' Balanced Salt Solutions |
Exact match. No differences. |
| 14175079 |
H6648 |
Hanks' Balanced Salt Solutions |
Exact match. No differences. |
| 14175095 |
H6648 |
Hanks' Balanced Salt Solutions |
Exact match. No differences. |
| 14175103 |
H6648 |
Hanks' Balanced Salt Solutions |
Exact match. No differences. |
| 14185052 |
H4641 |
Hanks' Balanced Salt Solutions |
Sigma contains phenol red, Invitrogen/Gibco is without phenol red |
| 14190136 |
D8537 |
Dulbecco's Phosphate Buffered Saline |
Exact match. No differences. |
| 14190144 |
D8537 |
Dulbecco's Phosphate Buffered Saline |
Exact match. No differences. |
| 14190235 |
D8537 |
Dulbecco's Phosphate Buffered Saline |
Exact match. No differences. |
| 14190250 |
D8537 |
Dulbecco's Phosphate Buffered Saline |
Exact match. No differences. |
| 14200075 |
D1408 |
Dulbecco's Phosphate Buffered Saline |
Exact match. No differences. |
| 15023021 |
G7021 |
D-(+)-Glucose anhydrous |
Exact match. No differences. |
| 15040066 |
E8008 |
Ethylenediaminetetraacetic acid solution |
Exact match. No differences. |
| 15050057 |
T4424 |
Trypsin solution from porcine pancreas |
Exact match. No differences. |
| 15050065 |
T4424 |
Trypsin solution from porcine pancreas |
Exact match. No differences. |
| 15070063 |
P4458 |
Penicillin - Streptomycin Solution (50W) Stabilized |
Exact match. No differences. |
| 15090046 |
T4549 |
Trypsin solution from porcine pancreas |
Exact match. No differences. |
| 15140122 |
P4333 |
Penicillin - Streptomycin Solution (100x) Stabilized |
Exact match. No differences. |
| 15140148 |
P4333 |
Penicillin - Streptomycin Solution (100x) Stabilized |
Exact match. No differences. |
| 15160054 |
K0129 |
Kanamycin solution from Streptomyces kanamyceticus |
Exact match. No differences. |
| 15210040 |
D1925 |
Colcemid. solution |
Exact match. No differences. |
| 15212012 |
D1925 |
Colcemid. solution |
Sigma in HBSS, Invitrogen/Gibco in PBS |
| 15230147 |
W3500 |
Water, Tissue Culture Grade |
Exact match. No differences. |
| 15230162 |
W3500 |
Water, Tissue Culture Grade |
Exact match. No differences. |
| 15230196 |
W3500 |
Water, Tissue Culture Grade |
Exact match. No differences. |
| 15230204 |
W3500 |
Water, Tissue Culture Grade |
Exact match. No differences. |
| 15240062 |
A5955 |
Antibiotic Antimycotic Solution, stabilized (100x) |
Exact match. No differences. |
| 15240096 |
A5955 |
Antibiotic Antimycotic Solution, stabilized (100x) |
Exact match. No differences. |
| 15250061 |
T8154 |
Trypan Blue solution |
Exact match. No differences. |
| 15260037 |
A8412 |
Albumin solution from bovine serum |
Exact match. No differences. |
| 15290018 |
A2942 |
Amphotericin B preparation |
Exact match. No differences. |
| 15295017 |
A9528 |
Amphotericin B solubilized |
Recommended alternative. |
| 15305014 |
T3924 |
Trypsin-EDTA solution |
Sigma is liquid in HBSS, Invitrogen/Gibco is lyophilized from 0.9% NaCl |
| 15400054 |
T4174 |
Trypsin solution from porcine pancreas |
Exact match. No differences. |
| 15630080 |
H0887 |
HEPES solution |
Exact match. No differences. |
| 15630106 |
H0887 |
HEPES solution |
Exact match. No differences. |
| 15640055 |
P4083 |
Penicillin - Streptomycin - Neomycin solution stabilized (100x) |
Exact match. No differences. |
| 15710064 |
G1272 |
Gentamicin solution |
Exact match. No differences. |
| 15710072 |
G1272 |
Gentamicin solution |
Exact match. No differences. |
| 15750060 |
G1397 |
Gentamicin solution |
Exact match. No differences. |
| 15750078 |
G1397 |
Gentamicin solution |
Exact match. No differences. |
| 16000036 |
F2442 |
Fetal Bovine Serum |
Exact match. No differences. |
| 16000044 |
F2442 |
Fetal Bovine Serum |
Exact match. No differences. |
| 16000069 |
F2442 |
Fetal Bovine Serum |
Exact match. No differences. |
| 16010142 |
N4637 |
Newborn Calf Serum |
Sigma is US origin, Invitrogen/Gibco is New Zealand origin |
| 16010159 |
N4637 |
Newborn Calf Serum |
Sigma is US origin, Invitrogen/Gibco is New Zealand origin |
| 16010167 |
N4637 |
Newborn Calf Serum |
Sigma is US origin, Invitrogen/Gibco is New Zealand origin |
| 16030074 |
12133C |
Calf Serum |
Sigma is US origin, Invitrogen/Gibco is New Zealand origin |
| 16050114 |
H1270 |
Horse Serum |
Sigma is US origin, Invitrogen/Gibco is New Zealand origin |
| 16050122 |
H1270 |
Horse Serum |
Sigma is US origin, Invitrogen/Gibco is New Zealand origin |
| 16050130 |
H1270 |
Horse Serum |
Sigma is US origin, Invitrogen/Gibco is New Zealand origin |
| 16070096 |
S2263 |
Sheep Serum |
Sigma is US origin, Invitrogen/Gibco is New Zealand origin |
| 16110082 |
C5405 |
Chicken Serum |
Sigma is US origin, Invitrogen/Gibco is New Zealand origin |
| 16120099 |
R4505 |
Rabbit serum |
Sigma is US origin, Invitrogen/Gibco is New Zealand origin |
| 16120107 |
R4505 |
Rabbit serum |
Sigma is US origin, Invitrogen/Gibco is New Zealand origin |
| 16140063 |
F4135 |
Fetal Bovine Serum, Heat Inactivated |
Recommended higher quality alternative. |
| 16140071 |
F4135 |
Fetal Bovine Serum, Heat Inactivated |
Recommended higher quality alternative. |
| 16140089 |
F4135 |
Fetal Bovine Serum, Heat Inactivated |
Recommended higher quality alternative. |
| 16170060 |
12133C |
Calf Serum |
Sigma is US origin, Invitrogen/Gibco is New Zealand origin |
| 16170078 |
12133C |
Calf Serum |
Sigma is US origin, Invitrogen/Gibco is New Zealand origin |
| 16170086 |
12133C |
Calf Serum |
Sigma is US origin, Invitrogen/Gibco is New Zealand origin |
| 16210064 |
G6767 |
Goat Serum Donor Herd |
Sigma is US origin, Invitrogen/Gibco is New Zealand origin |
| 16210072 |
G6767 |
Goat Serum Donor Herd |
Sigma is US origin, Invitrogen/Gibco is New Zealand origin |
| 16600082 |
M8403 |
McCoy's 5A Medium |
Sigma is without L-glutamine, Invitrogen/Gibco contains L-glutamine |
| 16600108 |
M8403 |
McCoy's 5A Medium |
Sigma is without L-glutamine, Invitrogen/Gibco contains L-glutamine |
| 17005042 |
S0196 |
Stemline® Keratinocyte Medium |
Recommended product, consult Certificate of Analysis for exact specifications |
| 17018029 |
C9891 |
Collagenase from Clostridium histolyticum |
Recommended product, consult Certificate of Analysis for exact specifications |
| 17075029 |
T6522 |
Trypsin inhibitor from Glycine max (soybean) |
Recommended alternative. |
| 17100017 |
C6885 |
Collagenase from Clostridium histolyticum |
Recommended product, consult Certificate of Analysis for exact specifications |
| 17101015 |
C9263 |
Collagenase from Clostridium histolyticum |
Recommended product, consult Certificate of Analysis for exact specifications |
| 17104019 |
C5138 |
Collagenase from Clostridium histolyticum |
Recommended product, consult Certificate of Analysis for exact specifications |
| 17105041 |
P3417 |
Dispase from Bacillus polymyxa |
Recommended product, consult Certificate of Analysis for exact specifications |
| 18180059 |
Y1000 |
Yeast extract |
Sigma is powder, Invitrogen/Gibco is liquid |
| 18200048 |
Y4375 |
Yeast Extract Ultrafiltrate (50W) |
Exact match. No differences. |
| 20012027 |
P3288 |
Phosphate buffer pH 7.2 at 25° C |
Sigma is powder, Invitrogen/Gibco is liquid |
| 20012043 |
P3288 |
Phosphate buffer pH 7.2 at 25° C |
Sigma is powder, Invitrogen/Gibco is liquid |
| 20012050 |
P3288 |
Phosphate buffer pH 7.2 at 25° C |
Sigma is powder, Invitrogen/Gibco is liquid |
| 21010046 |
B1522 |
Basal Medium Eagle |
Sigma is without L-glutamine, Invitrogen/Gibco contains L-glutamine |
| 21013024 |
D0422 |
Dulbecco's Modified Eagle's Medium - high glucose |
Exact match. No differences. |
| 21051024 |
G8540 |
L-Glutamine |
Exact match. No differences. |
| 21051040 |
G8540 |
L-Glutamine |
Exact match. No differences. |
| 21063029 |
D1145 |
Dulbecco's Modified Eagle's Medium - high glucose |
Sigma is without L-glutamine, Invitrogen/Gibco contains L-glutamine |
| 21083027 |
L1518 |
L-15 Medium (Leibovitz) |
Sigma contains phenol red, Invitrogen/Gibco is without phenol red |
| 21127022 |
N3520 |
Nutrient Mixture F-12 Ham |
Sigma is powder, Invitrogen/Gibco is liquid |
| 21300025 |
D5773 |
Dulbecco's Phosphate Buffered Saline |
Sigma's does not contain CaCl2, Invitrogen/Gibco supplies a bottle of CaCl2. |
| 21300058 |
D5773 |
Dulbecco's Phosphate Buffered Saline |
Exact match. No differences. |
| 21340039 |
N1140 |
NCTC 109 Medium |
Exact match. No differences. |
| 21600010 |
D5652 |
Dulbecco's Phosphate Buffered Saline |
Exact match. No differences. |
| 21600044 |
D5652 |
Dulbecco's Phosphate Buffered Saline |
Exact match. No differences. |
| 21600069 |
|