Chemical Attributes of Thiamine that make it a Useful Serum-Free Medium Supplement: Thiamine is a substituted pyrimidine linked to a substituted thiazole by a methylene group. It is generally added to cell culture as thiamine-2HCl.
- Formula C12H18Cl2N4OS
- Molecular Weight = 337.28
- Very water soluble
- Unstable in aqueous solutions above pH 5.5.
- May react with other media components-pyridoxal, iron, pyruvate (other alpha keto acids), and free radicals.
- Oxidizes to thiochrome.
The metabolically active form of thiamine is the diphosphate. It is formed by the enzymatic replacement of the hydroxyl group of the thiazole 5-hydroxyethyl side chain with pyrophosphate. Its biochemical activity is mediated by the thiazole moiety. The reactive chemistry of thiamine and thiamine pyrophosphate should be similar in solution.
Possible Chemical Instability in Cell Culture: Thiamine is not a very stable molecule. It can participate in a wide range of reactions that result in its destruction.
The metabolically relevant reaction site of thiamine and TPP is carbon 2 of the thiazole ring. It is situated between nitrogen and sulfur atoms. The proton on this carbon is acidic and at pH above 5.5 it dissociates to form a carbanion which undergoes nucleophilic addition to carbonyl groups. In the presence of alpha keto acids, such as pyruvate or alpha keto glutarate, thiamine may form thiamine:keto-acid adducts. The redox and pH conditions in the media will affect the degradation of these keto acids into aldehydes (reducing conditions) or acids (oxidizing conditions) and carbon dioxide.
Thiamine and TPP contain a primary amine on the pyrimidine moiety. This primary amine may form a Schiff base with pyridoxal or other aldehydes. In the presence of transition metals, such as iron, Schiff bases may cause the destruction of thiamine.
Oxidizing and reducing agents can destroy thiamine. One oxidation product is thiochrome. This molecule that may be detected by its fluorescence under uv light. Hypochlorite, sulfites and SO2 degrade thiamine. Sulfur dioxide reacts irreversibly with thiamine to yield pyrimidine sulfonic acid and 4-methylhydroxyethyl thiazole. These molecules can form in oxidatively stressed media.
Mechanism of Action: TPP is a coenzyme for two types of enzymes, alpha-keto acid dehydrogenases and transketolases, both of which cleave a C-C bond adjacent to a carbonyl group releasing either carbon dioxide or an aldehyde. The resulting product is then transferred to an acceptor molecule.
Alpha-keto acid dehydrogenases decarboxylate alpha-keto acids. The decarboxylation product is then transferred to coenzyme A (CoA).
Transketolase cleaves the C-C bond adjacent to the carbonyl group of an alpha-keto sugar to give an activated glycoaldehyde. The glycoaldehyde is then combined with an aldose to give a new ketose.
|