Product Lines
Antibody Expression
Classic Media & Salts
Coated Plates
ECACC Cell Lines
Embryo Culture
Gene Therapy
Primary Human Culture
Protein Expression/CHO
Reagents & Supplements
Sera
Serum Free Media
Stem Cells
SyntheChol NS0
TrypZean Solution
Vaccine Production
Product Highlights
Key Resources
Cell Signaling and Neuroscience
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 Cell Lines
Cell Culture
Sigma–Aldrich, a world leader in the development, optimization and manufacture of cell culture media and reagents, has formed a working partnership with ECACC, a world leader and recognized expert in the maintenance, cultivation and distribution of authenticated cell lines.
By working together we offer a comprehensive range of cell culture and genomic DNA products along with services available to satisfy your research needs.
Cell lines not offered for sale in the United States through Sigma-Aldrich may be ordered directly from ECACC.
Cell Name
Product # 
Species of Origin
Basal Media to Supply (Europe Only)
Hep G2
85011430
Human
M2279, Minimum Essential Medium Eagle
THP 1
88081201
Human
MRC–5
84101801
Human
M2279, Minimum Essential Medium Eagle
HeLa
93021013
Human
M2279, Minimum Essential Medium Eagle
A549
86012804
Human
D6546, Dulbecco's Modified Eagle's Medium – high glucose
CHO–K1
85051005
Hamster
N4888, Nutrient Mixture F–12 Ham
Vero
84113001
Monkey
D6546, Dulbecco's Modified Eagle's Medium – high glucose
U937
85011440
Human
RAW 264.7
91062702
Mouse
D6546, Dulbecco's Modified Eagle's Medium – high glucose
SH–SY5Y
94030304
Human
N4888:M2279 (1:1), Nutrient Mixture F–12 Ham:Minimum Essential Medium Eagle
MCF7
86012803
Human
M2279, Minimum Essential Medium Eagle
K562
89121407
Human
NSO – Exclusive
85110503
Mouse
C2C12
91031101
Mouse
D6546, Dulbecco's Modified Eagle's Medium – high glucose
MDCK
84121903
Dog
M2279, Minimum Essential Medium Eagle
BHK 21 (clone 13)
85011433
Hamster
G5154, Glasgow Minimum Essential Medium
A431
85090402
Human
M2279, Minimum Essential Medium Eagle
COS–1
88031701
Monkey (SV40 transformed)
D6546, Dulbecco's Modified Eagle's Medium – high glucose
LNCap.FGC
89110211
Human
U373 MG
89081403
Human
M2279, Minimum Essential Medium Eagle
MDA–MB–231
92020424
Human
L5520, L–15 Medium (Leibovitz)
SP2/0–Ag14
85072401
Mouse
J774.2 – Exclusive
85011428
Mouse
D6546, Dulbecco's Modified Eagle's Medium – high glucose
Fao – Exclusive
89042701
Rat
VOUCHER
ISHIKAWA – Exclusive
99040201
Human
M2279, Minimum Essential Medium Eagle
OE33 – Exclusive
96070808
Human
RPMI 8866 – Exclusive
95041316
Human
Hep–2C (HeLa Derivative) – Exclusive
85020207
Human
NB2–11 – Exclusive
97041101
Rat
A2780cis – Exclusive
93112517
Human
MDBK
90050801
Bovine
M2279, Minimum Essential Medium Eagle
L929
85011425
Mouse
D6546, Dulbecco's Modified Eagle's Medium – high glucose
EL4
85023105
Mouse
D6546, Dulbecco's Modified Eagle's Medium – high glucose
TF1
93022307
Human