Families of basal salt solutions, generally named after
the inventor, have been developed around the early discovery that cells
have a basic requirement for five ions: calcium, magnesium, potassium, sodium,
and phosphate. Each balanced salts family of products differs in the concentration and
"salt form" of these ions.
Please note that products
listed as "Other Salt Solutions" may have uses other than the
culturing of cells and that they may not contain all ions listed above.
Probably the three most important basal salt solutions for cell culture are
Dulbecco's phosphate buffered saline (D-PBS); Earle's balanced salts (EBSS) and
Hanks' balanced salts (HBSS). Dulbecco's formulation contains no
bicarbonate and hence is less susceptible to pH shift during filtration.
Earle's formulation contains 2.2 gm/L of sodium bicarbonate, and Hanks'
formulation contains 0.35 gm/L of sodium bicarbonate. Both of these
formulations may require pH adjustment after filtration.
D-PBS is offered with and without glucose as an energy source. The complete formulae for EBSS and HBSS both contain 1.0 mg/L of glucose.
| Product # |
Product Name |
Description |
Application |
| D 8662 |
Dulbecco's Phosphate Buffered Saline |
- Liquid
- With calcium chloride and magnesium chloride
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Known as D-PBS, this is a general use,
isotonic saline solution for washing cells and tissues. |
| D 1283 |
Dulbecco's Phosphate Buffered Saline
(10x) |
- Liquid
- With calcium chloride and magnesium chloride
- pH 4.2-4.6 at 10x
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Concentrated
form of D 8662. |
| D 8537 |
Dulbecco's Phosphate Buffered Saline Modified |
- Liquid
- Without calcium chloride and magnesium chloride
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Calcium and magnesium promote
cell adhesion. Use this formulation of D-PBS when tissue or cell dissociation or release is the objective.
|
| D 1408 |
Dulbecco's Phosphate Buffered Saline
(10x) |
- Liquid
- Sterile-filtered
- pH 6.6-7.0 at 10x
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Concentrated
form of D 8537. |
| D 5652 |
Dulbecco's Phosphate Buffered Saline Modified |
- Powder
- Without calcium chloride and magnesium chloride
- Formulated at 9.6 grams of powder per liter of medium
- Related products: Dulbecco's Phosphate Buffered Saline
Hybri-Max (D 7030)
- Cell culture tested
|
Powder
form of D 8537.
Calcium and magnesium
promote cell adhesion.
Use this formulation of
D-PBS when tissue or cell dissociation or release is the objective.
|
| D 7030 |
Dulbecco's Phosphate Buffered Saline Modified
Hybri-Max. |
- Powder
- Without calcium chloride and magnesium chloride
- Formulated at 9.6 grams of powder per liter of medium
- Endotoxin tested
- Hybridoma tested
|
Same
as D 5652 except it is tested for use with hybridoma cells. |
| D 5773 |
Dulbecco's Phosphate Buffered Saline |
- Powder
- Without calcium chloride
- Formulated at 9.7 grams of powder per liter of medium
- Cell culture tested
|
Calcium
promotes cell adhesion.
Use this formulation of
D-PBS when tissue or cell dissociation or release is the objective.
Also, suitable for
studies that involve calcium metabolism.
|
| D 4031 |
Dulbecco's Phosphate Buffered Saline Modified |
- Liquid
- With 36 mg/L sodium pyruvate, 50 mg/L streptomycin
sulfate, 100 mg/L kanamycin monosulfate, 1000 mg/L
glucose, calcium chloride, and magnesium chloride.
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Fully
supplemented isotonic solution.
Contains energy sources and
antibiotics.
Frequently used with
primary cells where risk of contamination may be high.
|
| D 6650 |
Dulbecco's Phosphate Buffered Saline Modified |
- Powder
- With 36 mg/L sodium pyruvate, 50 mg/L streptomycin
sulfate and 1.000 mg/L glucose.
- w/o calcium chloride and penicillin G
- Penicillin G supplied separately with each package for
addition at a final concentration of 1 million units/L.
Calcium chloride di-hydrate is supplied separately with
each package for final concentration of 133 mg/L.
- Cell culture tested
|
Isotonic saline
solution that contains energy sources and antibiotics.
Provides the option to
formulate with or without penicillin G or calcium.
Frequently used with
primary cells where risk of contamination may be high and where tissue
dissociation is an objective.
|
|
back to top |
|
|
| Product
# |
Product |
Description |
Application |
|
E 2888 |
Earle's Balanced Salt
Solution |
- Liquid
- With sodium bicarbonate
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Complete
and ready to use formulation of EBSS. |
|
E 7510 |
Earle's Balanced Salt
Solution (10x) |
- Liquid
- Without sodium bicarbonate; supplement with 2.2 gm/L sodium bicarbonate at 1x
- pH 4.0-4.4 at 10x
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Concentrated
version of E 2888. Must be diluted to 1x, supplemented with sodium
bicarbonate and pH adjusted before use.
|
|
E 6132 |
Earle's Balanced Salts |
- Powder
- Without sodium bicarbonate; supplement with 2.2 gm/L sodium bicarbonate
- Formulated at 8.7 grams of powder per liter of medium
- Cell culture tested
|
Powder
version of the complete EBSS formulation (E 2888).
Sodium bicarbonate is
added after the EBSS is dissolved in tissue culture grade water. The pH
is adjusted after addition of sodium bicarbonate.
|
|
E 3024 |
Earle's Balanced Salt
Solution |
- Liquid
- With sodium bicarbonate
- Without phenol red
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Same
as E 2888, except without phenol red.
Phenol red has been
shown to interfere with the growth of some cells at cloning densities.
Use this medium when
working with stem cells or when growing cells at low densities.
|
|
E 3261 |
Earle's Balanced Salt
Solution Modified |
- Powder
- Without phenol red and sodium bicarbonate; supplement with 2.2 gm/L sodium bicarbonate
- Formulated at 8.7 grams of powder per liter of medium
- Cell culture tested
|
Powder
version of E 3024.
Sodium bicarbonate is
added after the EBSS is dissolved in tissue culture grade water. The pH
is adjusted after addition of sodium bicarbonate.
Phenol red has been
shown to interfere with the growth of some cells at cloning densities.
Use this medium when
working with stem cells or when growing cells at low densities.
|
|
E 6267 |
Earle's Balanced Salt
Solution |
- Liquid
- With sodium bicarbonate
- Without calcium chloride, magnesium sulfate
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Same
as E 2888, except that calcium and magnesium have been removed.
Calcium and magnesium
support cell adhesion.
Recommended for
suspension cultures or whenever cell clumping is a problem.
|
|
E 2899 |
Earle's Balanced Salt Solution |
- Liquid
- With sodium bicarbonate
- Without calcium chloride, magnesium sulfate, and phenol red
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Same
as E 6267, except that phenol red has also been removed.
Phenol red has been
shown to interfere with the growth of some cells at cloning densities.
Use this medium when working with stem cells or when growing cells at low densities.
|
back to top |
|
|
| Product
# |
Product |
Description |
Application |
|
H 9269 |
Hanks'
Balanced Salt Solution |
- Liquid
- With sodium bicarbonate
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Complete
and ready to use formulation of HBSS.
|
|
H 1641 |
Hanks'
Balanced Salt Solution (10x) |
- Liquid
- Without sodium bicarbonate; supplement with 0.35 gm/L sodium bicarbonate at 1x
- pH 5.3-6.3 at 10x
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Concentrated
version of H 9269. Must be diluted to 1x, supplemented with sodium
bicarbonate and pH adjusted before use. |
|
H 6136 |
Hanks'
Balanced Salts |
- Powder
- Without sodium bicarbonate; supplement with 0.35 gm/L sodium bicarbonate
- Formulated at 9.8 grams of powder per liter of medium
- Cell culture tested
|
Powder
version of the complete HBSS formulation (H 9269).
Sodium bicarbonate is
added after the HBSS is dissolved in tissue culture grade water. The pH
is adjusted after addition of sodium bicarbonate.
Same
formulation as Hanks' Balanced Salts, Hybri-Max. (H 6393).
|
|
H 6393 |
Hanks'
Balanced Salts, Hybri-Max. |
- Powder
- Without sodium bicarbonate; supplement with 0.35 gm/L sodium bicarbonate
- Formulated at 9.8 grams of powder per liter of medium
- Endotoxin tested
- Hybridoma tested
|
Related
to H 6136. Specifically, tested for use with hybridoma cells.
Labeled
for in vitro diagnostic use.
|
|
H 8264 |
Hanks'
Balanced Salt Solution Modified |
- Liquid
- With sodium bicarbonate
- Without phenol red
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Same
as H 9269, except without phenol red.
Phenol red has been
shown to interfere with the growth of some cells at cloning densities.
Use this medium when
working with stem cells or when growing cells at low densities.
|
|
H 1387 |
Hanks'
Balanced Salts Modified |
- Powder
- Without phenol red and sodium bicarbonate; supplement with 0.35 gm/L sodium bicarbonate
- Formulated at 9.8 grams of powder per liter of medium
- Cell culture tested
|
Powder
version of H 8264.
Sodium bicarbonate is
added after the HBSS is dissolved in tissue culture grade water. The pH
is adjusted after addition of sodium bicarbonate.
Phenol red has been
shown to interfere with the growth of some cells at cloning densities.
Use this medium when
working with stem cells or when growing cells at low densities.
|
|
H 9394 |
Hanks'
Balanced Salt Solution Modified |
- Liquid
- With sodium bicarbonate
- Without calcium chloride, magnesium sulfate,
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Same
as H 6649, except that calcium and magnesium have been removed.
Calcium and magnesium
support cell adhesion.
Recommended for
suspension cultures or whenever cell clumping is a problem.
|
|
H 4641 |
Hanks'
Balanced Salt Solution (10x) |
- Liquid
- Without calcium chloride, magnesium sulfate, and sodium bicarbonate; supplement with 0.35 gm/L sodium bicarbonate
- pH 5.4-6.4 at 10x
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Concentrated
version of H 9394. Must be diluted to 1x, supplemented with sodium
bicarbonate and pH adjusted before use.
Calcium and magnesium
support cell adhesion.
Recommended for
suspension cultures or whenever cell clumping is a problem.
|
|
H 2387 |
Hanks'
Balanced Salts Modified |
- Powder
- Without calcium chloride, magnesium sulfate, and sodium bicarbonate; supplement with 0.35 gm/L sodium bicarbonate
- Formulated at 9.5 grams of powder per liter of medium
- Cell culture tested
|
Powder
version of H 9394.
Sodium bicarbonate is
added after the HBSS is dissolved in tissue culture grade water. The pH
is adjusted after addition of sodium bicarbonate.
Calcium and magnesium
support cell adhesion.
Recommended for
suspension cultures or whenever cell clumping is a problem.
|
|
H 6648 |
Hanks'
Balanced Salt Solution Modified |
- Liquid
- With sodium bicarbonate
- Without calcium chloride, magnesium sulfate, and phenol red
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Same
as H 9394, except that phenol red has been removed.
Calcium and magnesium
support cell adhesion. Phenol red has been shown to interfere with the growth of some cells at cloning densities.
Use this medium when
working with stem cells or when growing cells at low densities. Use to
minimize cell attachment or aggregation.
|
|
H 4891 |
Hanks'
Balanced Salts Modified |
- Powder
- Without calcium chloride, magnesium sulfate, phenol red and sodium bicarbonate; supplement with 0.35 gm/L sodium bicarbonate
- Formulated at 9.5 grams of powder per liter of medium
- Cell culture tested
|
Powder
version of H 6648.
Sodium bicarbonate is
added after the HBSS is dissolved in tissue culture grade water. The pH
is adjusted after addition of sodium bicarbonate.
Calcium and magnesium
support cell adhesion.
Phenol red has been
shown to interfere with the growth of some cells at cloning densities.
Use this medium when
working with stem cells or when growing cells at low densities. Use to
minimize cell attachment or aggregation.
|
back to top |
| Other Salt Mixtures
The following salts
solutions are available for use in specific cell culture or related
applications. Generally, the use of one of these formulations for an
application is specified in publications or technical protocols.
|
|
| Product # |
Product |
Description |
Application |
|
A 3551 |
Alsever's Solution |
- Liquid
- Sterile-filtered
- Endotoxin Tested
- Cell culture tested
|
Essentially
a solution containing citrate, sodium chloride and glucose. Provided
without bicarbonate, which may be added.
Hypotonic solution for
lysing cells. Especially, red blood cells.
|
|
G 9779 |
Gey's Balanced Salt Solution |
- Liquid
- Sterile-filtered
- Endotoxin Tested
- Cell culture tested
|
A
complete balanced salt solution with a unique combination of "salt
forms". Contains the same five basic ions as EBSS, HBSS and D-PBS. |
|
K 3753 |
Kreb's-Henseleit Buffer Modified |
- Powder
- With 2000 mg/L glucose
- Without calcium chloride and sodium bicarbonate; supplement with 2.1 gm/L of sodium bicarbonate;
- Formulated at 9.6 grams of powder per liter of medium
- Cell culture tested
|
Contains
the same components as K 4002. However, these ion components are at different
concentrations and in different salt forms. (see the formulation for
details). |
|
K 4002 |
Kreb's-Ringer Bicarbonate Buffer |
- Powder
- With 1800 mg/L glucose
- Without calcium chloride and sodium bicarbonate; supplement with 1.26 gm/L sodium bicarbonate
- Formulated at 9.5 grams of powder per liter of medium
- Cell culture tested
|
Bicarbonate
buffered isotonic saline containing magnesium, potassium, sodium and phosphate
ions.
|
|
P 2917 |
Puck's Saline |
- Liquid
- With sodium bicarbonate
- Without calcium chloride, magnesium sulfate, potassium phosphate and sodium phosphate
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
A very simple isotonic salt
solution. Contains sodium and potassium chloride, glucose and low concentration of bicarbonate
buffer (0.35 gm/L)
For in vitro diagnostic use
|