CLASSIC MEDIA AND SALTS
Cell Culture
Dulbecco's Media and Salts Selection Guide
Dulbecco's
Phosphate Buffered Saline (D-PBS)
The use of balanced salt solution
(BSS) in cell and tissue culture is generally attributed to early workers in the
field. In 1885, Sydney Ringer developed a solution of inorganic salts designed
to maintain contractility of mammalian heart tissue. A less specific salt solution
was designed by Tyrode for use in work with primary mammalian cells. Tyrode's
salt solution became the accepted fluid for diluting protein components of media
of natural origin. Since that time, many other balanced salt solutions have been
developed for use in cell and tissue culture. The current role of balanced salt
solution in cell culture is multi-faceted and can be divided into four principle
functions.
- As an irrigating, transporting and diluting fluid while maintaining intra- and extra-cellular osmotic balance;
- Provides cells with water and certain bulk inorganic ions essential for normal cell metabolism;
- Combined with a carbohydrate, such as glucose, provides the principle energy source for cell metabolism; and
- Provides buffering system to maintain the medium within the physiological pH range (7.2-7.6)
| Product # |
Product Name |
Description |
Application |
|
D 8662 |
Dulbecco's
Phosphate Buffered Saline |
- Liquid
- With calcium chloride and magnesium chloride
- Cell culture tested
- Sterile-filtered
- Endotoxin teste
|
Known
as D-PBS, this is a general use, isotonic saline solution for washing cells and
tissues. |
|
D 1283 |
Dulbecco's Phosphate Buffered Saline (10X) |
- Liquid
- With calcium chloride and magnesium chloride
- Cell culture tested
- Sterile-filtered
- pH 4.2-4.6 at 10X
- Endotoxin tested
|
Concentrated form of D 8662. |
|
D 8537 |
Dulbecco's
Phosphate Buffered Saline Modified |
- Liquid
- w/o calcium chloride and magnesium chloride
- Cell culture tested
- Sterile-filtered
- Endotoxin tested
|
Calcium and magnesium promote cell adhesion.
Use this formulation of D-PBS when tissue
or cell dissociation or release is the objective. |
|
D 1408 |
Dulbecco's Phosphate Buffered Saline (10X) |
- Liquid
- Cell culture tested
- Sterile-filtered
- pH 6.6-7.0 at 10X
- Endotoxin tested
|
Concentrated form of D 8537. |
|
D 5652 |
Dulbecco's Phosphate Buffered Saline Modified |
- Powder
- w/o calcium chloride and magnesium chloride
- Cell culture tested
- Formulated at 9.6 grams of powder per liter of medium
- Related products: Dulbecco's Phosphate Buffered Saline Hybri-Max (D7030)
|
Powder form of D 8537
Calcium and magnesium promote cell adhesion.
Use this formulation of D-PBS when tissue or cell dissociation or release is the objective.
|
| D
7030 |
Dulbecco's
Phosphate Buffered Saline Modified Hybri-Max. |
- Powder
- w/o calcium chloride and magnesium chloride
- Hybridoma tested
- Formulated at 9.6 grams of powder per liter of medium
- Endotoxin tested
|
Same
as D 5652 except it is tested for use with hybridoma cells. |
|
D 5773 |
Dulbecco's Phosphate Buffered Saline |
- Powder
- w/o calcium chloride
- Cell culture tested
- Formulated at 9.7 grams of powder per liter of medium
|
Calcium
promotes cell adhesion.
Use this formulation of D-PBS when tissue or cell dissociation or release is the objective.
Also, suitable for studies that involve calcium metabolism.
|
|
D 4031 |
Dulbecco's Phosphate Buffered Saline Modified |
- Liquid
- With 36 mg/L sodium pyruvate, 50 mg/L streptomycin sulfate, 100 mg/L kanamycin monosulfate, 1000 mg/L glucose, calcium chloride, and magnesium chloride.
- Sterile-filtered
- Endotoxin tested
|
Fully
supplemented isotonic solution.
Contains energy sources and antibiotics.
Frequently used with primary cells where risk of contamination may be high.
|
|
D 6650 |
Dulbecco's Phosphate Buffered Saline Modified |
- Powder
- With 36 mg/L sodium pyruvate, 50 mg/L streptomycin sulfate and 1.000 mg/L glucose.
- w/o calcium chloride and penicillin G
- Cell culture tested
- Penicillin G supplied separately with each package for addition at a final concentration of 1 million units/L.
Calcium chloride di-hydrate is supplied separately with each package for final concentration of 133 mg/L.
|
Isotonic saline solution that contains energy sources and antibiotics.
Provides the option to formulate with or without penicillin G or calcium.
Frequently used with primary cells where risk of contamination may be high and where tissue dissociation
is an objective.
|
back to top
Dulbecco's Modified Eagle's Medium (DME)
Many modifications of Eagle's Medium have been developed since the original formulation appeared in the literature.
Among the most widely used of these modifications is Dulbecco's Modified Eagle's medium (DME). DME is a modification of Basal Medium Eagle (BME) that contains
a four-fold higher concentration of amino acids and vitamins, as well as additional supplementary components. The original DME formula contains 1000 mg/L of glucose
and was first reported for culturing embryonic mouse cells. A further alteration with 4500 mg/L glucose has proved to be optimal for cultivation of certain cell
types.
| Product # |
Product |
Description |
Application |
|
D 5546 |
Dulbecco's Modified Eagle's Medium |
- Liquid
- With 1000 mg/L glucose and sodium bicarbonate
- Without L-glutamine
- With pyridoxine (substitutes pyridoxine HCl for pyridoxal HCl)
- Cell culture tested
- Endotoxin tested
- Sterile-filtered
|
Liquid
form of the original Dulbecco's formulation, offered as D 5523, with one key exception.
Pyridoxal, which is an aldehyde, has been replaced with pyridoxine. This
modification has been shown to improve the stability of the medium. |
|
D 6046 |
Dulbecco's Modified Eagle's Medium |
- Liquid
- With 1000 mg/L glucose, L-glutamine and sodium bicarbonate
- With pyridoxine (substitutes pyridoxine HCl for pyridoxal HCl)
- Cell culture tested
- Endotoxin tested
- Sterile-filtered
|
Fully
supplemented liquid form of the original Dulbecco's formulation, offered as D 5523, with one key exception. Pyridoxal, which is an aldehyde, has been
replaced with pyridoxine. This modification has been shown to improve the stability of the medium. |
|
D 2429 |
Dulbecco's Modified Eagle's Medium (10X) |
- Liquid
- With 1000 mg/L glucose at 1x
- With pyridoxine (substitutes pyridoxine HCl for pyridoxal HCl)
- Without L-glutamine, supplement with 0.584 gm/L L-glutamine, at 1x
- Without sodium bicarbonate, supplement with 3.7 gm/L sodium bicarbonate, at 1x
- Without folic acid, supplement with 0.004 gm/L folic acid, at 1x
- pH 1.8-2.0 at 10x
- Cell culture tested
- Endotoxin tested
- Sterile-filtered
|
Concentrated liquid version of the original low glucose Dulbecco's formulation, with
the pyridoxine substitution (see D 5546).
Because of its low solubility, folic acid is added after the medium is diluted to 1x and the pH has been adjusted.
|
|
D 5523 |
Dulbecco's Modified Eagle's Medium |
- Powder
- With L-glutamine and 1000 mg/L glucose
- Without sodium bicarbonate, supplement with 3.7 gm/L sodium bicarbonate
- Formulated at 10.0 grams of powder per liter of medium
- Cell culture tested
|
Provides investigator with the option to modify the buffering system. Also, this is Dulbecco's
original low glucose formulation. |
|
D 5921 |
Dulbecco's Modified Eagle's Medium Modified |
- Liquid
- With 1000 mg/L glucose, and sodium bicarbonate
- Without phenol red and L-glutamine, supplement with 0.584 gm/L L-glutamine
- With pyridoxine (substitutes pyridoxine HCl for pyridoxal HCl)
- Cell culture tested
- Endotoxin tested
- Sterile-filtered
|
Liquid form of the original Dulbecco's formulation, offered as D 5523, with one key exception.
Pyridoxal, which is an aldehyde, has been replaced with pyridoxine. This modification has been shown to improve the stability of the medium.
See D 2902 for note on phenol red
|
|
D 2902 |
Dulbecco's Modified Eagle's Medium Modified |
- Powder
- With L-glutamine and 1000 mg/L glucose
- Without sodium bicarbonate, supplement with 3.7 gm/L of sodium bicarbonate
- Without phenol red
- Formulated at 10.0 grams of powder per liter of medium
- Cell culture tested
|
Investigators who work with stem cells or cell clones frequently prefer media without phenol
red. |
|
D 5671 |
Dulbecco's Modified Eagle's Medium |
- Liquid
- With 4500 mg/L glucose and sodium bicarbonate
- Without L-glutamine, supplement with 0.584 gm/L L-glutamine
- Cell culture tested
- Hybridoma tested
- Endotoxin tested
- Sterile-filtered
|
Complete
liquid form of the high glucose modification of Dulbecco's original formulation.
Fully tested.
|
|
D 5796 |
Dulbecco's Modified Eagle's Medium |
- Liquid
- With 4500 mg/L glucose, L-glutamine and sodium bicarbonate
- With pyridoxine (substitutes pyridoxine HCl for pyridoxal HCl)
- Cell culture tested
- Endotoxin tested
- Sterile-filtered
|
Complete
liquid form of the high glucose version of Dulbecco's original formulation, with
one key exception. Pyridoxal, which is an aldehyde, has been replaced
with pyridoxine. This modification has been shown to improve the stability
of the medium. |
|
D 2554 |
Dulbecco's Modified Eagle's Medium (10X) |
- Liquid
- With 4500 mg/L glucose at 1x
- With pyridoxine (substitutes pyridoxine HCl for pyridoxal HCL)
- Without L-glutamine, supplement with 0.584 gm/L L-glutamine, at 1x
- Without sodium bicarbonate, supplement with 3.7 gm/L sodium bicarbonate, at 1x
- Without folic acid, supplement with 0.004 gm/L folic acid, at 1x
- pH 1.8-2.0 at 10x
- Cell culture tested
- Endotoxin tested
- Sterile-filtered
|
Concentrated
liquid version of the modified high glucose Dulbecco's formulation, with the pyridoxine
substitution (see D 5796).
Because of its low solubility, folic acid is added after the medium is diluted to 1x and the pH has been adjusted.
|
|
D 5648 |
Dulbecco's Modified Eagle's Medium |
- Powder
- With L-glutamine and 4500 mg/L glucose
- Without sodium bicarbonate, supplement with 3.7 gm/L of sodium bicarbonate
- Formulated at 13.4 grams of powder per liter of medium
- Cell culture tested
- Related to DME, Hybridoma tested (D 6655)
|
Modified
version of D 5523. Many cell lines have been found to require a higher glucose
level than was provided in the original formulation. |
|
D 6655 |
Dulbecco's Modified Eagle's Medium Hybri-Max. |
- Powder
- With L-glutamine and 4500 mg/L glucose
- Without sodium bicarbonate, supplement with 3.7 gm/L of sodium bicarbonate
- Formulated at 13.4 grams of powder per liter of medium
- Cell culture tested
- Hybridoma tested
- Endotoxin tested
|
Same as D 5648, except that it is tested with and recommended for use with hybridoma
cells. It is the powder version of D 5671. |
|
D 6546 |
Dulbecco's Modified Eagle's Medium |
- Liquid
- With 4500 mg/L glucose, 110 mg/L sodium pyruvate and sodium bicarbonate
- Without L-glutamine, supplement with 0.584 gm/L L-glutamine
- With pyridoxine (substitutes pyridoxine HCl for pyridoxal HCl)
- Cell culture tested
- Endotoxin tested
- Sterile-filtered
|
Pyruvate
supplemented, complete, liquid form of the high glucose version of Dulbecco's
original formulation, with one key exception. Pyridoxal, which is an aldehyde,
has been replaced with pyridoxine. This modification has been shown to improve
the stability of the medium.
Glutamine to be added by the investigator
|
|
D 6429 |
Dulbecco's Modified Eagle's Medium |
- Liquid
- With 4500 mg/L glucose, L-glutamine and sodium pyruvate
- With pyridoxine (substitutes pyridoxine HCl for pyridoxal HCl)
- Cell culture tested
- Endotoxin tested
- Sterile-filtered
|
Pyruvate
supplemented, complete, liquid form of the high glucose version of Dulbecco's original formulation, with one key exception. Pyridoxal, which is an aldehyde,
has been replaced with pyridoxine. This modification has been shown to improve the stability of the medium. |
|
D 7777 |
Dulbecco's Modified Eagle's Medium |
- Powder
- With L-glutamine, 4500 mg/L glucose and 110 mg/L sodium pyruvate
- Without sodium bicarbonate, supplement with 3.7 gm/L of sodium bicarbonate
- Formulated at 13.5 grams of powder per liter of medium
- Cell culture tested
- Related to DME, Hybridoma tested (D 6780)
- Cell culture tested
|
Same
as D 5648, except that it is supplemented with the additional energy source
pyruvate. |
|
D 6780 |
Dulbecco's Modified Eagle's Medium Hybri-Max. |
- Powder
- With L-glutamine, 4500 mg/L glucose and 110 mg/L sodium pyruvate
- Without sodium bicarbonate, supplement with 3.7 gm/L of sodium bicarbonate
- Formulated at 13.5 grams of powder per liter of medium
- Hybridoma tested
- Endotoxin tested
|
Same
as D 7777, except that it is tested with and recommended for use with hybridoma cells. |
|
D 6171 |
Dulbecco's Modified Eagle's Medium HEPES Modified |
- Liquid
- With 4500 mg/L glucose, 25 mM HEPES and sodium bicarbonate
- Without L-glutamine, supplement with 0.584 gm/L L-glutamine
- With pyridoxine (substitutes pyridoxine HCl for pyridoxal HCl)
- Cell culture tested
- Endotoxin tested
- Sterile-filtered
|
Pyruvate
supplemented, complete, liquid form of the high glucose version of Dulbecco's
original formulation, with one key exception. Pyridoxal, which is an aldehyde,
has been replaced with pyridoxine. This modification has been shown to improve
the stability of the medium.
See D 1152 for discussion of HEPES.
|
|
D 1152 |
Dulbecco's Modified Eagle's Medium HEPES Modification |
- Powder
- With L-glutamine and 4500 mg/L glucose and 25 mM HEPES
- Without sodium bicarbonate, supplement with 3.7 gm/L of sodium bicarbonate
- Formulated at 17.4 grams of powder per liter of medium
- Cell culture tested
|
Same
as D 5648, except that it is supplemented with HEPES. HEPES supplementation increases the buffering capacity of the medium. Recommended for cells that
require a higher pH (7.3-7.6) for optimal growth. |
|
D 0422 |
Dulbecco's Modified Eagle's Medium |
- Liquid
- With 4500 mg/L glucose, and sodium bicarbonate
- Without L-methionine, L-cystine and L-glutamine, supplement with 0.584 gm/L L-glutamine
- Cell culture tested
- Endotoxin tested
- Sterile-filtered
|
Deficient
liquid form of the high glucose version of Dulbecco's original formulation.
Key amino acids have been left out to allow researchers to set up metabolic studies
with tagged amino acids. |
|
D 3656 |
Dulbecco's Modified Eagle's Medium |
- Powder
- With L-glutamine and 4500 mg/L glucose
- Without sodium phosphate and sodium bicarbonate; supplement with 3.7 gm/L of sodium bicarbonate, 0.109 gm/L sodium phosphate mono-basic (anhydrous)
- Formulated at 13.3 grams of powder per liter of medium
- Cell culture tested
|
Provides
the investigator with the option and flexibility to control, modify or develop the medium buffering system. |
|
D 5280 |
Dulbecco's Modified Eagle's Medium Auto-Mod™ |
- Powder
- Modified for autoclaving
- With 1000 mg/L glucose
- Without L-glutamine and sodium bicarbonate; supplement with 0.584 gm/L of L-glutamine; 3.7 gm/L
of sodium bicarbonate
- Formulated at 9.6 grams of powder per liter of medium
- Cell culture tested
|
Same
as D 5523, except that L-glutamine and sodium bicarbonate have been removed so that this medium can be formulated and autoclaved. After autoclaving, the L-glutamine
and sodium bicarbonate can be added. |
|
D 5030 |
Dulbecco's Modified Eagle's Medium Base |
- Powder
- Without L-glutamine, glucose, phenol red, sodium pyruvate and sodium bicarbonate
- Supplement with 0.584 gm/L of L-glutamine; 1.0 gm/L glucose; 3.7 gm/L of sodium bicarbonate
- Formulated at 8.3 grams of powder per liter of medium
- Cell culture tested
|
The
most basic formulation offered. This formulation is used by investigators
who want to start with the essential components of DME, and have the flexibility
to optimize the formula for their own application. |
back to top
DME/Nutrient
Mixture F-12 Hams (DME/F-12)
During the past decade, researchers have reported the culture of a variety of cell lines in serum-free medium that
contained, instead of serum, a supplement of nutrients, growth factors and hormones. For example, Mather and Sato reported the successful cultivation of Leydig cells
and Sertoli cells in serum-free medium that contained insulin, transferrin,epidermal growth factor, leutinizing hormone or follicle stimulating hormone,
somatomedin and growth hormone. Although the hormones and their concentrations are specific for the type of cell under study, the medium found to be most satisfactory
for studies of this type was a 1:1 mixture of Dulbecco's Modified Eagle's Medium
(DME) and Ham's F-12 Nutrient Mixture. HEPES buffer is included in the formulation at a final concentration of 15 mM to compensate for the loss of buffering capacity incurred by eliminating serum.
| Product # |
Product |
Description |
Application |
|
D 6421 |
Dulbecco's Modified Eagle's Medium/Nutrient F-12 Ham |
- Liquid
- With 15 mM HEPES and sodium bicarbonate
- With pyridoxine (substitutes pyridoxine HCl for pyridoxal HCl)
- Without L-glutamine, supplement with 0.365 gm/L L-glutamine
- Endotoxin tested
- Cell culture tested
- Sterile-filtered
|
Liquid
form of original 1:1 mixture of DMEM/Ham's F-12, with one important exception. Pyridoxal, which is an aldehyde, has been replaced with pyridoxine. This
modification has been shown to improve the stability of the medium.
Suitable for growth of wide range of cell types in low serum conditions.
|
|
D 8437 |
Dulbecco's Modified Eagle's Medium/Nutrient F-12 Ham |
- Liquid
- With 15 mM HEPES, L-glutamine and sodium bicarbonate
- With pyridoxine (substitutes pyridoxine HCl for pyridoxal HCl)
- Endotoxin tested
- Cell culture tested
- Sterile-filtered
|
Same
as D 6421, except the L-glutamine is added to the medium at time of manufacture.
|
|
D 8900 |
Dulbecco's Modified Eagle's Medium/Nutrient F-12 Ham |
- Powder
- With L-glutamine and 15 mM HEPES
- w/o sodium bicarbonate, to be supplemented with 1.2 gm/L of sodium bicarbonate
- Formulated at 15.6 grams of powder per liter of medium
- Cell culture tested
|
Powder
form of original 1:1 mixture of DME/Ham's F-12. Suitable for growth of a wide range of cell types in low serum conditions. |
|
D 6905 |
Dulbecco's Modified Eagle's Medium/Nutrient F-12 Ham Hybi-Max. |
- Powder
- With L-glutamine and 15 mM HEPES
- w/o sodium bicarbonate, to be supplemented with 1.2 gm/L of sodium bicarbonate
- Formulated at 15.6 grams of powder per liter of medium
- Cell culture tested
|
Formulation
is identical to D 8900. This formulation has been tested with and is recommended
for use with hybridoma cells. |
|
D 6434 |
Dulbecco's Modified Eagle's Medium/Nutrient F-12 Ham |
- Liquid
- With 15 mM HEPES and sodium bicarbonate
- Without phenol red and L-glutamine, supplement with 0.365 gm/L L-glutamine
- With pyridoxine (substitutes pyridoxine HCl for pyridoxal HCl)
- Endotoxin tested
- Sterile-filtered
|
Pyridoxal, which is an aldehyde, has been replaced with pyridoxine. This modification has been
shown to improve the stability of the medium.
Suitable for growth of a wide range of cell types in low serum conditions. Absence of phenol red makes
this a preferred medium for clonal and stem cell culture.
|
|
D 2906 |
Dulbecco's Modified Eagle's Medium/Nutrient F-12 Ham |
- Powder
- With L-glutamine and 15 mM HEPES
- Without phenol red and sodium bicarbonate, to be supplemented with 1.2 gm/L of sodium bicarbonate
- Formulated at 15.6 grams of powder per liter of medium
- Cell culture tested
|
Suitable for growth of a wide range of cell types in low serum conditions.
The removal of phenol red is often preferred by investigators working with clonal and stem cell cultures.
|
|
D 6559 |
Dulbecco's Modified Eagle's Medium/Nutrient F-12 Ham |
- Liquid
- With sodium bicarbonate
- Without phenol red and L-glutamine, supplement with 0.365 gm/L L-glutamine
- With pyridoxine (substitutes pyridoxine HCl for pyridoxal HCl)
- Endotoxin tested
- Cell culture tested
- Sterile-filtered
|
Pyridoxal, which is an aldehyde, has been replaced with pyridoxine. This modification has been shown to improve the stability of the medium.
Suitable for growth of a wide range of cell types in low serum conditions. The absence of phenol red
makes this a preferred medium for clonal and stem cell culture.
The absence of HEPES allows the investigator to optimize the buffering system.
|
|
D 8062 |
Dulbecco's Modified Eagle's Medium/Nutrient F-12 Ham |
- Liquid
- With L-glutamine and sodium bicarbonate
- Without HEPES
- With pyridoxine (substitutes pyridoxine HCl for pyridoxal HCl)
- Endotoxin tested
- Cell culture tested
- Sterile-filtered
|
Same
as D 8437, except that HEPES has not been added to the formulation.
Allows the investigator to optimize the buffering system.
|
|
D 0547 |
Dulbecco's Modified Eagle's Medium/Nutrient F-12 Ham |
- Powder
- With L-glutamine and trace elements
- Without HEPES and sodium bicarbonate, to be supplemented with 1.2 gm/L of sodium bicarbonate
- Formulated at 12.1 grams of powder per liter of medium
- Cell culture tested
|
Powder
form of original 1:1 mixture of DME/Ham's F-12, supplemented with trace elements.
Trace element supplementation of the original formula (D 8900) is required for the completely serum free culture of a wide range of cell types.
|
|
D 9785 |
Dulbecco's Modified Eagle's Medium/Nutrient F-12 Ham Base |
- Powder
- With 15 mM HEPES
- Without L-glutamine, L-leucine, L-lysine, L-methionine, calcium chloride, magnesium chloride, magnesium sulfate, phenol red and sodium bicarbonate
- Supplement with 0.1545 gm/L calcium chloride; 0.365 gm/L L-glutamine; 0.05905 gm/L L-leucine, 0.09125 gm/L L-lysine; 0.0612 gm/L magnesium chloride, 0.04884 gm/L magnesium sulfate, 0.0172 gm/L L-methionine, and 1.2 gm/L
sodium bicarbonate
- Formulated at 14.8 grams of powder per liter of medium
- Cell culture tested
|
The
most basic formulation offered. This formulation is used by investigators who want to start with the essential components of (1:1) DME/Ham's F-12, and have
the flexibility to optimize the formula for their own application. This base formulation is particularly suitable for studies of cell protein metabolism
with label-tagged amino acids. |