Product Lines
Antibody Expression
Classic Media & Salts
Coated Plates
ECACC Cell Lines
Embryo Culture
Gene Therapy
Primary Human Culture
Protein Expression/CHO
Reagents & Supplements
Sera
Serum Free Media
Stem Cells
SyntheChol NS0
TrypZean Solution
Vaccine Production
Product Highlights
Key Resources
Cell Signaling and Neuroscience
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
CLASSIC MEDIA AND SALTS
Cell Culture
Basal Salt Mixtures :: Dulbecco's Media :: Ham's Nutrient Mixtures :: MCDB Media
MEM Media :: Medium 199 :: MegaCell™ Media :: Other Classic Media :: RPMI Media
Ham's Nutrient Mixtures
Ham's Nutrient Mixtures were originally developed to support growth of several clones of Chinese hamster ovary (CHO) cells, as well as clones of HeLa and mouse L-cells.  Both mixtures were formulated for use with or without serum supplementation, depending on the cell type being cultured.
  • Ham's F-10 has been shown to support the growth of human diploid cells, white blood cells for chromosomal analysis, primary explants of rat, rabbit and chicken tissues.
  • Ham's F-12  has been used for the growth of primary rat hepatocytes and rat prostate epithelial cells.  A clonal toxicity assay using CHO cells has also been reported with Ham's F-12 as the medium of choice.  Ham's F-12 is also available with 25 mM HEPES buffer that provides more effective buffering in the optimum pH range of 7.2-7.4.
  • Coon's modification of Ham's F-12 was developed for culturing hybrid cells that were produced by viral fusion.  The modification consists of doubling the amino acids and pyruvate and including ascorbic acid.  The salt concentrations have also been altered.  The formula contains 0.863 mg/L zinc sulfate, which may render it unsuitable for culturing mouse L-cells.
  • Kaighn's modification of Ham's F-12 and Coon's F-12 has increased concentrations of amino acids and pyruvate, as well as modified salts (Konigsbergs).  This medium was designed to support the growth of differentiated rat and chicken cells, and primary human liver cells.
Product # Product Name Description Application
N 6013
Nutrient Mixture F-10 Ham (Ham's F-10)
  • Liquid
  • With sodium bicarbonate
  • Without L-glutamine; supplement with 0.146 gm/L L-glutamine
  • Sterile-filtered
  • Endotoxin tested
  • Cell culture tested
Supports the growth of human diploid cells, white blood cells for chromosomal analysis, primary explants of rat, rabbit and chicken tissues.
Add L-glutamine to formulation at time of use.
N 6908
Nutrient Mixture F-10 Ham  (Ham's F-10)
  • Liquid
  • With L-glutamine and sodium bicarbonate
  • Sterile-filtered
  • Endotoxin tested
  • Cell culture tested
Complete, fully supplemented version of N 6013.
N 6635
Nutrient Mixture F-10 Ham (Ham's F-10)
  • Powder
  • With L-glutamine
  • Without sodium bicarbonate; supplement with 1.2 gm/L sodium bicarbonate;
  • Formulated at 9.8 grams of powder per liter of medium;
  • Cell culture tested
Powder version of N 6908.
Sodium bicarbonate is added at the time that the liquid medium is formulated.
N 2147
Nutrient Mixture F-10 Ham (Ham's F-10)
  • Liquid
  • With 20 mM HEPES
  • Without L-glutamine and sodium bicarbonate: supplement with 0.146 gm/L L-glutamine;
  • Sterile-filtered
  • Endotoxin tested
  • Cell culture tested
Contains HEPES and no bicarbonate.
HEPES increases the buffer strength of the medium and raises the maximum buffering range.
Recommended for cells that grow better at a pH above 7.2.
N 1387
Nutrient Mixture F-10 Ham HEPES Modification (Ham's F-10)
  • Powder
  • With L-glutamine and 25 mM HEPES;
  • Without sodium bicarbonate; supplement with 1.2 gm/L sodium bicarbonate
  • Formulated at 15.2 grams of powder per liter of medium;
  • Cell culture tested
Contains both HEPES and bicarbonate.
HEPES increases the buffer strength of the medium and raises the maximum buffering range.
Recommended for cells that grow better at a pH above 7.2.
N 4888
Nutrient Mixture F-12 (Ham's F-12)
  • Liquid
  • With sodium bicarbonate
  • Without L-glutamine; supplement with 0.146 gm/L L-glutamine
  • Sterile-filtered
  • Endotoxin tested
  • Cell culture tested
Developed for growth of primary rat hepatocytes and rat prostate epithelial cells.  Also used in a clonal toxicity assay using CHO cells.
Sodium bicarbonate is added at the time of formulation.
N 6658
Nutrient Mixture F-12 (Ham's F-12)
  • Liquid
  • With L-glutamine and sodium bicarbonate
  • Sterile-filtered
  • Endotoxin tested
  • Cell culture tested
Complete, fully supplemented version of N 4888.
Developed for growth of primary rat hepatocytes and rat prostate epithelial cells.  Also used in a clonal toxicity assay using CHO cells.
N 6760
Nutrient Mixture F-12 Ham;(Ham's F-12)
  • Powder
  • With L-glutamine
  • Without sodium bicarbonate; supplement with 1.176 gm/L sodium bicarbonate;
  • Formulated at 10.7 grams of powder per liter of medium;
  • Cell culture tested
Powder version of N 6658.
Developed for growth of primary rat hepatocytes and rat prostate epithelial cells.  Also used in a clonal toxicity assay using CHO cells.
N 8641
Nutrient Mixture F-12 Ham HEPES Modification (Ham's F-12)
  • Liquid
  • With sodium bicarbonate and 25 mM HEPES;
  • Without L-glutamine; supplement with 0.146 gm/L L-glutamine
  • Sterile-filtered
  • Endotoxin tested
  • Cell culture tested
Modified buffering system based upon HEPES.
HEPES increases the buffer strength of the medium and raises the maximum buffering range.
Recommended for cells that grow better at a pH above 7.2.
N 4388
Nutrient Mixture F-12 Ham HEPES Modification (Ham's F-12)
  • Powder
  • With L-glutamine and 25 mM HEPES;
  • Without sodium bicarbonate; supplement with 1.176 gm/L sodium bicarbonate;
  • Formulated at 16.2 grams of powder per liter of medium;
  • Cell culture tested
Powder version of N8641.
Supplement with sodium bicarbonate at time of formulation.
F 6636
Nutrient Mixture F-12 Ham F-12, Coon's Modification (Ham's F-12 Coon's)
  • Powder
  • With L-glutamine and 0.863 mg/L zinc sulfate;
  • Without sodium bicarbonate; supplement with 2.676 gm/L sodium bicarbonate;
  • Formulated at 11.6 grams of powder per liter of medium;
  • Cell culture tested
Coon's modification of Ham's F-12 was developed for culturing hybrid cells that were produced by viral fusion.  The modification consists of doubling the amino acids and pyruvate and including ascorbic acid.  The salt concentrations have also been altered.  The formula contains 0.863 mg/L zinc sulfate, which may render it unsuitable for culturing mouse L-cells.
N 3520
Nutrient Mixture F-12 Ham Kaighn's Modification (Ham's F-12K)
  • Powder
  • With L-glutamine
  • Without sodium bicarbonate; supplement with 2.5 gm/L sodium bicarbonate;
  • Formulated at 11.3 grams of powder per liter of medium;
  • Cell culture tested
Kaighn's modification of Ham's F-12 and Coon's F-12 has increased concentrations of amino acids and pyruvate, as well as modified salts (Konigsbergs).  This medium was designed to support the growth of differentiated rat and chicken cells, and primary human liver cells.
back to top