| Product # |
Product Name |
Description |
Application |
|
M6395 |
MCDB 105 Medium |
- Powder
- Complete with trace elements, L-glutamine and 25 mM HEPES;
- Formulated at 11.9 grams of powder per liter of medium;
- Cell culture tested
|
Originally developed for long term growth of diploid fibroblast-like cells, such as
WI-38, MRC-5 and IMR-90.
Recommended for
production of virus in adherent cell cultures.
HEPES increases the
buffer strength of the medium and raises the maximum buffering range.
Recommended for cells that grow better at a pH above 7.2.
|
|
M 6520 |
MCDB 110 Medium |
- Powder
- Complete with trace elements, L-glutamine and 25 mM HEPES;
- Formulated at 15.3 grams of powder per liter of medium;
- Cell culture tested
|
Originally developed for rapid of human diploid fibroblast-like cells.
Recommended for production of virus in adherent cell cultures.
HEPES increases the buffer strength of the medium and raises the maximum buffering range.
Recommended for cells that grow better at a pH above 7.2.
|
|
M 8537 |
MCDB 131 Medium |
- Powder
- Complete with trace elements and L-glutamine;
- Without sodium bicarbonate: supplement with 1.18 gm/L sodium bicarbonate;
- Formulated at 11.7 grams of powder per liter of medium;
- Cell culture tested
|
Originally developed for the clonal growth of human micro-vascular endothelial cells (HMVEC).
Buffered with bicarbonate instead of HEPES.
|
|
M 6645 |
MCDB 151 Medium |
- Powder
- Complete with trace elements, L-glutamine; and 28 mM HEPES;
- Without sodium bicarbonate: supplement with 1.176 gm/L sodium bicarbonate;
- Formulated at 17.7 grams of powder per liter of medium;
- Cell culture tested
|
Originally developed for the growth of primary human keratinocytes.
HEPES increases the buffer strength of the medium and raises the maximum buffering range.
Recommended for cells that grow better at a pH above 7.2.
|
|
M 7403 |
MCDB 153 Medium |
- Powder
- Complete with trace elements, L-glutamine; and 28 mM HEPES;
- Without sodium bicarbonate: supplement with 1.176 gm/L sodium bicarbonate;
- Formulated at 17.7 grams of powder per liter of medium;
- Cell culture tested
|
Originally developed for clonal and long term growth of human epidermal keratinocytes
under serum free conditions.
M 7403 is more
extensively supplemented than M 6645, especially with trace elements.
HEPES increases the
buffer strength of the medium and raises the maximum buffering range.
Recommended for cells that grow better at a pH above 7.2.
|
|
M 6770 |
MCDB 201 Medium |
- Powder
- Complete with trace elements, L-glutamine; and 30 mM HEPES;
- Formulated at 17.7 grams of powder per liter of medium;
- Cell culture tested
|
Originally developed for clonal growth of chicken embryo fibroblasts (CEF). CEF
cells are used for virus production.
HEPES increases the
buffer strength of the medium and raises the maximum buffering range.
Recommended for cells that grow better at a pH above 7.2.
|
|
M 2021 |
MCDB 302 Medium |
- Powder
- Complete with trace elements and L-glutamine;
- Without nucleosides and sodium bicarbonate: supplement with 1.18 gm/L sodium bicarbonate;
- Formulated at 11.0 grams of powder per liter of medium;
- Cell culture tested
|
Originally developed for clonal growth of Chinese hamster ovary (CHO) cells.
Buffered with bicarbonate instead of HEPES.