| Product # |
Product Name |
Description |
Application |
|
M 2279 |
Minimum Essential Medium Eagle |
- Liquid
- With Earle's salts and sodium bicarbonate
- Without L-glutamine, supplement with 0.292 gm/L of L-glutamine
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Essentially
the same as the original formula.
Generally used to grow
attached cell lines, such as fibroblasts, in the presence of FBS, calf or
horse sera. |
| M
4655 |
Minimum Essential Medium Eagle |
- Liquid
- With Earle's salts,
L-glutamine and sodium bicarbonate
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Essentially
the same as the original formula.
Generally used to grow
attached cell lines, such as fibroblasts, in the presence of FBS, calf or
horse sera.
|
| M
0275 |
Minimum Essential Medium Eagle
(10x) |
- Liquid
- With Earle's salts
- Without L-glutamine
and sodium bicarbonate
- Supplement with
0.292 gm/L of L-glutamine and 2.2 gm/L of sodium bicarbonate at 1x
- pH 5.1-5.5 at 10x
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Essentially
the same as the original formula.
Generally used to grow
attached cell lines, such as fibroblasts, in the presence of FBS, calf or
horse sera.
The L-glutamine and
sodium bicarbonate are added to the medium after dilution to 1x and pH
adjustment. |
| M
0268 |
Minimum Essential Medium Eagle |
- Powder
- With Earle's salts
and L-glutamine
- Without sodium
bicarbonate, supplement with 2.2 gm/L sodium bicarbonate
- Formulated at 9.6
grams of powder per liter of medium
- Cell culture tested
|
Essentially
the same as the original formula, except it is provided as a powder.
Generally used to grow
attached cell lines, such as fibroblasts, in the presence of FBS, calf or
horse sera.
|
| M
5650 |
Minimum Essential Medium Eagle |
- Liquid
- With Earle's salts,
non-essential amino acids and sodium bicarbonate
- Without L-glutamine,
supplement with 0.292 gm/L of L-glutamine
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Same as M 2279,
with one major difference. It contains 19 amino acids. The
essential amino acids and the non-essential amino acids: L-ala; L-asn; L-asp;
L-glu; L-gly; L-pro and L-ser.
May be preferred for
growth of adherent cells in the presence of reduced serum or as a base for
development of a serum free medium.
|
| M
0643 |
Minimum Essential Medium Eagle |
- Powder
- With Earle's salts,
L-glutamine and non-essential amino acids
- Without sodium bicarbonate, supplement with 2.2 gm/L sodium bicarbonate
- Formulated at 9.7 grams of powder per liter of medium
- Cell culture tested
|
Offered
as powder form of M 5650
It
contains 19 amino acids. The essential amino acids and the non-essential
amino acids: L-ala; L-asn; L-asp; L-glu; L-gly; L-pro and L-ser.
May be preferred for
growth of adherent cells in the presence of reduced serum or as a base for
development of a serum free medium. |
| M
3024 |
Minimum Essential Medium Eagle
Modified |
- Powder
- With Earle's salts and non-essential amino acids
- Without L-glutamine,
phenol red, and sodium bicarbonate.
- Supplement with 0.292 gm/L L-glutamine; with 2.2 gm/L of sodium bicarbonate
- Formulated at 9.4 grams of powder per liter of medium
- Cell culture tested
|
It contains 19 amino acids. The essential amino acids and the non-essential amino
acids: L-ala; L-asn; L-asp; L-glu; L-gly; L-pro and L-ser.
May be preferred for
growth of adherent cells in the presence of reduced serum or as a base for
development of a serum free medium.
Same as M 0643, except
that L-glutamine may be added at time of use.
|
| M
4144 |
Minimum Essential Medium Eagle
Modified |
- Powder
- With Earle's salts
- Without phenol red, L-glutamine and sodium bicarbonate.
- Supplement with 0.292 gm/L L-glutamine; with 2.2 gm/L of sodium bicarbonate
- Formulated at 9.3 grams of powder per liter of medium
- Cell culture tested
|
Essentially the
same as the original formula, except it is provided as a powder and
without phenol red.
Generally used to grow
attached cell lines, such as fibroblasts, in the presence of FBS, calf or
horse sera.
Media without
phenol red is often preferred for work with stem cells and in cell
cloning.
|
| M
7278 |
Minimum Essential Medium Eagle
HEPES Modification |
- Liquid
- With Earle's salts,
25 mM HEPES and sodium bicarbonate
- Without L-glutamine; supplement with 0.292 gm/L of L-glutamine
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Same as M 2279,
except that it is supplemented with 25 mM HEPES.
HEPES increases the
buffer strength of the medium and raises the maximum buffering range.
Recommended for cells
that grow better at a pH above 7.2.
|
| M
2645 |
Minimum Essential Medium Eagle
HEPES Modification |
- Powder
- With Earle's salts,
L-glutamine and HEPES
- Without sodium
bicarbonate; supplement with 2.2 gm/L sodium bicarbonate
- Formulated at 14.2
grams of powder per liter of medium
- Cell culture tested
|
Similar to M 7278,
except that it is a powder without sodium bicarbonate.
HEPES increases the
buffer strength of the medium and raises the maximum buffer range.
Leaving the bicarbonate out gives the investigator greater control over the
buffer system.
Recommended for cells
that grow better at a pH above 7.2.
|
| M
8042 |
Minimum Essential Medium Eagle
Alpha Modification |
- Liquid
- With ribonucleosides
and deoxyribonucleosides and sodium bicarbonate
- Without L-glutamine,
supplement with 0.292 gm/L of L-glutamine
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
This is the most
enriched variation of the MEM formulation offered. It contains all 21
normal amino acids, some at increased concentrations. In addition, it
contains 5 additional vitamins. See formulation for details.
Often used in
cytogenetic analysis when supplemented with FBS.
|
| M
4526 |
Minimum Essential Medium Eagle
Alpha Modification |
- Liquid
- With sodium
bicarbonate
- Without L-glutamine, supplement with 0.292 gm/L of L-glutamine
- Without
ribonucleosides and deoxyribonucleosides
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Same as M 8042,
except that it lacks the ribonucleosides and deoxyribonucleosides.
This is the most
enriched variation of the MEM formulation offered. It contains all 21
normal amino acids, some at increased concentrations. In addition, it contains
5 additional vitamins.
|
| M
0644 |
Minimum Essential Medium Eagle
Alpha Modification |
- Powder
- With L-glutamine, ribonucleosides and deoxyribonucleosides.
- Without sodium bicarbonate; supplement with 2.2 gm/L sodium bicarbonate
- Formulated at 10.2 grams of powder per liter of medium
- Cell culture tested
|
This is the most
enriched variation of the MEM formulation offered. It contains all 21
normal amino acids, some at increased concentrations. In addition, it contains
5 additional vitamins.
Often used in
cytogenetic analysis when supplemented with FBS.
Offered as a powder
form of M 8042.
|
| M
0894 |
Minimum Essential Medium Eagle
Alpha Modification |
- Powder
- With L-glutamine
- Without ribonucleosides, deoxyribonucleosides and sodium bicarbonate; supplement with 2.2 gm/L sodium bicarbonate
- Formulated at 10.1 grams of powder per liter of medium
- Cell culture tested
|
This is the most
enriched variation of the MEM formulation offered. It contains all 21
normal amino acids, some at increased concentrations. In addition, it contains
5 additional vitamins.
Offered as a powder
form of M 4526.
|
| M
2414 |
Minimum Essential Medium Eagle
Modified |
- Liquid
- With Earle's salts
and reduced sodium bicarbonate
- Without L-glutamine; supplement with 0.292 gm/L of L-glutamine
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Same as M 2279, except that the sodium bicarbonate concentration is reduced to
0.85 gm/L.
Recommended for use
in a carbon dioxide-free culture system. May also be supplemented with
HEPES to raise the buffering range.
|
| M
3786 |
Minimum Essential Medium Eagle |
- Liquid
- With Earle's salts,
sodium bicarbonate and L-glutamine
- Without sodium
phosphate; supplement with 0.122 gm/L of sodium phosphate monobasic
(anhydrous)
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Same as M 2279, except that sodium phosphate has been left out of the
formulation.
Recommended for
investigators who want to use a formulation that is totally buffered by
sodium bicarbonate.
|
| M
8028 |
Minimum Essential Medium Eagle
Joklik Modification |
- Liquid
- With sodium
bicarbonate
- Without calcium chloride and L-glutamine, supplement with 0.292 gm/L of L-glutamine
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Calcium and magnesium support cell adhesion. Calcium has been removed
from this formulation. However, it still contains magnesium chloride.
Recommended for suspension cultures.
|
| M
0518 |
Minimum Essential Medium Eagle
Joklik Modification |
- Powder
- With
L-glutamine
- Without calcium
chloride and sodium bicarbonate, supplement with 2.0 gm/L sodium
bicarbonate
- Formulated at 11.2
grams of powder per liter of medium
- Cell culture tested
|
Calcium and magnesium support cell adhesion. Calcium has been removed
from this formulation. However, it still contains magnesium chloride.
Recommended for
suspension cultures.
Offered as the powder
form of M 8028
|
| M
8167 |
Minimum Essential Medium Eagle
Spinner Modification
(S-MEM) |
- Liquid
- With Earle's salts and sodium bicarbonate
- Without calcium chloride and L-glutamine; supplement with 0.292 gm/L of L-glutamine
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Calcium and
magnesium support cell adhesion. Calcium has been removed from this
formulation. However, it still contains magnesium sulfate.
Recommended for suspension cultures.
|
| M
4767 |
Minimum Essential Medium Eagle
Spinner Modification (S-MEM) |
- Powder
- With L-glutamine
- Without calcium chloride and sodium bicarbonate
- Formulated at 10.4 grams of powder per liter of medium
- Cell culture tested
|
Calcium and magnesium support cell adhesion. Calcium has been removed
from this formulation. However, it still contains magnesium sulfate.
Recommended for suspension
cultures
Offered as the powder
form of M 8167
|
| M
7395 |
Minimum Essential Medium Eagle
D-Valine Modification |
- Powder
- With Earle's salts, L-glutamine and D-valine substituted for L-valine.
- Without sodium bicarbonate; supplement with 2.2 gm/L sodium bicarbonate.
- Formulated at 9.6 grams of powder per liter of medium
- Cell culture tested
|
Same
formulation as M 2279 except that D-valine has been substituted for L-valine.
Recommended for the
preferential selection and growth of cells possessing D-amino acid oxidase,
such as epithelial and Schwann cells.
|
|
M 7270 |
Minimum Essential Medium Eagle |
- Powder
- With Earle's salts and L-glutamine.
- Without L-leucine, L-lysine, L-methionine and sodium bicarbonate
- Supplement with 0.052 gm/L of L-leucine; 0.0725 gm/L of L-lysine; 0.015 gm/L of L-methionine
and 2.2 gm/L of sodium bicarbonate.
- Formulated at 9.4 grams of powder per liter of medium
- Cell culture tested
|
Same as M 0268, except that 3 essential amino acids are not included in the
powder.
This base formulation
is particularly suitable for studies of cell protein metabolism and
label-tagged amino acid studies.
Offered as the powder
form of M 2289
|
| M
3911 |
Minimum Essential Medium Eagle |
- Liquid
- With Earle's salts,
sodium bicarbonate and L-glutamine
- Without L-methionine; supplement with 0.015 gm/L L-methionine
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Same as M 4655,
except that L-methionine is not included.
Useful for studies of
methionine metabolism.
|
| M
2289 |
Minimum Essential Medium Eagle
Modified |
- Liquid
- With Earle's salts and sodium bicarbonate
- Without L-methionine, L-cystine, and L-glutamine
- Supplement with 0.313 gm/L of L-cystine dihydrochloride; 0.292 gm/L of L-glutamine and 0.015 gm/L of L-methionine.
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Same as M 2279,
except that 3 essential amino acids are not included.
This base formulation
is particularly suitable for studies of cell protein metabolism and
label-tagged amino acid studies.
|
| M
0769 |
Minimum Essential Medium Eagle
Auto-ModTM |
- Powder
- Modified for autoclaving
- With Earle's salts
- Without L-glutamine and sodium bicarbonate.
- Supplement with 0.292 gm/L L-glutamine; with 2.2 gm/L of sodium bicarbonate
- Formulated at 9.4 grams of powder per liter of medium
- Cell culture tested
|
Modification of the
original MEM formulation. Contains a succinate buffering system. See
formulations table for specific information.
Medium may be
autoclaved.
|
| M
5775 |
Minimum Essential Medium Eagle |
- Liquid
- With Hanks' salts and sodium bicarbonate
- Without L-glutamine, supplement with 0.292 gm/L of L-glutamine
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Modification of the
original MEM formulation. Earle's salts have been replaced with Hanks'
salts.
Since the bicarbonate
concentration is substantially lower, this medium has a substantially reduced
buffering capacity.
Same as M 4780, except
that the L-glutamine may be added at time of use.
|
| M
4780 |
Minimum Essential Medium Eagle |
- Liquid
- With Hanks' salts, L-glutamine and sodium bicarbonate
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Modification of the
original MEM formulation. Earle's salts have been replaced with Hanks'
salts.
Since the bicarbonate
concentration is substantially lower, this medium has a substantially reduced
buffering capacity.
|
| M
9288 |
Minimum Essential Medium Eagle
(10x) |
- Liquid
- With Hanks' salts
- Without L-glutamine
and sodium bicarbonate
- Supplement with 0.292 gm/L of L-glutamine and 0.35 gm/L of sodium bicarbonate at 1x
- pH 5.2-5.4 at 10x
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
This is the 10x
concentrated version of M 5775 and M 4780.
L-glutamine and sodium
bicarbonate are added after the medium has been diluted to 1x and the pH has
been adjusted.
|
| M
4642 |
Minimum Essential Medium Eagle |
- Powder
- With Hanks' salts and L-glutamine.
- Without sodium bicarbonate; supplement with 0.35 gm/L sodium bicarbonate
- Formulated at 10.7 grams of powder per liter of medium
- Cell culture tested
|
Modification of the
original MEM formulation. Earle's salts have been replaced with Hanks'
salts.
Since the bicarbonate
concentration is substantially lower, this medium has a substantially reduced
buffering capacity.
|
| M
3149 |
Minimum Essential Medium Eagle
Modified |
- Powder
- With Hanks' salts and non-essential amino acids
- Without L-glutamine, phenol red and sodium bicarbonate
- Supplement with 0.292 gm/L of L-glutamine
- Formulated at 10.5 grams of powder per liter of medium
- Cell culture tested
|
Modification of the
original MEM formulation. Earle's salts have been replaced with Hanks'
salts.
Since the bicarbonate
concentration is substantially lower, this medium has a substantially reduced
buffering capacity.
It contains 19 amino
acids. The essential amino acids and the non-essential amino acids:
L-ala; L-asn; L-asp; L-glu; L-gly; L-pro and L-ser.
May be preferred for
growth of adherent cells in the presence of reduced serum or as a base for development of a serum free medium.
|
| M 1018 |
Minimum Essential Medium Eagle |
- Powder
- With Hanks' salts, L-glutamine and non-essential amino acids
- Without sodium bicarbonate; supplement with 0.35 gm/L sodium bicarbonate
- Formulated at 10.8 grams of powder per liter of medium
- Cell culture tested
|
Modification of the
original MEM formulation. Earle's salts have been replaced with Hanks'
salts.
Since the bicarbonate concentration is substantially lower, this medium has a substantially reduced
buffering capacity.
It contains 19 amino acids. The essential amino acids and the non-essential amino acids:
L-ala; L-asn; L-asp; L-glu; L-gly; L-pro and L-ser.
May be preferred for growth of adherent cells in the presence of reduced serum or as a base for
development of a serum free medium.
|
Most formulations are offered in both a powder and a liquid form. In addition, most formulations are offered as a complete medium or in deficient variations that allow the user to supplement with common components such as bicarbonate, glutamine or
calcium. These variations provide more flexibility and control over the ultimate use and suitability of the medium.
Most of the formulations offered contain the original Earle's salts. However, a number of formulations contain Hanks' salts. Both of these phosphate and bicarbonate buffered saline solutions contain the same ions: calcium; magnesium; potassium; sodium,and phosphate. They differ in the concentrations of calcium,
sodium and in the buffer component. Hanks' buffered salts solution contains a substantially lower concentration of sodium bicarbonate than does
Earle's buffered salts.
The original MEM formulation contained Earle's salts and a group of amino acids generally referred to as essential amino acids. They were considered essential because the cells for which the medium was developed would not grow in their absence. The 12 essential amino acids are: L-arginine; L-cystine; L-glutamine; L-histidine; L-isoleucine; L-leucine; L-methionine; L-phenylalanine; L-threonine; L-tryptophan; L-tyrosine; and L-valine.