| Product # |
Product Name |
Description |
Application |
|
M 4530 |
Medium 199 |
- Liquid
- With Earle's salts, L-glutamine, and sodium bicarbonate;
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Complete
liquid version of the original M 199 formulation published by Morgan.
One of the early media
formulations used for virus production.
|
|
M 2154 |
Medium 199 |
- Liquid
- With Earle's salts and sodium bicarbonate;
- Without L-glutamine; supplement with 0.1 gm/L L-glutamine
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Same
as M 4530, except that L-glutamine must be added by the investigator.
|
|
M 0650 |
Medium 199 (10x) |
- Liquid
- With Earle's salts
- Without L-glutamine and sodium bicarbonate; supplement with 0.1 gm/L L-glutamine,
2.2 gm/L sodium bicarbonate
- pH 3.1-4.1 at 10x
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
This
is the concentrated version (10x) of M 4530.
L-glutamine and sodium
bicarbonate are added to the medium after dilution to 1x and pH adjustment.
|
|
M 5017 |
Medium 199 |
- Powder
- With Earle's salts and L-glutamine
- Without sodium bicarbonate: supplement with 2.2 gm/L sodium bicarbonate;
- Formulated at 9.9 grams of powder per liter of medium;
- Cell culture tested
|
Powder
version of M 4530.
Sodium bicarbonate is
added at the time that the liquid medium is formulated.
|
|
M 3769 |
Medium 199 Modified |
- Powder
- With Earle's salts
- Without phenol red, L-glutamine and sodium bicarbonate: supplement with
0.1 gm/L L-glutamine, 2.2 gm/L sodium bicarbonate;
- Formulated at 9.8 grams of powder per liter of medium;
- Cell culture tested
|
Related
to M 4530, but without phenol red.
Sodium bicarbonate and
L-glutamine are added at the time that the liquid medium is formulated.
Phenol red has been
shown to interfere with the growth of some cells at low or cloning densities.
Use this version of
Medium 199 when working with stem cells or cells at low densities.
|
|
M 7528 |
Medium 199 HEPES Modified |
- Liquid
- With Earle's salts, sodium bicarbonate and 25 mM HEPES;
- Without L-glutamine; supplement with 0.1 gm/L L-glutamine
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Same
as M 2154, and M 4530, except the buffer system is modified.
HEPES increases the
buffer strength of the medium and raises the maximum buffering range.
Recommended for cells
that grow better at a pH above 7.2.
|
|
M 2520 |
Medium 199 HEPES Modified |
- Powder
- With Earle's salts, L-glutamine and 25 mM HEPES;
- Without sodium bicarbonate: supplement with 2.2 gm/L sodium bicarbonate;
- Formulated at 15.1 grams of powder per liter of medium;
- Cell culture tested
|
Powder
version of M 7528.
HEPES increases the
buffer strength of the medium and raises the maximum buffering range.
Recommended for cells
that grow better at a pH above 7.2
|
|
M 7653 |
Medium 199 |
- Liquid
- With Hanks' salts and sodium bicarbonate;
- Without L-glutamine; supplement with 0.1 gm/L L-glutamine
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Modification
of the original Medium 199. Earle's salts are replaced with Hanks'
salts. Hanks' salts use a lower concentration of bicarbonate and
different concentrations and salt forms of the essential inorganic ions.
|
|
M 9163 |
Medium 199 (10x) |
- Liquid
- With Hanks' salts;
- Without L-glutamine and sodium bicarbonate; supplement with 0.1 gm/L L-glutamine,
0.35 gm/L sodium bicarbonate
- pH 4.2-4.4 at 10x
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
This
is the concentrated version (10x) of M 7653.
L-glutamine and sodium
bicarbonate are added to the medium after dilution to 1x and pH adjustment.
|
|
M 0393 |
Medium 199 |
- Powder
- With Hanks' salts and L-glutamine
- Without sodium bicarbonate: supplement with 0.35 gm/L sodium bicarbonate;
- Formulated at 11.0 grams of powder per liter of medium;
- Cell culture tested
|
Powder
version of M 7653.
Sodium bicarbonate is
added at the time that the liquid medium is formulated.
|
|
M 3274 |
Medium 199 Modified |
- Powder
- With Hanks' salts
- Without phenol red, L-glutamine and sodium bicarbonate: supplement with
0.1 gm/L L-glutamine, 0.35 gm/L sodium bicarbonate;
- Formulated at 10.9 grams of powder per liter of medium;
- Cell culture tested
|
Related
to M 0393, but without phenol red.
Sodium bicarbonate and
L-glutamine are added at the time that the liquid medium is formulated.
Phenol red has been
shown to interfere with the growth of some cells at low or cloning densities.
Use this version of
Medium 199 when working with stem cells or cells at low densities.