CLASSIC MEDIA AND SALTS
Cell Culture
Other Classic Media
Ames' Medium
The retina is an important in vitro model for the central nervous system. It is more
readily accessible than most nervous tissue and is strong enough to remain
intact during manipulations. The retina of the rabbit commonly contains no
penetrating blood vessels, but is nurtured by diffusion from capillary networks
on either side. For this reason, the tissue will survive and functions
without circulating blood, as long as it is bathed in a medium that closely
resembles in vivo conditions.
Ames' medium was formulated to support retinal tissue in relatively short-term
culture. Rabbit retina has been incubated in Ames' medium for over 2 days
with its metabolism and electrical responses to light stimuli well maintained. This mixture is a medium of choice for maintaining central
nervous system tissue in vitro.
| Product # |
Product Name |
Description |
Application |
|
A 1420 |
Ames' Medium |
- Powder
- With L-glutamine
- Without sodium bicarbonate; supplement with 1.9 gm/L sodium bicarbonate
- Formulated at 8.9 grams of powder per liter of medium;
- Cell culture tested
|
Use
to maintain retinal tissues for in vitro studies on central nervous system.
|
back to top |
|
Medium BGJ
was originally developed by Biggers, Gwatkin and Judah in the early 1960's at
the Wistar Institute. Subsequent studies resulted in a modification
designated BGJb which has been used for supporting cultures of cartilaginous
embryonic bone. An additional modification, developed by Sylvia Fitton-Jackson
at Strangeways Laboratory in England, is further enriched over the prior
formulae. Additional amino acids and vitamins, and increased buffering
capacity conferred by the phosphates in the Fitton-Jackson modification, create
conditions that permit calcification, as well as, growth of cartilaginous
embryonic bone.
|
| Product # |
Product Name |
Description |
Application |
|
B 6644 |
BGJb Medium
(Fitton-Jackson Modification) |
- Powder
- With L-glutamine
- Without sodium bicarbonate; supplement with 3.5 gm/L sodium bicarbonate
- Formulated at 21.3 grams of powder per liter of medium;
- Cell culture tested
|
Culturing
of cartilaginous embryonic bone cells.
|
back to top |
|
|
| Product # |
Product Name |
Description |
Application |
|
C 5572 |
Click's Medium
(Eagle's Ham's amino acids, EHAA) |
- Liquid
- With sodium bicarbonate
- Without mercaptoethanol and L-glutamine; supplement with 0.0.584 gm/L L-glutamine
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
back to top |
|
CMRL-1066
is a chemically defined medium developed in the late 1960's at the Connaugh
Medical Research Laboratories. A less complex and extensively modified
version of Medium 199, CMRL-1066 was designed initially for use with mouse
L-cells in non-supplemented culture. Although developed for use in
serum-free culture, CMRL-1066 can be supplemented with serum and used to support
the growth of many cell types.
|
| Product # |
Product Name |
Description |
Application |
|
C 0422 |
CMRL-1066 Medium |
- Powder
- With L-glutamine
- Without sodium bicarbonate; supplement with 2.2 gm/L sodium bicarbonate
- Cell culture tested
|
Suitable
for growing a wide variety of cell types, especially fibroblast-like cells.
|
back to top |
|
Fischer's
Medium was originally formulated to support serial propagation of cells from
leukemic mice. In initial studies, cells in culture were examined for
resistance to chemotherapeutic agents. While these studies were typically
carried out in whole animals, Fischer's medium made it possible to conduct
parallel studies in vitro. Fischer's medium supports clonal reproduction
of cells, particularly lymphoblasts from primary explants or from cells in
culture.
|
| Product # |
Product Name |
Description |
Application |
|
F 5008 |
Fischer's Medium |
- Powder
- With L-glutamine
- Without sodium bicarbonate; supplement with 1.125 gm/L sodium bicarbonate;
- Formulated at 10.6 grams of powder per liter of medium;
- Cell culture tested
|
Useful
for cloning and low density culture of hematopoeitic cells, especially
lymphocytes.
|
back to top |
|
Glascow
Minimum Essential Medium was originally developed by Ian MacPherson and Michael
Stoker as a modification of Eagle's medium (BME). The modifications included
adding 10% tryptose phosphate and twice the normal concentration of amino acids
and vitamins. This medium
was used to study the genetic factors affecting cell competence. Polyoma
virus was used to transform four fibroblast clones from a culture of baby
hamster kidney cells.
|
| Product # |
Product Name |
Description |
Application |
|
G 5154 |
Glasgow Minimum Essential Medium |
- Powder
- With L-glutamine
- Without tryptose phosphate broth and sodium bicarbonate; supplement with
2.75 gm/L sodium bicarbonate;
- Formulated at 12.5 grams of powder per liter of medium;
- Cell culture tested
|
For
use with adherent kidney cell lines such as baby hamster kidney cells (BHK).
Complete GMEM
formulation by supplementing with tryptose phosphate broth (T 8159) at
100 mL per liter of medium.
|
|
G 6148 |
Glasgow Minimum Essential Medium |
- Liquid
- With sodium bicarbonate
- Without L-glutamine; supplement with 0.292 gm/L L-glutamine
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Liquid
version of G 5154.
Complete GMEM
formulation by supplementing with tryptose phosphate broth (T 8159) at
100 mL per liter of medium.
|
back to top |
|
Guilber and
Iscove demonstrated that precursor cells of erythrocytes and macrophages could
be cultured in a totally defined serum-free medium supplemented with albumin,
transferrin, lecithin, and selenium. This medium is a modification of
Dulbecco's Modified Eagle's medium (DME) and contains selenium, additional amino
acids and vitamins, sodium pyruvate, HEPES buffer and potassium nitrate instead
of ferric nitrate.
Studies
have demonstrated that Iscove's medium supports murine B lymphocytes,
hematopoeitic tissue from bone marrow, B cells stimulated with lipopolysaccharide, T lymphocytes, and a variety of hybrid cells.
|
| Product # |
Product Name |
Description |
Application |
|
I 2510 |
Iscove's Modified Dulbecco's Medium Hybri-Max. |
- Powder
- With L-glutamine and 25 mM HEPES
- Without sodium bicarbonate; supplement with 3.024 gm/L sodium bicarbonate;
- Formulated at 17.7 grams of powder per liter of medium;
- Endotoxin tested
- Hybridoma tested
|
Hybridoma
tested version of I 7633.
Use this product when
working with hybridoma cells.
|
|
I 3390 |
Iscove's Modified Dulbecco's Medium Hybri-Max. |
- Liquid
- With sodium bicarbonate
- Without L-glutamine; supplement with 0.584 gm/L L-glutamine
- Sterile-filtered
- Endotoxin tested
- Hybridoma tested
|
Liquid
and hybridoma tested version of I 7633
Use this product when
working with hybridoma cells.
|
|
I 7633 |
Iscove's Modified Dulbecco's Medium |
- Powder
- With L-glutamine and 25 mM HEPES
- Without sodium bicarbonate; supplement with 3.024 gm/L sodium bicarbonate;
- Formulated at 17.7 grams of powder per liter of medium;
- Cell culture tested
|
IMDM is frequently used for
serum-free growth of hematopoeitic cells. This includes fusion cell
lines such as hybridomas.
Recommended as a base
formulation for the development of unique serum-free formulations.
HEPES increases the
buffer strength of the medium and raises the maximum buffering range.
Recommended for cells
that grow better at a pH above 7.2.
|
back to top |
|
L-15 Medium
(Leibovitz) was originally formulated for use in carbon dioxide free systems
requiring sodium bicarbonate supplementation. L-15 is buffered by its
complement of salts, free base amino acids and galactose (in place of
glucose). When properly supplemented, L-15 medium supports
established cell lines, such as HEp-2 and LLC-MK2, and primary
explants of embryonic and adult human tissue. Many viruses have been
successfully cultivated in this medium.
|
| Product # |
Product Name |
Description |
Application |
|
L 4386 |
L-15 Medium (Leibovitz) |
- Powder
- With L-glutamine
- Formulated at 14.8 grams of powder per liter of medium;
- Cell culture tested
|
Primarily
used for the culture of adherent primary explants of embryonic and adult human
tissue; especially, when these explanted cells; are used for production
of virus.
|
|
L 5520 |
L-15 Medium (Leibovitz) |
- Liquid
- Without L-glutamine; supplement with 0.3 gm/L L-glutamine
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Liquid
formulation of L 4386.
Primarily used for the
culture of adherent primary explants of embryonic and adult human tissue;
especially, when these explanted cells are used for production of virus.
|
back to top |
|
In 1959,
McCoy and his coworkers reported the amino acid requirements for in vitro
cultivation of Novikoff Hepatoma cells. These studies were performed using
Basal Medium 5A, which was subsequently modified to create a new medium known as
McCoy's 5A medium. Hsu and Kellogg employed this medium to support the
growth of primary cultures derived from normal bone marrow, skin, gingiva,
testes, mouse kidney, omentum, adrenal glands, lung, spleen, rat embryos, and
other tissues.
|
| Product # |
Product Name |
Description |
Application |
|
M 4892 |
McCoy's 5A Modified |
- Powder
- With L-glutamine
- Without sodium bicarbonate; supplement with 2.2 gm/L sodium bicarbonate;
- Formulated at 12.0 grams of powder per liter of medium;
- Cell culture tested
|
Used
for the primary culture of cells from a wide range of tissues.
Recommend use for
production of virus in primary cell cultures.
|
|
M 8403 |
McCoy's 5A Modified |
- Liquid
- With sodium bicarbonate
- Without L-glutamine; supplement with 0.22 gm/L L-glutamine
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Liquid
form of M 4892.
Used for the primary
culture of cells from a wide range of tissues.
Recommend use for
production of virus in primary cell cultures.
|
|
M 9270 |
McCoy's 5A Modified |
- Powder
- With L-glutamine
- Without phenol red and sodium bicarbonate; supplement with 2.2 gm/L sodium bicarbonate;
- Formulated at 11.9 grams of powder per liter of medium;
- Cell culture tested
|
Same
as M 4892, except that phenol red is not included.
Phenol red has been
shown to interfere with the growth of some cells at cloning densities.
Use this version of
McCoy's 5A when working with stem cells or cells at low densities..
|
back to top |
|
NCTC-135
was developed by the Tissue Culture Section, Laboratory of Biology, National
Cancer Institute (NCI), Bethesda, MD. NCTC-135 is a modification of NCTC-109
medium, also developed by the NCI. NCTC-135 differs from NCTC-109 in that the
L-cysteine-HCl, which is toxic to certain cell types, was replaced with L-cystine. NCTC-109 and
the 135 modification were formulated to establish and maintain a strain of mouse
cells (L929) in a chemically defined and serum free medium.
Successful establishment of L 929 cells led to further nutritional and
metabolic studies and the establishment of at least ten other cell lines adapted
to this medium.
|
| Product # |
Product Name |
Description |
Application |
|
N 1140 |
NCTC 109 Medium |
- Liquid
- With sodium bicarbonate
- Without L-glutamine; supplement with 0.136 gm/L L-glutamine
- Sterile-filtered
- Endotoxin tested
- Cell culture tested>
|
NCTC-109 and
the 135 modification were formulated to establish and maintain a strain of mouse
cells (L929) in a chemically defined and serum free medium.
|
back to top |
|
Swim's S-77
medium (designated S-77) is a modification of MEM Eagle's medium that lacks biotin
and has increased levels of amino acids and inositol. This medium was
designed to support suspension cultures of cells derived from the Novikoff
hepatoma. It is an incomplete basal medium, requiring additions of
L-glutamine, L-cystine, calcium chloride and other components. When
properly supplemented, Swim's S-77 medium has been used to establish, in
culture, a number of stable cell lines (N1-31, N1-S1/FUdr) derived from the
Novikoff hepatoma.
|
| Product # |
Product Name |
Description |
Application |
|
S 2513 |
Swim's S-77 Medium |
- Powder
- With 12.5 mg/L streptomycin sulfate.
- Without penicillin G sodium, L-glutamine, L-cystine, calcium chloride, phenol red, and sodium
bicarbonate;
- Supplement with 0.265 gm/L calcium chloride; 0.012 gm/L L-cystine; 0.292 gm/L
L-glutamine; 2.2 gm/L sodium bicarbonate
- Formulated at 9.0 grams of powder per liter of medium;
- Cell culture tested.
|
Designed to support
suspension cultures of cells derived from the Novikoff hepatoma. |
|
|
|
back to top |
|
Waymouth Medium MB 752/1 was developed as a totally defined synthetic medium for the cultivation of mouse L929 cells in a serum-free environment. Clones of L929 cells, conditioned to grow in serum-supplemented or serum free medium were cultivated for more than thirty passages in serum-free Waymouth medium and
demonstrated a doubling time of approximately 24 hours. The applicability
of Waymouth Medium MF 752/1 has been extended to include whole organ culture,
establishment of carcinoma cell lines from pleural effusions, and the growth of
potentially tumorigenic cells prior to their assessment in vitro.
|
| Product # |
Product Name |
Description |
Application |
|
W 1625 |
Waymouth MB 752/1 Medium |
- Liquid
- With sodium bicarbonate
- Without L-glutamine; supplement with 0.3 gm/L L-glutamine
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
For the
cultivation of mouse L929 cells in a serum-free environment. Also used in whole organ culture,
establishment of carcinoma cell lines from pleural effusions, and the growth of
potentially tumorigenic cells prior to their assessment in vitro.
|
back to top |
|
In 1971,
Williams et. al. described a procedure for enriching isolated hepatocyte
cultures for poylgonal epithelial cells and reducing the number of contaminating
fibroblasts. This method was a sequential plating technique based on the
observation that fibroblast-like cells adhere to a substrate more rapidly than
epithelial cells. The isolated epithelial cells resulting from this
treatment could then be cultured in a rich medium designate William's Medium
D. Newborn animals were the source of cells used in these studies, as they
were in most studies of liver cell culture. Since newborn liver in not
functionally mature, further studies were conducted by Williams and Gunn to
explore the possibility of culturing adult liver cells on a long term
basis. The medium developed during the course of these studies was
designated William's Medium E. It has been shown to support the growth in
long-term culture of adult liver epithelial cells.
|
| Product # |
Product Name |
Description |
Application |
|
W 4125 |
Williams' Medium E |
- Powder
- With L-glutamine
- Without sodium bicarbonate; supplement with 2.2 gm/L sodium bicarbonate;
- Formulated at 10.8 grams of powder per liter of medium;
- Cell culture tested
|
For
long-term culture of adult liver epithelial cells.
|
|
W 4128 |
Williams' Medium E |
- Liquid
- With sodium bicarbonate
- Without L-glutamine; supplement with 0.292 gm/L L-glutamine
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Liquid
version of W 4125.
Add glutamine at time
of formulation.
|
|
W 1878 |
Williams' Medium E |
- Liquid
- With sodium bicarbonate
- Without L-glutamine and phenol red; supplement with 0.292 gm/L L-glutamine
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Same
as W 4128, except that phenol red has been removed.
Phenol red has been shown to interfere with the growth of some cells at low or cloning densities.
Use this version of William's E when working with stem cells or cells at low densities.