| Product # |
Product Name |
Description |
Application |
|
R 0883 |
RPMI-1640 Medium |
- Liquid
- With sodium bicarbonate
- Without L-glutamine; supplement with 0.3 gm/L L-glutamine
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Same
as R 8758, except that L-glutamine must be added by the investigator.
|
|
R 8758 |
RPMI-1640
Medium |
- Liquid
- With sodium bicarbonate and L-glutamine
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Complete
liquid version of the original RPMI-1640 formulation published by Moore et.
al. in 1967.
Developed for growth of
human normal and neoplastic leukocytes (peripheral blood lymphocytes).
Generally, supplemented with serum. |
|
R 1145 |
RPMI-1640
Medium (10x) |
- Liquid
- Without L-glutamine, folic acid and sodium bicarbonate
- Supplement with 0.3 gm/L L-glutamine, 0.001gm/L folic acid and 2.0 gm/L sodium bicarbonate at 1x
- pH 1.8-2.2 at 10x
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
This
is the concentrated version (10x) of R 8758.
The L-glutamine and
sodium bicarbonate are added to the medium after dilution to 1x and pH
adjustment.
|
|
R 6504 |
RPMI-1640 Medium |
- Powder
- With L-glutamine
- Without sodium bicarbonate; supplement with 2.0 gm/L sodium bicarbonate.
- Formulated at 10.4 grams of powder per liter of medium.
- Cell culture tested
|
Related to R 5382, RPMI-1640
Medium Hybri-Max.
|
|
R 7509 |
RPMI-1640
Medium Modified |
- Liquid
- With sodium bicarbonate
- Without phenol red and L-glutamine; supplement with 0.3 gm/L L-glutamine.
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Phenol
red has been shown to interfere with the growth of some cells at clonal
densities.
Use this medium when
working with stem cells or when growing cells at low densities.
Also recommended for in
vitro diagnostics use. |
|
R 5886 |
RPMI-1640 Medium HEPES Modification |
- Liquid
- With 25 mM HEPES and sodium bicarbonate
- Without; L-glutamine; supplement with 0.3 gm/L L-glutamine.
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Same
as R 8758, except that HEPES buffer has been added.
HEPES increases the
buffer strength of the medium and raises the maximum buffering range.
Recommended for cells
that grow better at a pH above 7.2. |
|
R 4130 |
RPMI-1640 Medium HEPES Modification |
- Powder
- With L-glutamine and 25 mM HEPES
- Without sodium bicarbonate; supplement with 2.0 gm/L sodium bicarbonate.
- Formulated at 16.4 grams of powder per liter of medium.
- Cell culture tested
|
Same
as R 8758, except that HEPES buffer has been added.
HEPES increases the
buffer strength of the medium and raises the maximum buffering range.
Recommended for cells
that grow better at a pH above 7.2. |
|
R 7638 |
RPMI-1640 Medium Dutch Modification |
- Liquid
- With 1 gm/L sodium bicarbonate and 20 mM HEPES
- Without L-glutamine; supplement with 0.3 gm/L L-glutamine.
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Same
as R 8758, except that the concentration of sodium bicarbonate has been cut in
half and HEPES buffer has been added.
HEPES increases the
buffer strength of the medium and raises the maximum buffering range.
Recommended for cells
that grow better at a pH above 7.2.
|
|
R 7388 |
RPMI-1640
Medium Modified |
- Liquid
- With 20 mM HEPES and L-glutamine
- Without sodium bicarbonate; supplement with 2.0 gm/L sodium bicarbonate.
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Same
as R 8758, except that HEPES buffer has been added.
HEPES increases the
buffer strength of the medium and raises the maximum buffering range.
Recommended for cells
that grow better at a pH above 7.2.
|
|
R 8755 |
RPMI-1640 Medium Modified |
- Powder
- With L-glutamine
- Without phenol red and sodium bicarbonate; supplement with 2.0 gm/L sodium bicarbonate.
- Formulated at 10.4 grams of powder per liter of medium.
- Cell culture tested
|
Phenol red has been shown to interfere with the growth of some cells at clonal densities.
Use this medium when
working with stem cells or when growing cells at low densities.
Also recommended for in
vitro diagnostics use. |
|
R 7755 |
RPMI-1640
Medium Auto-Mod™ |
- Powder
- Without L-glutamine and sodium bicarbonate; supplement with 0.3 gm/L of L-glutamine and 2.0 gm/L sodium bicarbonate.
- Formulated at 10.3 grams of powder per liter of medium.
- Cell culture tested
|
Modified
for autoclaving
|
|
R 1383 |
RPMI-1640 Medium |
- Powder
- With L-glutamine
- Without glucose and sodium bicarbonate; supplement with 2.0 gm/L sodium bicarbonate.
- Formulated at 8.4 grams of powder per liter of medium.
- Cell culture tested
|
Without
glucose, allows investigator to add own energy source.
|
|
R 7513 |
RPMI-1640 Medium Modified |
- Liquid
- Without sodium bicarbonate
- Without L-methionine, L-cystine, and L-glutamine
- Supplement with 0.652 gm/L L-cystine-2HCL, 0.015 gm/L L-methionine, and 0.3 gm/L of
L-glutamine
- Sterile-filtered
- Endotoxin tested
- Cell culture tested
|
Formulated
without sulfur containing amino acids.
Recommended for
metabolism studies using radio-labeled sulfur containing amino acids.