|
Sigma's line of coated plates for high throughput screening undergo extensive validation. Each lot is assayed to ensure the exceptional performance and consistent quality necessary to generate reliable results. For detailed information regarding the assay performed, click on the Technical Bulletin icon: 
Biotinylated Biomolecule Capture
Cell-Based Assays
Epitope Tag and Protein Capture Plates
Biotinylated Biomolecule Capture
SigmaScreen Streptavidin Coated Plates
| Description |
 |
Biotin Binding Capacity |
%CV well to well |
%CV plate to plate |
DNase |
Protease |
96-well clear,
M 5432
96-well white, M 5557 |
 |
>25 pmole biotin per well |
<10% |
<10% |
None detected |
None detected |
| 96-well black,
M 4058 |
 |
>25 pmole biotin per well |
<12% |
<12% |
None detected |
None detected |
384-well clear,
S 8686
384-well white, S 8561
384-well black, S 8811 |
 |
>4 pmole biotin per well |
<10% |
<10% |
None detected |
None detected |
SigmaScreen Streptavidin High Capacity Coated Plates
| Description |
 |
Biotin Binding Capacity |
%CV well to well |
%CV plate to plate |
DNase |
Protease |
| 96-well clear,
S 6940 |
 |
>300 pmole biotin per well |
<15% |
<15% |
None detected |
None detected |
| 96-well black,
S 8940 |
 |
>200 pmole biotin per well |
<15% |
<15% |
None detected |
None detected |
| 96-well white,
S 9065 |
 |
>250 pmole biotin per well |
<15% |
<15% |
None detected |
None detected |
| 384-well clear,
S 8815 |
 |
>150 pmole biotin per well |
<15% |
<15% |
None detected |
None detected |
| 384-well black,
S 9190 |
 |
>100 pmole biotin per well |
<15% |
<15% |
None detected |
None detected |
| 384-well white,
S 9315 |
 |
>125 pmole biotin per well |
<15% |
<15% |
None detected |
None detected |
SigmaScreen ExtrAvidin Coated Plates
| Description |
 |
Biotin Binding Capacity |
%CV well to well |
%CV plate to plate |
DNase |
Protease |
| 96-well clear,
E 3027 |
 |
>25 pmole biotin per well |
<10% |
<10% |
None detected |
None detected |
back to top
Cell-Based Assays
SigmaScreen Poly-D-Lysine Coated Plates
| Description |
 |
Bioburden |
Endotoxin |
Plate Performance |
96-well clear,
M 3555
96-well white, M 5682
96-well black, M 5307 |
 |
<0.52% wells contaminated (<10 wells out of 20 plates) |
<0.5 EU/mL for all wells tested |
Tested wells show >2X signal of negative control wells. Visual confirmation of greater number of cells (BHK 21) adhered to test wells than negative control wells. |
384-well clear,
S 7436
384-well white, S 7561
384-well black, S 7686 |
 |
<0.52% wells contaminated (<40 wells out of 20 plates) |
<0.5 EU/mL for all wells tested |
Tested wells show >1.5X signal of negative control wells. Visual confirmation of greater number of cells (BHK 21) adhered to test wells than negative control wells. |
SigmaScreen Collagen I Coated Plates
| Description |
 |
Bioburden |
Endotoxin |
Plate Performance |
96-well clear,
S 8813
96-well white, S 3315 96-well black, S 3190 |
 |
<0.52% wells contaminated (<10 wells out of 20 plates) |
<0.5 EU/mL for all wells tested |
Test wells show approximately 90% of HT 1080 cells spread, as compared to negative control wells which demonstrate rounded cellular morphology and poor spreading. |
384-well clear,
S 3440
384-well white, S 3565 384-well black, S 3690 |
 |
<0.52% wells contaminated (<40 wells out of 20 plates) |
<0.5 EU/mL for all wells tested |
Test wells show approximately 90% of HT 1080 cells spread, as compared to negative control wells which demonstrate rounded cellular morphology and poor spreading. |
back to top
Epitope Tag and Protein Capture Plates
HIS-Select HC Nickel Coated Plate
| Description |
 |
%CV well to well |
%CV plate to plate |
Binding Capacity |
| 96-well clear,
S 5563 |
 |
<15% |
<15% |
>4 5g protein per well |
| 384-well clear,
H 1661 |
 |
<15% |
<15% |
>2 5g protein per well |
HIS-Select HS Nickel Coated Plate
| Description |
 |
%CV well to well |
%CV plate to plate |
Minimum Detectable |
Protein Binding |
| 96-well strip,
S 5688 |
 |
<10% |
<10% |
Absorbance of 1 ng/well |
>0.20 |
Anti-FLAG HS M2 Coated Plate
| Description |
 |
%CV well to well |
%CV plate to plate |
Binding Sensitivity |
| 96-well clear,
P 2983 |
 |
<10% |
<10% |
Detection of <1 ng of FLAG-BAP-HIS Control Protein above background |
Protein G HS Coated Plate
| Description |
 |
%CV well to well |
%CV plate to plate |
Binding Sensitivity (signal/noise ratio) |
| 96-well clear,
S 2063 |
 |
<10% |
<10% |
>1.5 (test wells show detection of an antibody Alkaline Phosphatase conjugate of <1 ng above background) |
Glutathione Coated Plate
| Description |
 |
%CV well to well |
%CV plate to plate |
Binding Capacity |
Binding Sensitivity |
| 96-well clear,
P 3233 |
n/a |
<10% |
<10% |
>10 ng purified GST |
<1 ng purified GST detected |
back to top
Sigma's line of coated plates utilize Corning® base plates.
Corning is a registered trademark of Corning Incorporated, Corning, NY 14831-0001.
SigmaScreen is a trademark of Sigma-Aldrich Corporation, St. Louis, MO 63103.
|