CHO CELLS MEDIA AND MEDIUM SUPPLEMENTS |
 |
Cell Culture |
 |
 |
| |
|
|
The development of genetically-engineered cell lines to express therapeutically valuable
recombinant proteins has created a major break-through in health care.
While the earliest methods of expressing recombinant proteins relied on bacterial systems,
it is now recognized that the pharmacology and efficacy of recombinant proteins produced for therapeutic applications are affected by post-translational
modifications such as glycosylation and phosphorylation. This has led to the use of higher order cell systems.
The Chinese Hamster Ovary cell line (CHO) was one of the first mammalian cell lines successfully developed for use in the production of therapeutically valuable
proteins. Today, variations of this cell line are widely used for this purpose. To accommodate the variety among different CHO clones, Sigma
offers several different formulations to meet the needs of specific end users.
For more information on medium selections click here
|
| Product # |
Product Name |
Description |
Application |
|
CH0001 |
CHO Kit 1
Performance Data
|
- Liquid
- Kit consists of 6 proprietary medium formulations containing inorganic salts, HEPES and sodium
bicarbonate buffers, essential and non-essential amino acids, vitamins, recombinant human insulin,
plant hydrolysates, other organic compounds, trace elements, and surfactants.
- Medium formulations do not contain antibiotics, antimycotics, L-glutamine, or transferrin.
- Animal Component-free
- Sterile filtered
- Cell culture tested
|
To determine which medium is optimal for an individual CHO cell clone, it is
often necessary to screen multiple formulations. This kit provides a platform from which the user
can screen several unique CHO modifications. Initial screening of the six media for growth and
productivity may offer enough improvement over your current culture system to suffice. However, if
further medium optimization is desired, the user can select the top three performing media based on
cell growth or productivity. These three formulations can be combined in mixture experiments as
described in the kit and can be analyzed to determine a best-fit media based on growth and productivity.
|
|
C 8862 |
CHO DHFR- Medium
Performance Data
|
- Liquid
- Proprietary formulation containing inorganic salts, HEPES and sodium bicarbonate buffers, essential
and non-essential amino acids, vitamins, recombinant human insulin, plant hydrolysates, other organic
compounds, trace elements, and surfactants.
- Does not contain antibiotics, antimycotics, L-glutamine, or transferrin.
- Animal Component-free
- Sterile filtered
- Cell culture tested
|
This animal component-free medium (containing no hypoxanthine or thymidine) is formulated to maximize cell growth and recombinant protein production using the Dihydrofolate Reductase (DHFR) gene amplification system in DHFR- Chinese Hamster Ovary (CHO) cells.
This medium supports rapid initial cell growth and high levels of protein expression in suspension cultures. This medium supports and maintains high cell densities for extended periods, resulting in increased productivity.
|
| C 5467 |
CHO Protein-free Medium, Animal Component-free Medium for Suspension Culture (PF-ACF-CHO)
Performance Data
|
- Liquid
- Proprietary formulation
- With inorganic salts, HEPES and sodium bicarbonate buffers, essential and non-essential amino acids, vitamins, rHu-Insulin, other organic compounds, trace elements, and surfactants.
- With plant hydrolysates
- Without antibiotics, anti-mycotics, transferrin, or L-glutamine: add 20-40 mL of 200 mM L-glutamine per liter, prior to use.
- Without hypoxanthine and thymidine;
- pH 7.1-7.7
- Sterile filtered.
- Endotoxin tested.
- Cell culture tested.
|
Optimized for the production of recombinant proteins by cultured CHO cells, under protein-free and animal derived component-free conditions; designed to support the rapid growth, high cell densities and extended viability of genetically engineered CHO cells in suspension culture.
This formulation eliminates regulatory concerns about the risks of biological contamination of recombinant proteins that are associated with the use of animal
components during cell culture.
May be used in Dihydrofolate Reductase (DHFR) gene amplification systems. |
| C 9098 |
CHO Protein-free Medium, Animal Component-free Medium for Suspension Culture (PF-ACF-CHO) |
- Powder
- Proprietary formulation
- Formula contains inorganic salts, HEPES and sodium bicarbonate buffers, essential and non-essential amino acids, vitamins, rHu-Insulin, other organic compounds, trace elements, and surfactants.
- Formula contains plant hydrolysates
- Formula without antibiotics, anti-mycotics, hypoxanthine, thymidine or transferrin.
- Without L-glutamine and sodium bicarbonat: supplement with 0.584 to 1.168 gm/L of L-glutamine and 1.5 gm/L of sodium bicarbonate.
- pH 6.0 +/- 1.0 without sodium bicarbonate, and 7.1-7.7 fully formulated.
- Formulated at 21.5 grams per liter of medium.
- Sterile filtered.
- Endotoxin tested.
- Cell culture tested.
|
This is the powder version of C 5467. It is only available as a custom formulation.
Optimized for the production of recombinant proteins by cultured CHO cells, under protein-free and
animal derived component-free conditions; designed to support the rapid growth, high cell densities and extended
viability of genetically engineered CHO cells in suspension culture.
This formulation eliminates regulatory concerns about the risks of biological contamination of recombinant proteins that are associated with the use of animal
components during cell culture.
May be used in Dihydrofolate Reductase (DHFR) gene amplification systems. |
|
C 8730 |
CHO Protein-free Medium, Animal Component-free Medium for Adherent Culture (PF-ACF-CHO)
Performance Data
|
- Liquid
- Proprietary formulation
- With inorganic salts, HEPES and sodium bicarbonate buffers, essential and non-essential amino acids, vitamins, rHu-Insulin, other organic compounds, trace elements, and surfactants.
- With plant hydrolysates
- Without antibiotics, phenol red, and L-glutamine: add 20 mL of 200 mM L-glutamine per liter, prior to use.
- Without hypoxanthine and thymidine;
- Sterile filtered.
- Endotoxin tested.
- Cell culture tested.
|
Modified version of C 5467, divalent cation concentrations have been
optimized to promote better attachment. Designed to support the growth of, and expression of recombinant proteins by, adherent CHO cells in a variety of
systems, such as; T-flasks, roller bottles and bioreactors using fiber discs or micro-carrier beads.
This formulation eliminates regulatory concerns about the risks of biological contamination of recombinant proteins that are associated with the use of animal components during cell culture.
May be used in Dihydrofolate Reductase (DHFR) gene amplification systems. |
| C 4726 |
CHO Chemically-defined, Animal Component-free Medium
(CD-ACP-CHO) |
- Liquid
- Proprietary formulation
- With inorganic salts, HEPES and sodium bicarbonate buffers, essential and non-essential amino acids, vitamins, rHu-insulin, other organic compounds, trace elements, and surfactants.
- Without plant hydrolysates
- Without antibiotics, anti-mycotics, transferrin, any undefined nutrients or supplement and L-glutamine: add 20-40 mL of 200 mM L-glutamine per liter, prior to use.
- Without hypoxanthine and thymidine;
- pH 7.1-7.7
- Sterile filtered.
- Endotoxin tested.
- Cell culture tested.
|
All components in this animal protein free medium are chemically defined. This allows biotechnologists to have
greater control over the biological risk factors associated with the production of CHO expressed proteins; AND to optimize cell growth, protein production, and the down-stream purification process. Furthermore, this formulation offers the biotechnologist the best opportunity to manage and control lot-to-lot variations in productivity.
This formulation eliminates regulatory concerns about the risks of biological contamination of recombinant proteins that are associated with the use of animal components during cell culture.
May be used in Dihydrofolate Reductase (DHFR) gene amplification systems.
|
| C 1707 | CHO Serum Free Medium (CHO-SF) |
- Liquid
- With fetuin, insulin, BSA, transferrin, and HEPES and sodium bicarbonate buffers;
- Without L-glutamine: add 10.3 mL of 200 mM L-glutamine per liter, prior to use
- Without hypoxanthine and thymidine;
- Protein content: 100 ug/mL
- Sterile filtered
- Endotoxin tested
- Cell culture tested
|
Optimized for the production of recombinant proteins, under serum-free conditions, which are expressed in either suspension or attached cultured CHO cells.
This formulation reduces regulatory concerns, about the risks of biological contamination of recombinant therapeutics, that are associated with the use of serum.
May be used in Dihydrofolate Reductase (DHFR) gene amplification systems. |
|
|
|
back to top |
|
|
|
Medium Selection Information |
|
The FDA requires that all new drugs meet stringent safety requirement. The potential for infecting a patient with an adventitious biological agent that contaminates a recombinant protein drug is among their greatest concerns. The most likely potential sources of biological contamination of these drugs manufactured in well designed cGMP facility are the animal-derived components and supplements
used in the cell culturing process.
Production of recombinant proteins using CHO cells is a complex process. The growth and productivity of a cell line (derived from a cell bank) must be fully characterized and understood in the context of the specific protein being developed. Furthermore, no single medium formulation will meet the requirements of all situations. It is frequently necessary to balance the need for maximum performance with the regulatory agencies requirements for safety.
Sigma's CHO formulations offer choices ranging from fully supplemented medium to fully defined
medium. This strikes a good balance between maximal productivity and maximal safety from adventitious agents.
Product C 1707 contains animal-derived components. Products C 5467, C8730 and C9098
contain no animal-derived components. Product C 4726 contains no animal-derived components and is also free from any chemically-undefined
components, like plant hydrolysates. Customers who are indifferent to the presence of animal-derived components may want to try C 1707 as a starting
point. Customers who would like to work animal-component free should start with C 5467 in their initial trials.
|
back to top
|
Chinese Hamster Ovary (CHO) cell expression systems are complex, and are typically modified (or optimized) to meet the specific priorities and needs of the biotechnologist. In addition to offering the above varieties of CHO media, Sigma provides supplements that you may find useful in your culture system. The table below identifies some of the most useful supplements for your evaluation and use.
Other important potential supplements such as the transferrins, albumins, fetuins, and protein hydrolysates are offered by Sigma in many varieties. Please use the Sigma search engine to find these product families.
|
| Product # |
Product Name |
Description |
Application |
| A 1345 |
Anti-oxidant Supplement (1000x) |
- Liquid
- Sterile filtered.
- Endotoxin tested.
- Cell culture tested.
|
Proprietary medium supplement specially formulated to boost cell growth for enhanced protein expression. May be used in combination
with Polyamine Supplement (P 8483)
|
| F 7050 |
Fatty Acid Supplement |
- Liquid
- Sterile filtered.
- Endotoxin tested.
- Cell culture tested.
|
Animal-component free. Proprietary aqueous mixture of fatty acids that have been found to be optimal
for cell growth and protein expression.
|
| G 7513 |
L-Glutamine Solution, 200 mM |
- Liquid
- Sterile filtered.
- Endotoxin tested.
- Cell culture tested.
|
Prepared in cell culture grade water. Useful for supplementing liquid CHO media.
|
| G 9785 |
GSEM Supplement (Glutamine Synthetase Expression Medium Supplement) (50x) |
- Liquid
- Sterile filtered.
- Endotoxin tested.
- Cell culture tested.
|
The glutamine synthetase (GS) system is used to express recombinant proteins from mammalian cells.
Glutamine-dependent cells transfected with a glutamine synthetase gene can be selectively grown in glutamine-deficient medium.
|
| P 8340 |
Protease Inhibitor Cocktail (1000x) for Use in Mammalian Cell Expression Systems |
- Liquid, in DMSO
- Contains 4-(2-aminoethyl) benzene sulfonyl fluoride (AEBSF), pepstatin A, trans epoxysuccinyl L-leucylamido(4-guanidino) butane
(E-64), bestatin, leupeptin, and aprotinin.
- Contains no metal chelators.
|
Endogenous enzymes, notably proteases and phosphatases, can degrade proteins. The best way to improve
the yield of fully functional proteins is to add inhibitors of these lytic enzymes. This formulation contains
inhibitors of serine, cysteine, and aspartic proteases, as well as inhibitors of aminopeptidases.
This cocktail may be used effectively to increase the quality of protein expressed in CHO cells. However,
it should always be thoroughly tested for specific cell culture applications.
|
| P 1300 |
Pluronic F-68, Non-ionic Detergent |
- Liquid
- Cell culture tested.
|
A media additive often used to protect cells growing in suspended culture systems from damage which may occur during
agitation and aeration.
|
| P 8483 |
Polyamine Supplement (1000x) |
- Liquid
- Sterile filtered.
- Endotoxin tested.
- Cell culture tested.
|
Medium supplement formulated to boost cell growth for enhancement of protein expression.
|
| S 5761 |
Sodium Bicarbonate |
- Powder
- Molecular Weight 84.01
- Purity >99.5%
- Cell culture tested.
|
Use to supplement cell culture media.
|
| S 8761 |
Sodium Bicarbonate, 7.5% solution |
- Liquid
- Sterile filtered.
- Endotoxin tested.
- Cell culture tested.
|
Use to supplement cell culture media.