Normal attachment, growth, and development of many cell types are dependent on attachment factors and extracellular matrix components. While some cells are able to synthesize these components, others require an exogenous source, particularly when grown in serum-free culture.
To help facilitate attachment, cell spreading, growth, morphology, differentiation, and motility of your cells, Sigma offers an extensive line of attachment and matrix factors. Each lot is cell culture tested to assess its ability to promote cell attachment and spreading.
Collagen
Chondroitin Sulfate A
ECM gel
Fibronectin
Gelatin
Laminin
Superfibronectin
Thrombospondin
Vitronectin
Proteoglycans
Poly-Lysine
Disintegrins
Fibronectin Proteolytic Fragments
Collagen Type I
Target cells for attachment:
- Epithelial cells
- Muscle cells
- Nerve cells
Mediate the attachment of many cell types
Product Use
Collagen Type I, Acid soluble powder
| Product Number |
Source |
Storage |
Concentration for Use |
| C 1809 |
Kangaroo Tail |
2-8°C |
6-10 µg/cm2 |
| C 7661 |
Rat Tail |
2-8°C |
6-10 µg/cm2 |
| C 9791 |
Calf Skin |
2-8°C |
6-10 µg/cm2 |
Collagen Type I,
0.1% Solution, Sterile-filtered (Not suitable for 3D gel formation)
| Product Number |
Source |
Storage |
Concentration for Use |
| C 8919 |
Calf Skin |
2-8°C |
6-10 µg/cm2 |
back to top
Collagen Type II
Target cells for attachment:
Product Use
Collagen Type II,
Powder
| Product Number |
Source |
Storage |
Concentration for Use |
| C 9301 |
Chicken Sternal Cartilage |
2-8°C |
6-10 µg/cm2 |
back to top
Collagen Type IV
Target cells for attachment:
- Epithelial cells
- Enodothelial cells
- Muscle cells
- Nerve cells
Product Use
Collagen Type IV, Powder
| Product Number |
Source |
Storage |
Concentration for Use |
| C 0543 |
Engelbreth-Holm-Swarm mouse sarcoma |
-20°C; store solubilized product at 2-8°C |
6-10 µg/cm2 |
Collagen Type IV, Lyophilized
| Product Number |
Source |
Storage |
Concentration for Use |
| C 5533 |
Human placenta |
-20°C |
6-10 µg/cm2 |
back to top
Chondroitin Sulfate A
Appears to play a regulatory role for:
- Chondrocytes
- Neural cells
- Some tumor cells
| Product Number |
Source |
Storage |
Concentration for Use |
| C 9819 |
Bovine trachea |
2-8°C |
20 µg-2 mg/ml |
back to top
ECM gel
Target cells for attachment:
- Epithelial cells
- Enodothelial cells
- Muscle cells
- Nerve cells
- Tumor cells
| Product Number |
Source |
Storage |
Concentration for Use |
| E 1270 |
Engelbreth-Holm-Swarm mouse sarcoma |
-20°C; store solubilized product at 2-8°C |
6-10 µg/cm2 |
back to top
Fibronectin
Target cells for attachment:
- Epithelial cells
- Mesenchymal cells
- Neuronal cells
- Fibroblasts
- Neural crest cells
- Endothelial cells
Product Use
Fibronectin, 0.1% Solution
| Product Number |
Source |
Storage |
Concentration for Use |
| F 0895 |
Human plasma |
2-8°C |
1-5 µg/cm2 |
| F 1141 |
Bovine plasma |
2-8°C |
1-5 µg/cm2 |
Fibronectin, Fragment III1 -C
| Product Number |
Source |
Storage |
Concentration for Use |
| F 3542 |
Recombinant |
-20°C |
0.45 µg/ml with 0.5 g/ml fibronectin |
Fibronectin, Lyophilized
| Product Number |
Source |
Storage |
Concentration for Use |
| F4759 |
Bovine plasma |
-0°C |
1-5 µg/cm2 |
| F2006 |
Human plasma |
-0°C |
1-5 µg/cm2 |
| F0635 |
Rat plasma |
-0°C |
1-5 µg/cm2 |
Fibronectin Cellular, Lyophilized
| Product Number |
Source |
Storage |
Concentration for Use |
| F2518 |
Human foreskin |
-0°C |
1-5 µg/cm2 |
Fibronectin Cellular, Aseptically Processed, Lyophilized
| Product Number |
Source |
Storage |
Concentration for Use |
| F6277 |
Human foreskin |
-0°C |
1-5 µg/cm2 |
Fibronectin-Like Engineered Protein Polymer
| Product Number |
Source |
Storage |
Concentration for Use |
| F5022 |
Recombinant |
Room temperature |
2-10 µg/cm2 |
Fibronectin-Like Engineered Protein Polymer-plus Genetically Engineered, Lyophilized, Autoclaved
| Product Number |
Source |
Storage |
Concentration for Use |
| F8141 |
Recombinant |
Room temperature |
2-10 µg/cm2 |
back to top
Gelatin
Used for attachment of a variety of cell types
Product Use
Gelatin 2% Solution, Approx. 225 Bloom, Autoclaved
| Product Number |
Source |
Storage |
Concentration for Use |
| G 1393 |
Bovine skin |
Store solubilized product at 2-8°C |
0.1-0.2 mg/cm2 (Solution: 5-10 µg/cm2) |
Gelatin, Approx. 225 Bloom
| Product Number |
Source |
Storage |
Concentration for Use |
| G 9391 |
Bovine skin |
Store powder at room temperature |
0.1-0.2 mg/cm2 (Solution: 5-10 µg/cm2) |
Gelatin, Approx. 300 Bloom
| Product Number |
Source |
Storage |
Concentration for Use |
| G 1890 |
Porcine skin |
Store powder at room temperature |
0.1-0.2 mg/cm2 (Solution: 5-10 µg/cm2) |
Gelatin, Approx. 300 Bloom, Lyophilized, Sterilized by g-irradiation
| Product Number |
Source |
Storage |
Concentration for Use |
| G 9136 |
Porcine skin |
Store powder at room temperature |
0.1-0.2 mg/cm2 (Solution: 5-10 µg/cm2) |
back to top
Laminin
Target cells for attachment:
- Epithelial cells
- Endothelial cells
- Muscle cells
- Tumor cells
- Hepatocytes
- Schwannoma
Product Use
| Product Number |
Source |
Storage |
Concentration for Use |
| L 2020 |
Basement membrane of Engelbreth-Holm-Swarm mouse sarcoma |
-20°C |
1-2 µg/cm2 |
| L 6274 |
Human placenta |
-20°C |
1-2 µg/cm2 |
back to top
Superfibronectin
Target cells for attachment:
- Epithelial cells
- Mesenchymal cells
- Neuronal cells
- Fibroblasts
- Neural crest cells
- Endothelial cells
| Product Number |
Source |
Storage |
Concentration for Use |
| S 5171 |
Human plasma and recombinant |
2-8°C |
1 µg/ml |
back to top
Thrombospondin, Lyophilized
Target cells for attachment:
- Osteoblasts
- Bovine aortic endothelial cells
- Neurons
- Human melanoma cells
Enhances proliferation of mitogen-stimulated smooth
muscle cells and fibroblasts
Product Use
| Product Number |
Source |
Storage |
Concentration for Use |
| T 7043 |
Human platelets |
-20°C |
25 ng-50 µg/ml |
back to top
Vitronectin
Cells with integrin receptors that bind vitronectin:
- Platelets
- Endothelial cells
- Melanoma cells
- Osteosarcoma
Product Use
| Product Number |
Source |
Storage |
Concentration for Use |
| V 8379 |
Human plasma |
2-8°C; store solubilized product at 2-8°C |
0.1 µg/cm2 |
| V 0132 |
Rat plasma |
2-8°C; store solubilized product at 2-8°C |
0.1 µg/cm2 |
| V 9881 |
Bovine plasma |
2-8°C; store solubilized product at 2-8°C |
0.1 µg/cm2 |
back to top
PRODUCT USE
Collagen Type I (Product Nos.
C 1809,
C 7661,
C 9791,
C 8919*)
Optimal conditions for attachment must be determined for each cell line and application.
- Add collagen to 0.1 M acetic acid to obtain 0.1% (w/v)
collagen solution. Allow to stir at room temperature 1-3 hours
until dissolved. Collagen solution (Product No. C 8919) should
be diluted 1:10 to obtain a working concentration of 0.01%.
- We recommend transferring the collagen solution to a glass
bottle with a screw cap and carefully layering chloroform at the
bottom. The amount of chloroform to use should be
approximately 10% of the volume of collagen solution. DO NOT
SHAKE OR STIR. Allow to sit overnight in the cold. Aseptically
remove the top layer containing your collagen solution. We do
not recommend sterilizing the collagen solution by membrane
filtration. We have found substantial protein loss by this method.
- Coat dishes with 6-10 µg/cm2. Allow the protein to bind for
several hours at room temperature, 37°C, or overnight at 2-8°C.
- Remove excess fluid from the coated surface, and allow it to
dry overnight. If the collagen solution is not sterile, the dried,
coated surface can be sterilized easily by overnight exposure to
UV light in a sterile tissue culture hood.
- Rinse with sterile tissue culture grade water or a balanced salt
solution before introducing cells and medium.
*Note: Steps 1 and 2 are not necessary for the Collagen Solution, Product No. C 8919.
back to top
Collagen Type II and Type IV (Product Nos.
C 9301,
C 0543,
C 5533)
Optimal conditions for attachment must be determined for each cell line and application.
- Collagen Types II and IV may be reconstituted to a
concentration of 0.5-2.0 mg/ml in 0.25% acetic acid. Dissolve
for several hours at 2-8°C, occasionally swirling.
- Coating of tissue culture plastic dishes may be performed by
air drying the above protein solution, or by preincubating the
same solution overnight at 2-8°C (or several hours at 37°C)
without air drying.
- Dried coated dishes can be sterilized overnight by exposure to
UV light in a sterile tissue culture hood or by rinsing with 70%
ethanol. Alternatively, the collagen solution may be sterilized by
dialysis in a 0.25% acetic acid and 0.5% chloroform solution.
back to top
Fibronectin (Product Nos.
F 1141*,
F 0895*,
F 4759,
F 2006,
F 0635)
Optimal conditions for attachment must be determined for each cell line and application.
- Reconstitute with 1 ml sterile H2O/mg of protein. Allow to
dissolve for at least 30 minutes at 37°C. A small amount of
undissolved material may remain. This will not affect product
performance.
- Dilute fibronectin in sterile balanced salt solution and coat the
culture surface with a minimal volume.
- Allow to air dry for at least 45 minutes at room temperature.
Excess fibronectin may be removed by aspiration, but is not
necessary.
*Note: Step 1 is not necessary for the Fibronectin Solutions, Product Nos. F 1141 and F 0895.
- Freezing and thawing of reconstituted fibronectin is not recommended as
breakdown of protein will occur.
back to top
Cellular Fibronectin (Product Nos.
F 2518,
F 6277)
Optimal conditions for attachment must be determined for each cell line and application.
- Reconstitute with 0.5 ml of sterile water. Do not agitate. Allow
the solution to stand 30 minutes to solubilize. A small amount of
undissolved material may remain. This will not affect product
performance.
- Dilute fibronectin in sterile balanced salt solution and coat the
culture surface with a minimal volume.
- Allow to air dry for at least 45 minutes at room temperature.
Excess fibronectin may be removed by aspiration, but is not
necessary.
back to top
Fibronectin-Like Engineered Protein Polymer
(Product No. F 5022)
Optimal conditions for attachment must be determined for each cell line and application.
The diluent and the stock solution contain 4.5 M LiClO4 which is
near saturation at room temperature. During shipping (diluent)
and storage (diluent and stock solution) it is likely that there will
be some recrystallization in the vials. Warming will return the
crystals into solution. Store tightly sealed at room temperature.
- Reconstitute the lyophilized product to a 1 mg/ml stock solution by combining with the
provided diluent. Swirl occasionally until dissolved.
- Dilute to working concentration with a balanced salt solution in the range of 1:10 to 1:50, depending upon the cell line and
application. The optimal range may be established by titer dilution.
- Working concentration range is 2-10 µg/cm2. If currently using
fibronectin, use the polymer at the same concentration. Some cell lines show equivalent
performance using from 2 to 5 times less protein polymer than fibronectin.
- Coat surfaces with 0.1 ml/cm2 of the working solution for 5 minutes at room temperature.
- Remove the coating solution and immediately rinse twice with a balanced salt solution or tissue
culture medium. Coated substrates can be used immediately or stored for up to 4 months
at room temperature.
back to top
Gelatin Solution, 2% (Product No.
G 1393)
Optimal conditions for attachment must be determined for each cell line and application.
- Allow gelatin solution to completely liquefy at 37°C.
- Coat culture surface with 5-10 µl gelatin solution/cm2
(i.e., 0.1-0.2 mg/cm2 gelatin).
- Allow to dry at least 2 hours before introducing cells and medium.
back to top
Gelatin (Product Nos.
G 9391,
G 1890,
G 9136)
Optimal conditions for attachment must be determined for each cell line and application.
- Prepare a 2% (w/v) solution by dissolving gelatin in tissue culture grade water.
- Sterilize by autoclaving at 121°C, 15 psi for 30 minutes.
- Coat culture surface with 5-10 µl gelatin solution/cm2
(i.e. 0.1-0.2 mg/cm2 gelatin).
- Allow to dry at least 2 hours before introducing cells and medium.
back to top
Laminin (Product Nos.
L 2020,
L 6274)
Optimal conditions for attachment must be determined for each cell line and application.
- Slowly thaw laminin at 2-8°C to avoid the formation of a gel.
- Dilute in a balanced salt solution and coat culture surface with a minimal volume.
- Allow to air dry at least 45 minutes before introducing cells and medium.
back to top
Thrombospondin (Product No.
T 7043)
Optimal conditions must be determined for each cell line and application.
- Reconstitute with 0.5 ml of tissue culture grade water and sterilize by filtration.
- The resulting solution is slightly hazy and has a concentration of 40 µg/ml.
- Use concentration for thrombospondin in culture has been reported in the range of
25 ng to 50 µg/ml depending upon the application.
- Please refer to the literature for more specific information.
back to top
Vitronectin (Product Nos.
V 8379,
V 0132,
V 9881)
Optimal conditions must be determined for each cell line and application.
- Reconstitute with tissue culture grade water and sterilize by filtration.
- Material is reported to be active at a concentration of 0.1 mg/cm2 of
surface area. Optimal concentrations vary with each cell line.
- Coat culture surface for 1-2 hours at 37°C. Remove any remaining solution and
wash with a balanced salt solution before introducing cells and medium.