Product Lines
Antibody Expression
Classic Media & Salts
Coated Plates
ECACC Cell Lines
Embryo Culture
Gene Therapy
Primary Human Culture
Protein Expression/CHO
Reagents & Supplements
Sera
Serum Free Media
Stem Cells
SyntheChol NS0
TrypZean Solution
Vaccine Production
Product Highlights
Key Resources
Cell Signaling and Neuroscience
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 REAGENTS AND SUPPLEMENTS
Cell Culture
Albumin/Transport Prtns
Amino Acids & Vitamins
Antibiotics
Attachment Factors
Biological Buffers
Biological Detergents
Cell Dissociation
Cell Freezing
Cell Separation
Cell Viability
Coated Plates
Growth Factors
Hormones
Hybridoma Reagents
ITS & Supplements
Lipids & Lipid Carriers
Misc. Reagents
Mycoplasma Detection
Mycoplasma Detection
LookOut™ Products for Mycoplasma Screening
Mynox® Mycoplasma Elimination Kit
Product List
LookOut™ Products for Mycoplasma Screening
Mycoplasma contamination in cell culture is a serious and wide-spread problem. Detection of contamination by classical culture methods can take weeks and often cell lines are discarded. due to ineffective antibiotics typically used in clean up.. Contamination can result in lost experimental data, time, and money. Sigma-Aldrich now offer the LookOut™ series of products developed for detection, surface elimination, and culture clean-up.
Product No. Product Name
Size
MP0035 LookOut™ Mycoplasma PCR Detection Kit
1 KT
MP0030 LookOut™ Mycoplasma Elimination Kit
1 KT
L8917 LookOut™ DNA Erase Spray Reagent
250 ml
L9042 LookOut™ DNA Erase Refill
1 L
LookOut™ Mycoplasma PCR Detection Kit (Product No. MP0035)
Product Description
The LookOut™ Mycoplasma PCR Detection Kit utilizes the polymerase chain reaction (PCR), which is established as the method of choice for highest sensitivity in the detection of Mycoplasma and Acholeplasma contamination in cell cultures and other cell culture derived biologicals. Detection requires as little as 1 to 5 fg of mycoplasma DNA corresponding to 10 mycoplasma particles per microliter of sample.
The primer set is specific to the highly conserved rRNA operon, or more specifically, the 16S rRNA coding region in the mycoplasma genome. This allows for detection of all Mycoplasma, Acholeplasma, and Ureaplasma species tested so far and usually encountered as contaminants in cell cultures. Eukaryotic and bacterial DNA are not amplified.
Only one protocol is needed for the detection of all mycoplasma species. Because the reaction tubes included with the kit are pre-coated with appropriate dNTPs and primers, the total assay time is greatly reduced compared to general protocols that require individual loading of reaction tubes. The reaction tubes also contain DNA to serve as an internal control. For the internal control DNA, a successfully performed reaction is indicated by a distinct 481 bp band on the agarose gel.
Components
  • Test Reaction Tubes, 3 strips of 8 tubes each
    (transparent tubes pre-coated with primers, dNTPs, and internal control DNA)
  • Positive Control Reaction Tubes, 1 strip of 8 tubes
    (pink tubes pre-coated with primers, dNTPs, internal control DNA, and non-infectious DNA fragments of Mycoplasma orale genome, prepared by PCR)
  • Mycoplasma PCR Rehydration Buffer, 1.2 ml
  • Caps for PCR Tubes, 4 strips of 8 caps
back to top
Equipment/Reagents required but not provided
  • JumpStart™ Taq DNA Polymerase, Product No.D9307
  • Microcentrifuge tubes, Product No. T0447
  • GenElute™ Blood Genomic DNA Kit, Product No. NA2000
    (optional, for DNA extraction and purification)
  • Gel Loading Solution, Product No. G7654
  • 1.2% Agarose Gel
Procedure
  1. Preparation of Sample Material
    Cell lines should be pre-cultured in the absence of mycoplasma active antibiotics for several days to maximize test sensitivity. Samples should be derived from cultures that are at 90-100% confluence. PCR inhibiting substances may accumulate in the medium of older cultures. For a sample from an older culture, an DNA extraction is strictly recommended prior to testing. The GenElute™ Blood Genomic DNA Kit (Product No. NA2000) is recommended.
  2. To avoid false positive results, the use of deionized, DNA-free water, aerosol-preventive filter tips, and gloves is recommended.
    Templates for PCR analysis are prepared by boiling the supernatant of cell cultures or other biologicals for 5 minutes as follows:
    1. Transfer 100 µl of supernatant from the test culture to a sterile microcentrifuge tube (Product No. T0447). The lid should be tightly sealed to prevent opening during heating.
    2. Boil or incubate the sample supernatant at 95 °C for 5 minutes.
    3. Briefly centrifuge (5 seconds) the sample supernatant to pellet cellular debris before adding to the PCR mixture. The templates are stable at 2–8 °C for at least 1 week.
  3. PCR Setup
    Total volume for each PCR is 25 µl. It is recommended to perform a positive (step 4) and a negative control reaction.
    1. Polymerase/Rehydration Buffer Preparation – Determine the total volume of Polymerase/ Rehydration Buffer required for the reactions (see Table 1). Calculations should also include an additional reaction volume (23 ml) to compensate for pipetting losses. One unit of DNA polymerase is required per reaction. For Jumpstart Taq DNA polymerase with 2.5 units/ml, a volume of 0.4 ml is required per reaction. Pipette the required volume of DNA polymerase into a clean microcentrifuge tube (Product No. T0447). Add Rehydration Buffer to bring the volume to the total required volume. Mix the Polymerase/Rehydration Buffer carefully by flicking the tube. DO NOT VORTEX!
      Table 1: PCR Schemes
        Test Reaction Positive Control Negative Control
      Polymerase/Rehydration Buffer 23 µl 25 µl 23 µl
      Sample Volume 2 µl    
      DNA-free Water     2 µl
    2. Reaction Tube rehydration - Remove strip of Test Reaction Tubes (transparent) from bag and cut off the appropriate number of tubes. Replace remaining tubes in bag and seal. Peel off protective film from tubes. Label tubes as appropriate. Add 23 ml of the prepared Polymerase/Rehydration Buffer to each test reaction tube.
    3. Sample addition - Add 2 ml of DNA-free water to the negative control and 2 ml of sample to each test reaction tube. Close tubes with Caps for PCR Tubes included in kit.
    4. Positive Control Preparation - Remove strip of Positive Control Reaction Tubes (pink) from bag and cut off the appropriate number of tubes. Replace remaining tubes in bag and seal. Peel off protective film from tubes. Label tubes as appropriate. Add 25 ml of of the prepared Polymerase/Rehydration Buffer to each tube. Close tubes with Caps for PCR Tubes included in kit.
    5. Incubation - Mix contents of each tube thoroughly by flicking tubes. DO NOT VORTEX! Incubate at room temperature for 5 minutes. Proceed immediately to thermal cycling.
  4. Thermal Cycler Profile
    The programming process of your cycler is explained in the instrument manual. Two programs, a normal thermal program (std program) and a short program, are possible depending on the heating speed of your cycler.
    1. The incubation time depends on the DNA polymerase used. Hot start enzymes need to be activated at 94 °C. Please see DNA polymerase data sheet for duration.
      Standard Program Short Program
      1 cycle 94 ° for 2 min
      55 ° for 2 min
      72 ° for 2 min
      1 cycle 94 ° for 2 min
      34 cycles 94 ° for 30 sec
      55 ° for 1 min
      72 ° for 1 min
      35 cycles 94 ° for 30 sec
      55 ° for 30 sec
      72 ° for 30 sec
      1 cycle 72 ° for 4 min
      cooldown
      to 4-8 °C
      cool down
      to 4-8 °C
  5. Agarose Gel
    1. Use 1.2 % standard agarose gel with 5 mm comb.
    2. Load 8 µl of each PCR reaction, mixed with Gel Loading Solution (Product No. G7654) per lane.
    3. Stop electrophoresis after migration of 3 cm (depending on the electrophoresis chamber used e.g., run for 20 minutes at 100 V).
Results
Gel Evaluation (Figure 1)
  1. The internal control DNA and negative control samples show a distinct 481 bp band. Internal controls should appear in every lane indicating a successfully performed PCR. This band may be less intense with increased amounts of amplicons formed, caused by mycoplasma DNA loads of >5 x 106 copies/ml.
  2. The positive control shows a band at 270 bp and depending on the activity of the DNA polymerase used, an additional band at 481 bp due to the internal control.
  3. Mycoplasma positive samples show bands in the range of 265–278 bp.
Figure 1: Relevant Amplicon Bands
back to top
Mynox® Mycoplasma Elimination Reagent
An innovative method for fast and efficient elimination of mycoplasmas in cell cultures and virus stocks. It’s time for efficient mycoplasma elimination!
Mycoplasma contamination of cell cultures is a serious and widespread problem. Contaminated cultures are frequently discarded or the contamination ignored since treatment with standard antibiotics is undesirable, time-consuming, and an ineffective means to eliminate mycoplasmas. Sigma-Aldrich now offers the Mynox® Mycoplasma Elimination reagent with the following outstanding characteristics:
  • Kills mycoplasmas, but safe for cells
  • Effective with only one treatment
  • Not an antibiotic
  • Effective against most mycoplasma species
  • Applicable for most cell and virus types
  • Ready-to-use solution
Contents
  • Mynox® is packaged in a premeasured sterile, single-use tube.
Requirements
  • Standard cell culture equipment
  • Mycoplasma detection system to verify the elimination success
Mynox® and VenorGeM® are registered trademarks of Minerva Biolabs, Germany.
Available only in North and South America. Mynox® and VenorGem® are intended for research use only. Not recommended for clinical treatment.
back to top
Additional Products for Mycoplasma Screening
Detection  
Product Name Product # 
Bisbenzimide H 33258, cell culture, minimum 98%, Powder B1155
4',6-Diamidino-2-phenylindole dihydrochloride, cell culture, ³98%, Powder D8417
Hanks' Balanced Salt Solutions H5899
Hoechst Stain solution H6024
Mounting Medium M1289
Mycoplasma Agar, Microbiologically, Powder M0660
Mycoplasma Broth, Microbiologically, Powder M0535
Mycoplasma Control Slides M1414
Mycoplasma Stain Kit MYC-1
back to top