Product Lines
Antibody Expression
Classic Media & Salts
Coated Plates
ECACC Cell Lines
Embryo Culture
Gene Therapy
Primary Human Culture
Protein Expression/CHO
Reagents & Supplements
Sera
Serum Free Media
Stem Cells
SyntheChol NS0
TrypZean Solution
Vaccine Production
Product Highlights
Key Resources
Cell Signaling and Neuroscience
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 TiterHigh™ Sf Insect Medium
Cell Culture
Insect Cell Media :: Formulation :: TiterHigh™ Sf Insect Medium
TiterHigh Sf Insect Medium, Animal Component-Free (I 5408) is the latest addition to the fastest growing line of serum-free media in the industry.
With TiterHigh Sf Insect Medium You Get:
  • Exceptional cell growth - viable cell densities of up to 2 x 107 cells/mL can be achieved in shaker cultures, with little or no adaptation required
  • Excellent baculoviral expression - beats the competition in terms of per cell productivity with wild type and recombinant baculoviruses
  • No serum or any animal-derived components - satisfies regulatory requirements and simplifies downstream processing
Sƒ21 cells were grown in TiterHigh Sf Insect Medium and the leading copetitor's medium in shaker flasks, at 27° C with a shaker speed of 130 rpm. As is shown, TiterHigh Sf supports much higher viable cell densities than the competition.
Sƒ21 cells grown in TiterHigh Sf Insect Medium and the leading competitor's medium were infected with a recombinant baculovirus expressing b-galactosidase. Cells were infected at a cell concentration of 1x106 cells per mL, with a multiplicity of infection of 5 virus particles per cell. Enzyme levels were measured at days 2, 3, and 4 post-infection. As is demonstrated, TiterHigh Sf beats the competition in terms of per cell baculovirus productivity.
Product Code Description Package Size Price US $
I 5408 TiterHigh Sf Insect Medium 1 L
6 X 1 L
53.45
240.55
Product Code Description Package Size Price US $
L 3287 ESCORT™ IV Transfection Reagent 1 mL 86.05
E 9770 ESCORT™ Transfection Reagent .25 mL
.5 mL
44.35
78.95
G 1397 Gentamicin Solution 10 mL
100 mL
41.00
340.35
back to top
Cell Culture Home