Product Lines
Antibody Expression
Classic Media & Salts
Coated Plates
ECACC Cell Lines
Embryo Culture
Gene Therapy
Primary Human Culture
Protein Expression/CHO
Reagents & Supplements
Sera
Serum Free Media
Stem Cells
SyntheChol NS0
TrypZean Solution
Vaccine Production
Product Highlights
Key Resources
Cell Signaling and Neuroscience
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 TrypZean™ Solution
Cell Culture
Non-animal recombinant trypsin solution for dissociating attached cells
TrypZean Solution, 1X (Product Code T 3449) is the first animal component-free trypsin solution optimized for cell dissociation. It is formulated with TrypZean, a recombinant bovine trypsin expressed in corn and manufactured by Sigma-Aldrich utilizing ProdiGene’s proprietary transgenic plant protein expression system.
Percent Viability
TrypZean Solution, 1X provides the following features and benefits:
  • Animal component-free
    TrypZean eliminates the risk of viruses, BSE, or other potential adventitious agents.
  • TrypZean is recombinant trypsin
    Using the same enzyme (with the same kinetics) for cell detachment means minimal protocol changes for you.
  • Enzyme inhibition
    Soybean trypsin inhibitor and other inhibitors work the same with TrypZean as they do with native trypsins (on a weight-to-weight basis).
  • High purity
    TrypZean provides increased specificity and eliminates contaminating activities found in lower purity enzymes.
  • Convenience
    TrypZean Solution is formulated at the optimal concentration to dissociate adherent cells.
Kinetics of TrypZean
Trypsin vs. TrypZean
Product Name
Product # 
Package Size
TrypZean™ Solution, 1X
T 3449
100 ml
500 ml
back to top
Back to Cell Culture Home