Sigma is proud to offer a selection of media, many of which are serum-free, for the in vitro culture of human primary cells, such as keratinocytes, endothelial cells, and epithelial hepatocytes. These primary cell types have potential commercial value in
a wide range of emerging technologies and therapeutic applications. Among these are: tissue
engineering applications, such as wound healing and burn treatment; high throughput screening for drug toxicology and efficacy studies; gene therapy; pseudo-organs (e.g.. artificial liver) and as model cell systems for developing new therapeutic approaches to human diseases.
Keratinocytes, EpiLife™
Keratinocyte Cell Culture Media
Sigma proudly offers a new "formulation break-through" serum-free medium, EpiLife™
Extended-Lifespan medium, that greatly extends the lifespan of cultured human keratinocytes and corneal
epithelial cells when compared to other commercial serum-free media. EpiLife™
Extended-Lifespan medium is the only commercially available serum-free medium which is able to substantially
extend the replicating lifespan of epithelial cells such as keratinocytes and corneal epithelial cells to
40 or more population doublings.
The following tables provide details to allow you to select the epithelial cell culture products that
are most appropriate to your needs. You can go directly to the topic of greatest interest by using
the hyperlinks at the top of this page.
For more information about keratinocyte epidermal cells please click this link:
Keratinocyte Cell Culture Media
Background
| |
|
|
back to top
| | Product #
| Product Name |
Description |
Application |
|
E 0151 |
EpiLife™ Extended-LifespanSerum-Free
Medium |
- Liquid, basal medium
- Proprietary formulation
- Contains essential and non-essential amino acids, vitamins, trace minerals, organic and inorganic salts.
- Without antibiotics, antimycotics, hormones, growth factors, or proteins.
- Formulated with 0.06 mM calcium chloride.
- Required growth supplement provided separately
- Buffered with HEPES and sodium bicarbonate, equilibrates in 5% carbon dioxide to pH = 7.4.
- Sterile filtered
- Endotoxin tested
- Cell culture tested.
| This
basal medium, when appropriately supplemented, will support the proliferation of normal human keratinocytes.
All of the growth factors, hormones and extracts necessary for the growth of
human keratinocytes are provided separately The basal medium may be supplemented with either
Keratinocyte Medium Supplement (K 2007), or Keratinocyte Medium Supplement, BPE-free (K 3136).
K 3136 should be used when a chemically defined culture system is a requirement.
When fully supplemented, this proprietary cell culturing system can dramatically extend the life
span of actively dividing human keratinocyte cells.
Keratinocytes grown in the EpiLife™ Extended-Lifespan culture systems will successfully complete
from 35 to 45 doublings. | |
K 4131 |
Keratinocyte Basal Medium |
- Liquid
- Contains essential and non-essential amino acids, vitamins, trace minerals, organic and
inorganic salts.
- Without antibiotics, antimycotics, hormones, growth factors, or proteins.
- Required growth supplements are provided separately
- Buffered with HEPES and sodium bicarbonate
- Sterile filtered
- Endotoxin tested
- Cell culture tested.
|
A component of Keratinocyte Medium Kit (KMK-2). This
basal medium, when appropriately supplemented with Keratinocyte Medium Supplement
(K 2007), will support the growth of normal human keratinocytes.
Since K 2007 contains bovine pituitary extract (BPE), it
is not fully defined. However, BPE effectively supports the proliferation of human keratinocytes.
|
| EpiLife™ is a trademark of Cascade Biologics, Inc.,
Portland, OR. |
back to top |
Keratinocyte Cell Culture Companion Products and Supplements
The serum free culture of keratinocyte cells is supported by
a series of products offered by Sigma. The table below provides a partial list of Sigma products
recommended for this purpose. Also, see the "Keratinocyte Cell Culture Media" table above
and the "EpiLife™ Extended-Lifespan Keratinocyte Culture Systems" table below for more
detailed information.
| |
|
|
back to top
| |
Product # |
Product Name |
Description |
Application |
|
K 2007 |
Keratinocyte Medium Supplement (100x)
|
- Liquid
- Contains bovine pituitary extract (BPE), bovine insulin, hydrocortisone, bovine transferrin, and
human recombinant EGF.
- Sterile filtered
- Endotoxin tested
- Cell culture tested.
|
This supplement is a component of Keratinocyte Proliferation Kit
(KMK-2). It is designed to be used in conjunction with Keratinocyte Basal
Medium (K 4131) or EpiLife™ serum-free medium (E 0151)
Since it contains BPE, it is not fully defined and hence, should only be used
when this is not a concern. This supplement is a component of
a unique proprietary cell culturing system that dramatically extends the life span of actively dividing
human keratinocyte cells. Keratinocytes grown in the EpiLife™
culture systems will successfully complete from 35 to 45 doublings. |
|
K 3136 |
Keratinocyte Medium Supplement,
BPE-free |
- Liquid
- Contains recombinant growth factors, hydrocortisone, bovine serum albumin and
bovine transferrin
- Sterile filtered
- Endotoxin tested
- Cell culture
tested.
| This supplement
is designed to be used in conjunction with, EpiLife™ Keratinocyte medium (E 0151).
Since it does not contain BPE, the culture system is chemically defined. It
should be used when a chemically defined cell culturing system is a requirement.
This supplement is a component of a unique proprietary cell culturing
system that dramatically extends the life span of actively dividing human keratinocyte cells.
Keratinocytes grown in the EpiLife™ culture systems will successfully
complete from 35 to 45 doublings. |
|
K 3276 |
EpiLife™ Cell Freezing Medium |
- Liquid
- Animal-component free
- Ready-to-use formula containing EpiLife™ medium, dimethyl
sulfoxide (DMSO) and substituted methylcellulose
- Sterile filtered
- Endotoxin tested
- Cell culture tested.
|
Useful for the cryopreservation of
keratinocytes and other epithelial derived cell types. The absence
of animal-derived components in this medium reduces the potential for contamination of cryopreserved
cells with adventitious biological agents such as viruses. |
|
T 4299 |
Trypsin-EDTA Solution for Adherent Cell Culture
|
- Liquid
- Contains 500 BAEE units porcine trypsin and 180 ug of EDTA74Na per mL in DPBS without calcium
and magnesium
- Powder negative for porcine parvovirus by 9 CFR
- Sterile filtered
- Mycoplasma tested
- Cell culture tested
|
Useful for the dissociation of, or release of,
adherent cells types, such as endothelial, epithelial, and fibroblast cells from support surfaces.
| |
T 7659 |
Trypsin Inhibitor Defined |
- Liquid
- Phosphate buffered saline solution containing (1x) 0.025% soybean trypsin inhibitor
- pH approximately 7.4 at room temperature
- Sterile filtered
|
Use to inhibit the proteolytic
action of Trypsin-EDTA (T 4299) after adherent cells, such as epithelial cells,
are released from a substrate or dissociated.
Chemically defined and
free of animal derived components. The absence of animal derived components reduces the
potential for contamination of cells with adventitious biological agents such as viruses.
| | |
|
|
back to top
|
EpiLife™Extended-Lifespan
Keratinocyte Culture Systems
The following table provides a guide for selection of the appropriate components to grow keratinocytes under
expanded replicating lifespan conditions in both defined and undefined culture systems.
|
For Keratinocytes |
|
Undefined |
Defined |
| Basal Medium
|
EpiLife™ E 0151
|
EpiLife™ E 0151
| |
Growth Supplement |
Keratinocyte Medium
Supplement (100x) K 2007 |
Keratinocyte
Medium Supplement, BPE-free K 3136
|
Subculture Reagent-1 Cell Release |
Trypsin Inhibitor Chemically Defined
T 7659 |
Trypsin Inhibitor Chemically Defined
T 7659 |
Subculture
Reagent-2 Trypsin Inhibitor |
Trypsin-EDTA
Solution T 4299 |
Trypsin-EDTA
Solution T 4299 |
| EpiLife™ is a trademark of Cascade Biologics, Inc.,
Portland, OR. |
back to top |
Epidermal
(Skin) Cell Background Information
Skin is composed of an external epithelial component called the epidermis which is separated
from an underlying connective tissue component, the dermis, by a basal membrane. The
portion of the dermis adjacent to the epidermis is called the reticular dermis and is composed
primarily of collagen fibers produced by fibroblasts, micro vessels and a few migrating
leukocytes. The reticular dermis supplies all of the nutrition to the epidermis which is
devoid of blood vessels. The great majority of cells in the epidermis are keratinocytes,
which are arranged in stratified layers. At the dermal-epidermal junction is a single layer
of keratinocytes with small interspersed melanocytes (ratio of approximately 1:30) called the
stratum basale. This basal layer of keratinocytes is also called the
stratum germinativium, because it is where new keratinocytes are generated by cell proliferation.
Three types of keratinocytes in the stratum basale have been
defined by kinetic analysis: stem cells; transient-amplifying cells and committed cells. Stem cells,
which represent approximately 10% of the basal cell population, generate daughter cells from mitosis that
are either stem cells themselves or transient-amplifying cells. Transient-amplifying cells, which
represent approximately 40% of the basal cell population, replicate with much higher frequency than stem
cells, but are capable of only a few population doublings. Transient-amplifying cells produce
daughter cells that are committed to terminally differentiate. Committed cells detach from the
basement membrane, differentiate, and ultimately cease to proliferate as they migrate toward the skin
surface where they are sloughed off as dead cornified cells called squames.
back to top
Keratinocyte
Cell Culture Media Background
It has been approximately 25 years since
replicating in vitro cultures of human epidermal keratinocytes were first established.
The basal medium used for these first studies was developed using immortalized cells lines and
initial success in establishing replicating epidermal keratinocyte cultures relied on the use of
fibroblast feeder layers and on the inclusion of animal serum in the culture medium, both for
establishing primary cultures and for subcultures. Using these culture conditions, keratinocytes
could be sub-cultured until they achieved 20-50 population doublings. By modifying the
culture conditions, many more population doublings could be obtained from epidermal keratinocytes
in vitro, although feeder layers and serum were still required. While many investigators
still use fibroblast feeder layers and animal sera for keratinocyte culture, it is generally considered
desirable to remove these ill-defined components from cell culture systems, particularly those intended
for therapeutic applications.
Richard Ham and his colleagues were the first to develop an in vitro system that did not require serum or
a feeder layer to support clonal growth, serial propagation, or differentiation of epidermal keratinocytes.
This system was the basis for the development of current commercialmedia that do not rely on the use of
serum or feeder layers. In this class, Sigma offers the keratinocyte serum-free medium kit, KMK-2,
which is a multi-component kit composed of basal medium, K 4131 and medium supplement, K 2007.
While serum-free media systems such as KMK-2 are very useful for growing and studying primary
keratinocytes, they have one substantial limitation. They are typically only able to support 15
to 20 population doublings after primary culture.
Since replicating cultures of keratinocytes were first established, several applications for these cells have
been investigated. Cultured human keratinocytes are now being used for in vitro toxicity assays, which
require that large standardized banks of these cells
be available, for autografting and allografting to aid in would healing, and for investigations of the
feasibility of ex vivo gene therapy approaches. Unfortunately, the development of many of these
applications has been hindered because of the limited in vitro replicating lifespan of keratinocytes in the
available commercial media.
EpiLife™ keratinocyte medium potentially
solves this problem by enabling the extension of the replicating lifespan of keratinocytes to 40
population doublings or more in vitro.
back to top |