|
// We only want a grey bar if c3_headline has a value...
$headline="Stemline Media and Cytokines";
if ($headline) {
print "| ";
} else {
print " | ";
}
?>
Stemline Media and Cytokines |
|
The use of human hematopoietic stem cells (HSC) isolated from umbilical cord blood as a source of cellular reconstitution following high-dose chemotherapy is now a common therapeutic modality for the treatment of malignancy. However, due to the low yield of HSC from cord blood, HSC transfusion is most effective in children and has limited application in adults. In order to obtain sufficient numbers of cells for applications of this therapeutic approach in adults, ex vivo expansion has been utilized to ensure successful engraftment and minimize the short-term effects of neutropenia and thrombocytopenia. To this end, we have developed a serum-free, animal protein-free medium, Stemline Hematopoietic Stem Cell Expansion Medium for the optimal expansion of cord blood HSC.
Stemline Hematopoietic Stem Cell Expansion Medium has been developed to promote growth of CD34+ hematopoietic stem cells derived from bone marrow, cord blood or mobilized peripheral blood. This medium supports high viable cell densities. The elimination of serum reduces performance variability in the medium and eliminates safety risks associated with possible adventitious agents in serum.
back to top
|
| Prod. # |
Product Name |
Description |
Application |
|
S 0189 |
Stemline Hematopoietic Stem Cell Expansion Medium |
Liquid
Sterile filtered
The medium does not contain antibiotics or cytokines. Medium must be supplemented with desired cytokines and/or antibiotics. Human serum albumin is the only human origin material. No other animal-derived components. Cell culture tested with normal human bone marrow CD34+ cells in a 14-day culture expansion. Medium was supplemented with 100 ng/ml each of SCF (Stem Cell Factor,
S7901), G-CSF (Granulocyte-Colony Stimulating Factor,
G0407) and TPO (Thrombopoietin,
T1568) for cell expansions.
|
Stemline HSC Expansion Medium shows excellent expansion of total nucleated cells (TNC), committed progenitors (GM-CFC) and primitive, high proliferative potential progenitors (HPP-CFC).
For clinical-scale studies, cord blood CD34+ cells were cultured for 7 days in 100 ml Teflon® culture bags containing 50 ml of culture medium, plus cytokines. Cells were harvested from these bags and a 5 ml aliquot was transferred to a second 100 ml Teflon® bag containing 50 ml of selected medium, plus cytokines, and cultured for an additional 7-day period. At the end of both culture protocols, cells were harvested and assayed for their ability to promote the growth of cord blood CD34+ cells. The data are summarized in this
Data Sheet.
|
| back to top |
|
Methylcellulose-supplemented Iscove's Modified Dulbecco's Medium (IMDM) forms a semi-solid growth
medium that, when supplemented with cytokines, supports the growth
and differentiation of hematopoietic progenitor cells and allows the clonal
progeny of a single cell to form a distinct cluster or colony. Methylcellulose
based, semi-solid culture media are routinely used for the
enumeration and evaluation of stem cell-derived progeny which typically manifest
as Colony Forming Units (CFU) or Burst Forming Units (BFU).
The Stemline Methylcellulose
Media family of products are manufactured under tightly controlled conditions using rigorously-qualified raw materials to ensure reliable and predictable results from the culture of human and murine hematopoietic cells. This group of
products will support the growth of granulocyte colonies (CFU-G), macrophage colonies (CFU-M), granulocyte/macrophage colonies (CFU-GM), erythroid colonies (BFU-E
and CFU-E), and pluripotent mixed colonies (CFU-GEMM).
back to top
|
| Prod. # |
Product Name |
Description |
Application |
|
S 3562 |
Stemline Complete Methylcellulose Medium with Erythropoietin, Human |
Liquid, frozen
Aseptically processed
Cell culture tested in a 12 to 16 day assay, using normal human CD 34 positive progenitor cells.
|
Use for culture of human hematopoietic cells.
Supports the following colony types: CFU-G; CFU-M; CFU-GM; BFU-E; CFU-E and CFU-GEMM.
Recommended for LTC-IC assays of purified populations of very primitive human
clonogenic, hematopoietic cells. |
|
S 3687 |
Stemline Complete Methylcellulose Medium without Erythropoietin, Human |
Liquid, frozen
Aseptically processed
Cell culture tested in a 12 to 16 day assay, using normal human CD 34 positive progenitor cells.
|
Same as S 3562, except that erythropoietin has not been added.
Use for culture of human hematopoietic cells.
Supports the following colony types: CFU-G; CFU-M; and CFU-GM.
Recommended for LTC-IC assays of purified populations of very primitive human clonogenic, hematopoietic cells.
|
| S 3812 |
Stemline Complete Methylcellulose Medium with Erythropoietin, Mouse |
Liquid, frozen
Aseptically processed
Cell culture tested in a 7 to 10 day CFU assay, using normal mouse bone marrow cells
|
Use for culture of mouse hematopoietic cells.
Supports the following colony types: CFU-G; CFU-M; CFU-GM; BFU-E; CFU-E and CFU-GEMM.
Recommended for assay of murine clonogenic hematopoietic progenitor cells isolated from murine bone
marrow and other murine hematopoietic tissues.
|
| S 3937 |
Stemline Methylcellulose Medium, without Growth Factors, Human |
Liquid, frozen
Without erythropoietin
Aseptically processed
Cell culture tested in a 12 to 16 day CFU assay using normal human CD 34 positive progenitor cells
|
Use for culture of human hematopoietic cells.
Supports the following colony types: CFU-G; CFU-M; CFU-GM; BFU-E; CFU-E and CFU-GEMM, only if supplemented with appropriate growth factors.
Recommended for assay of human clonogenic, hematopoietic progenitor cells from human blood and
bone marrow. It allows the researcher to add growth factors or cytokines
of their
choice.
|
| S 4062 |
Stemline Methylcellulose Medium, without Growth Factors, Mouse |
Liquid, frozen
Without erythropoietin
Aseptically processed
Cell culture tested in a 7 to 10 day CFU assay, using normal mouse bone marrow cells.
|
Use for culture of mouse hematopoietic cells.
Supports the following colony types: CFU-G; CFU-M; CFU-GM; BFU-E; CFU-E and CFU-GEMM, only if supplemented with appropriate growth factors.
Recommended for assay of murine clonogenic hematopoietic progenitor cells isolated from murine bone
marrow and other hematopoietic tissues. It allows the researcher to add
growth factors of their choice.
|
| back to top |
|
The
following table lists a group of cytokines whose predominant actions involve the stimulation or regulation of hematopoietic cells.
The term "colony stimulating factor" (CSF) was originally given to agents recognized to stimulate the growth of colonies containing differentiated myeloid cells from a single bone marrow-derived precursor cell plated in semi-solid agar. Glycoproteins considered to be CSFs include: granulocyte colony-stimulating factor (G-CSF); granulocyte-macrophage colony-stimulating factor (GM-CSF); macrophage colony-stimulating factor (M-CSF); interleukin-3
(multi-CSF or IL-3); interleukin-5 (IL-5); erythropoietin (EPO), and thrombopoietin (TPO).
The formation and maturation of hematopoietic cells are also affected by cytokines of the structural class
know as the "4-alpha-helical bundle" family. These include
stem cell factor (SCF) and flt-3/flk-2 Ligand (FL). |
| back to top |
| Prod. # |
Description |
Source |
EC50 Value |
Quality Assurance |
Stimulated Cells |
|
E5627 |
Erythropoietin (EPO) |
Human; recombinant host: CHO cells |
0.05 to 0.1 units/mL |
10 units
Endotoxin tested Proliferative activity tested using the human cell line TF-1.
|
Erythroid progenitor cells.
EPO receptors are lost during cell differentiation.
|
|
F3422 |
FLT-3/FLK-2 Ligand |
Human; recombinant host: NSO mouse cells |
0.5 to 1.0 ng/mL |
5 µg >
97% pure by (SDS-PAGE) Endotoxin tested Biological activity measured by its ability to stimulate the proliferation of an Flt3 transfected pro-B cell line.
|
Primitive hematopoietic (multi-lineage) progenitor cells that express the flt3 receptor. Myeloid and pro-B cells
of mouse, rat, cat, rabbit, primate and human origin.
|
|
G0407 |
Granulocyte Colony-Stimulating Factor (G-CSF) |
Human; recombinant host: Escherichia coli |
0.01 to 0.1 ng/mL |
2 µg >
95% pure by (SDS-PAGE) and N-terminal analysis. 18.8 kDa monomer; 175 amino acids; N-termianl methionine. Endotoxin tested Proliferative activity tested using the
cytokine-dependent murine NFS-60 cell line.
|
Proliferation, differentiation, survival and activation factor for hematopoietic restricted granulocyte lineage cells.
Neutrophils, myeloid leukemic cells, neutrophilic granulocytes from bone marrow and placental trophoblasts.
|
|
G8160 |
Granulocyte Colony-Stimulating Factor (G-CSF) |
Mouse; recombinant host: Escherichia coli |
0.005 to 0.05 ng/mL |
5 µg >
97% pure by (SDS-PAGE) 19 kDa monomer; 179 amino acids; N-terminal methionine. Endotoxin tested
Proliferative activity tested using the cytokine-dependent murine NFS-60 cell line.
|
Proliferation, differentiation, survival and activation factor for hematopoietic restricted granulocyte lineage cells.
Neutrophils, myeloid leukemic cells, neutrophilic granulocytes from bone marrow and placental trophoblasts.
|
|
G5035 |
Granulocyte-Macrophage Colony-Stimulating Factor (GM-CSF) |
Human; recombinant host: Escherichia coli |
0.02 to 0.2 ng/mL |
5 µg >
97% pure by (SDS-PAGE) 14 kDa monomer; 127 amino acids; N-terminal methionine.
Endotoxin tested Proliferative activity tested using human TF-1 cells
|
Growth, differentiation, and essential survival factor for granulocyte, macrophage and eosinophil lineage
cells from progenitor stage to maturity. Receptors on most types of myeloid progenitor cells, mature monocytes, neutrophils, eosinophils,
basiophils, and dendritic cells. Stimulates some epithelial cells and osteoclasts.
|
|
G0282 |
Granulocyte-Macrophage
Colony-Stimulating Factor (GM-CSF) |
Mouse; recombinant host: Escherichia coli |
0.05 to 0.5 ng/mL |
5 µg >
95% pure by (SDS-PAGE) and N-terminal analysis. 14.8 kDa monomer; 125 amino acids; N-terminal methionine. Endotoxin tested
Proliferative activity tested using murine DA-3 cells
|
Growth, differentiation, and essential survival factor for granulocyte, macrophage and eosinophil lineage
cells from progenitor stage to maturity. Receptors on most types of myeloid progenitor cells, mature monocytes, neutrophils, eosinophils,
basiophils, and dendritic cells. Stimulates some epithelial cells and osteoclasts.
|
|
I1646 |
Interleukin-3 (IL-3) |
Human; recombinant host: Escherichia
coli |
0.04 to 0.4 ng/mL |
10 µg >
97% pure by (SDS-PAGE)
Endotoxin tested
Proliferative activity tested using human TF-1 cells
|
Multifunctional cytokine.
Supports early development of multi-potential hematopoietic progenitor cells: macrophage, neutrophils, mast cells and megakaryocytes from
bone marrow. Stimulate T-cells and induce IgG secretion from activated B-cells.
|
|
I4144 |
Interleukin-3 (IL-3) |
Mouse; recombinant
host: Escherichia coli |
0.025 to 0.10 ng/mL |
10
µg >
97% pure by (SDS-PAGE)
Endotoxin tested Proliferative activity tested using murine NFS-60 cells
|
Multifunctional cytokine.
Supports early development of multi-potential hematopoietic progenitor cells: macrophage, neutrophils, mast cells and megakaryocytes from
bone marrow. Stimulate T-cells and induce IgG secretion from activated B-cells.
|
| I1395 |
Interleukin-6 (IL-6) |
Human; recombinant host: Escherichia coli |
0.4 to 2.0 ng/mL |
10 µg >
97% pure by (SDS-PAGE)
Endotoxin tested Proliferative activity tested using the mouse plasmacytoma cell line T1165.85.2.1.
|
Multifunctional cytokine which acts on a wide range of cell types. In combination with other cytokines, it acts upon; fibroblasts, myeloid progenitor cells, T-cells, B-cells, and hepatocytes.
|
|
I9646 |
Interleukin-6 (IL-6) |
Mouse; recombinant host: Escherichia coli |
0.03 to 0.30 ng/mL |
5 µg > 97% pure by (SDS-PAGE)
Endotoxin tested Proliferative activity tested using the mouse plasmacytoma cell line T1165.85.2.1.
|
Multifunctional cytokine which acts on a wide range of cell types.
In combination with other cytokines, it acts upon; fibroblasts, myeloid progenitor cells, T-cells, B-cells, and hepatocytes.
|
|
M6518 |
Macrophage Colony-Stimulating Factor (M-CSF) |
Human |
0.1 to 3.0 ng/mL |
5 µg >
97% pure by (SDS-PAGE) and N-terminal analysis., 37 kDa homodimer; each monomer contains 159 amino acids including the
N-terminal methionine. Endotoxin tested Proliferative activity tested using cytokine-dependent monocytic M-NFS-60 cells.
|
Primarily targets
macrophages and stimulates multiple responses, including; proliferation, cytokine and inflammatory modulator release, cytotoxicity
and pinocytosis. Also, supports osteoclast differentiation and may act on placental trophoblasts.
|
|
M9170 |
Macrophage Colony-Stimulating Factor (M-CSF) |
Mouse
|
0.3 to 3.0 ng/mL |
5
µg >
97% pure by (SDS-PAGE) 52 kDa homodimer; each monomer contains 231 amino acids including the
N-terminal methionine. Endotoxin tested Proliferative activity tested using cytokine-dependent
monocytic M-NFS-60 cells.
|
Primarily targets
macrophages and stimulates multiple responses, including; proliferation, cytokine and inflammatory modulator release, cytotoxicity
and pinocytosis. Also, supports osteoclast differentiation and may act on placental trophoblasts.
|
|
S7901 |
Stem Cell Factor (SCF) |
Human; recombinant host: Escherichia coli |
2.0 to 5.0 ng/mL |
10
µg >
97% pure by (SDS-PAGE)
Endotoxin tested Proliferative activity tested using TF-1 cells.
|
Broad activities involving
hematopoietic, pigment and primordial germ cell lineages. A potent synergistic co-stimulating growth factor which increase the size, but
not the differential, of myeloid, erythroid, and lymphoid lineage colonies.
|
|
T4309 |
Thrombopoietin (TPO) |
Human; recombinant host: NSO cells |
0.4 to 4.0 ng/mL |
5
µg >
97% pure by (SDS-PAGE) Predicted 35 kDa from sequence 332 amino acids: glycosylated recombinant product has apparent MW of 82 kDa by (SDS-PAGE).
Endotoxin tested Proliferative activity tested using MO7e cells.
|
Primary regulatory factor for megakaryocytopoiesis and thrombopoiesis. Regulates the growth and maturation of megakaryocytes and their progenitors.
|
|
T4184 |
Thrombopoietin (TPO) |
Mouse; recombinant host: NSO cells |
0.4 to 4.0 ng/mL |
5 µg >
97% pure by (SDS-PAGE) Predicted 35 kDa from sequence 335 amino acids: glycosylated recombinant product has apparent MW of
75 kDa by (SDS-PAGE).
Endotoxin tested Proliferative activity tested using MO7e cells.
|
Primary regulatory factor for megakaryocytopoiesis and thrombopoiesis. Regulates the growth and maturation of megakaryocytes and their progenitors.
|
| back to top |
|
© 2003 Sigma-Aldrich Co. Reproduction forbidden without permission.
Sigma-Aldrich brand products are sold exclusively through Sigma-Aldrich, Inc. Best viewed in
IE5 or higher