Product Highlights
New Literature
Ion Channels
Key Resources
Cell Signaling Web Tools
New Products
PathFinder
Pathway Slides & Charts
Pharm. Action Table
Poster Gallery
Protocols
Search Tools
eHandbook
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
Caspase Activation Intrinsic Pathway
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Caspase Activation Intrinsic Pathway
Cytochrome c is released from the mitochondria of pre-apoptotic cells and binds to Apaf-1 in the presence of dATP/ATP. The interaction results in a conformational change in Apaf-1 allowing the molecules of Apaf-1 to associate with each other. This formation results in a wheel-like structure that contains 7 molecules each of Apaf-1, cytochrome c and ATP. Pro-Caspase-9 contains a caspase recruitment domain (CARD) that is used to mediate specific interactions with Apaf-1 CARD, which becomes exposed on the apoptosome during assembly. Pro-Caspase-9 is autoactivated and after cleavage, mature caspase-9 remains bound to the apoptosome where it is able to activate executioner caspases such as caspase-3 and caspase-7.
Back to Top