Automation
Biological Buffers
Cancer Research
Cell Culture
Cell Signaling and Neuroscience
Product Lines
Product Highlights
New Literature
Ion Channels
Key Resources
Cell Signaling Web Tools
New Products
PathFinder
Pathway Slides & Charts
Pharm. Action Table
Poster Gallery
Protocols
Search Tools
eHandbook
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
Formation of Activated 20S Proteasome
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Formation of Activated 20S Proteasome
Intracellular Proteolytic Systems Recognize and Destroy misfolded or damaged proteins, unassembled polypeptide chains, and short-lived regulatory proteins. There are a number of mechanisms of protein degradation within cells. Two systems that play an important role in proteolysis resulting from cell stress are calpain proteases and the ubiquitin-proteasome pathway.
The calpains are a family of heterodimeric, calcium-activated cysteine proteases. They are composed of a common 30 kDa subunit and an isoform specific, catalytic 80 kDa subunit. The large subunit of Calpain I m-isoform) is activated by micromolar calcium levels, while Calpain II (m-isoform) requires millimolar levels of calcium for activation. Cell stress or injury can lead to sustained elevation of intracellular calcium levels, causing sustained activation of calpains. The common, late-stage of the cell death pathway induced by excitotoxic compounds in the nervous system involves calpain-mediated proteolysis. The ubiquitin-proteasome pathway functions widely in intracellular protein turnover. It plays a central role in degradation of short-lived and regulatory proteins important in a variety of basic cellular processes, including regulation of the cell cycle, modulation of cell surface receptors and ion channels, and antigen processing and presentation. The pathway employs an enzymatic cascade by which multiple ubiquitin molecules are covalently attached to the protein substrate. The polyubiquitin modification marks the protein for destruction and directs it to the 26S proteasome complex for degradation.