Product Highlights
New Literature
Ion Channels
Key Resources
Cell Signaling Web Tools
New Products
PathFinder
Pathway Slides & Charts
Pharm. Action Table
Poster Gallery
Protocols
Search Tools
eHandbook
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
PDGF Receptors and Actions
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
PDGF Receptors and Actions
The α-PDGF receptor can be activated by either PDGF-A or –B chains, but the β-PDGF receptor can only be activated by PDGF-B chain. Ligand binding to one subunit induces receptor homodimerization or heterodimerization and their autophosphorylation. Cellular responses of the two receptors indicate some differences, e.g., only β-PDGF receptor is anti-apoptotic.