Product Highlights
New Literature
Ion Channels
Key Resources
Cell Signaling Web Tools
New Products
PathFinder
Pathway Slides & Charts
Pharm. Action Table
Poster Gallery
Protocols
Search Tools
eHandbook
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
Phosphorylation Driven Regulatory Switch from Transcription Initiation to Elongation
Top Border
Left Border Phosphorylation-driven Regulatory Switch from Transcription Initiation to Elongation Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
Phosphorylation Driven Regulatory Switch from Transcription Initiation to Elongation
The RNA polymerase II (pol II) A form contains an unphosphorylated carboxy-terminal domain (CTD) which initiates transcription producing 20-25 nucleotide 5’-triphosphorylated transcripts. The CTD is then phosphorylated to the RNA pol II O form, changing conformation and binding to RNA capping enzyme (CE). Stimulated by the elongation factor SPT5 and the phosphorylated CTD, CE which has RNA triphosphatase and RNA guanylyltransferase activities modifies the exposed end of the nascent transcript to GpppN. Methyl transferase (MT) binds to CE and to the terminal GpppN. Finally, Importin-a (Impa) stimulates N7 methylation of the cap GpppN by MT, and RNA pol II O form switches from the initiation process to elongation. In addition, the pol II CTD also binds splicing factors and polyadenylation factors. Hence, the pol II CTD is central to the cotranscriptional nature of RNA processing (5’ capping, splicing and 3’polyadenylation).
Back to Top
Adapted from Shatkin, A.J. et al., Nature Structural Biology, 7, 838, (2000) and McCracken, S. et al., Genes & Development 11, 3306, (1997).