Product Highlights
New Literature
Ion Channels
Key Resources
Cell Signaling Web Tools
New Products
PathFinder
Pathway Slides & Charts
Pharm. Action Table
Poster Gallery
Protocols
Search Tools
eHandbook
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
TNF-α and TNF-β (LT) Forms and Receptor Binding
Top Border
Left Border  Right Border
Left Corner Back Download Forward Sigma-Aldrich Right Corner
TNF-α and TNF-β (LT) Forms and Receptor Binding
TNF-α is expressed on the cell membrane and then hydrolyzed to release the soluble form, which forms homotrimers. TNF-β (LT-α) has no cell membrane attachment domain but can form either membrane-anchored heterotrimers with LT-β or soluble homotrimers. Receptor binding specificities are indicated.