| Adenosines/P2 Nucleotide Receptors (Purinergics) | | |
| Product
Name | Product # |
Specificity |
| Adenosine agonists | | |
| Adenosine amine congener solid | A-111 | A1 |
| 2-Chloro-N6-cyclopentyladenosine
solid | C-142 | A1 |
| N6-Cyclohexyladenosine | C9901 | A1 |
| N6-Cyclopentyladenosine solid | C8031 | A1 |
| (S)-ENBA | E-111 | A1 |
| (-)-N6-(2-Phenylisopropyl)adenosine solid | P4532 | A1 |
| N6-p-Sulfophenyladenosine sodium salt
solid | S-198 | A1 |
| (+)-N6-(2-Phenylisopropyl)adenosine
| P7665 | A1; less active
enantiomer |
| 2-Chloroadenosine | C5134 | A1,A2 |
| N6-Phenyladenosine solid | P-108 | A1,A2 |
| N6-(2-Phenylethyl)adenosine | P-107 | A1,A2 |
| Metrifudil | M-225 | A2 |
| MECA solid | A-024 | A2,A1 |
| 2-Phenylaminoadenosine >97 % solid | P-101 | A2,A1 |
| HE-NECA solid | H8034 | A2,A3 |
| CGS-21680 hydrochloride solid | C-141 | A2A |
| DPMA solid | D-130 | A2A |
| 5'-(N-Ethylcarboxamido)adenosine powder | E2387 | A2B |
| N6-(4-Aminobenzyl)-N-methylcarboxamidoadenosine
solid | A-236 | A3 |
| N6-2-(4-Aminophenyl)ethyladenosine >95
% | A-202 | A3 |
| N6-Benzyl-5'-N-ethylcarboxamidoadenosine
solid | B-152 | A3 |
| Chloro-IB-MECA solid | C-277 | A3 |
| IB-MECA solid | I-146 | A3 |
| I-AB-MECA solid | I-145 | A3
probe |
| Adenosine min. 99 % | A9251 | Endogenous |
| Adenosine hemisulfate salt min. 98 % | A7636 | Endogenous |
| Adenosine antagonists | back to top |
| CGS-15943 solid | C-199 | A1 |
| 7-Chloro-4-hydroxy-2-phenyl-1,8-naphthyridine
solid | C5982 | A1 |
| 8-Cyclopentyl-1,3-dimethylxanthine crystalline | C-102 | A1 |
| 8-Cyclopentyl-1,3-dipropylxanthine solid | C-101 | A1 |
| 1,3-Dimethyl-8-phenylxanthine crystalline | P2278 | A1 |
| FSCPX | F7927 | A1 |
| Xanthine amine congener | X-103 | A1 |
| 1,7-Dimethylxanthine approx. 98 % solid | D5385 | A1 >
A2 |
| 1,3-Dipropyl-8-(p-sulfophenyl)xanthine solid | A-022 | A1 >
A2 |
| 3-Isobutyl-1-methylxanthine min. 99 %
(HPLC) powder | I5879 | A1 >
A2 |
| 3-Propylxanthine min. 99 % solid | P5679 | A1 >
A2 |
| 8-(p-Sulfophenyl)theophylline solid | A-013 | A1 >
A2 |
| Theophylline anhydrous min. 99 % powder | T1633 | A1 >
A2 |
| 3,7-Dimethyl-1-propargylxanthine solid | D-134 | A2 |
| Aminophylline hydrate min. 98 % powder | A1755 | A2 >
A1 |
| 8-(3-Chlorostyryl)caffeine solid | C-197 | A2A |
| SCH-58261 99 % (HPLC) solid | S4568 | A2A |
| Alloxazine solid | A-242 | A2B |
| MRS 1754 >98 % (HPLC) | M6316 | A2B |
| MRS 1191 solid | M-227 | A3 |
| MRS 1220 solid | M-228 | A3 |
| MRS 1523 >98 % (HPLC) | M1809 | A3 |
| VUF 5574 | V5888 | A3 |
| Enzyme inhibitors | back to top |
| erythro-9-(2-Hydroxy-3-nonyl)adenine hydrochloride
solid | E-114 | Adenosine
deaminase |
| P2 Receptor agonists | back to top |
| Cytidine 5'-triphosphate disodium salt approx. 98
% | C1506 | Endogenous |
| Adenosine 5'-[g-thio]triphosphate tetralithium salt min. 75 % powder | A1388 | P2 |
| Adenosine 5'-triphosphate disodium salt Grade I min.
99 % | A2383 | P2 |
| 2-(Methylthio)adenosine 5'-diphosphate trisodium salt >89
% solid | M-152 | P2 |
| 2-(Methylthio)adenosine 5'-triphosphate tetrasodium salt
>91 % solid | A-023 | P2 |
| Adenosine 5'-[b-thio]diphosphate trilithium salt min. 80 % | A8016 | P2X |
| 8-Bromoadenosine 5'-triphosphate sodium salt approx. 95
% | B3756 | P2X |
| P1,P4-Di(adenosine-5')
tetraphosphate ammonium salt approx. 95 % (HPLC) powder | D1262 | P2X |
| a,b-Methyleneadenosine 5'-triphosphate lithium salt min. 95 % solid | M6517 | P2X > P2Y |
| b,g-Methyleneadenosine 5'-triphosphate disodium salt min. 95 % solid | M7510 | P2X > P2Y |
| Adenosine 5'-diphosphate monopotassium salt dihydrate
bacterial 98-100 % crystalline | A5285 | P2X1 |
| Cytidine 5'-triphosphate disodium solution
| C8552 | P2X3 |
| 2-Chloroadenosine triphosphate tetrasodium salt
solid | C-145 | P2Y |
| Uridine 5'-triphosphate trisodium salt solution
| U1006 | P2Y2 |
| Uridine 5'-diphosphate sodium salt from Saccharomyces
cerevisiae 95-100 % | U4125 | P2Y6 |
| P2 Receptor antagonists | back to top |
| P2 Receptor antagonists solid | P-178 | P2 |
| Suramin sodium salt min. 95 % solid | S2671 | P2 |
| NF 023 crystalline | N8652 | P2X1/3 |
| TNP-ATP solid | T4193 | P2X1/3 |
| Diinosine pentaphosphate pentasodium salt | D6938 | P2X1 |
| MRS 2159 >98 % (HPLC) solid | M7684 | P2X1 |
| PPNDS solid | P2738 | P2X1 |
| Brilliant Blue G min. 90 % (TLC) | B5133 | P2X7 |
| Adenosine 2',5'-diphosphate sodium salt min. 95
% | A5638 | P2Y |
| Adenosine 3',5'-diphosphate sodium salt approx. 97
% | A5763 | P2Y |
| Reactive Blue 2 solid | R-115 | P2Y |
| MRS 2179 >98 % (HPLC) | M3808 | P2Y1 |
| MRS 2395 ³98 %
(HPLC) solid | M5942 | P2Y12 |
| Transporter | | |
| Adenosine Transporter Receptor human Membrane
suspension | A8352 | |
| Transport inhibitors | back to top |
| Dilazep dihydrochloride powder | D5294 | |
| Dipyridamole min. 98 % (TLC) powder | D9766 | |
| 5-Iodotubericidin solid | I-100 | |
| S-(4-Nitrobenzyl)-6-thioinosine min. 98
% solid | N2255 | |
| Adrenergics | | back to top |
| Product
Name | Product # | Specificity |
| Agonists | | |
| 6-Fluoronorepinephrine hydrochloride solid | B-012 | a |
| Mephentermine hemisulfate salt | M3656 | a |
| Metaraminol (+)-bitartrate salt | M4773 | a |
| (±)-Octopamine hydrochloride solid | O0250 | a |
| (±)-Synephrine min. 99 % | S0752 | a |
| Tetrahydrozoline hydrochloride min. 98 % | T4264 | a |
| Xylometazoline hydrochloride | X6000 | a |
| Dihydroergotamine methanesulfonate salt powder | D2763 | a
(Partial) |
| (R)-(-)-Phenylephrine hydrochloride
| P6126 | a1 |
| Cirazoline hydrochloride solid | C-223 | a1A |
| Methoxamine hydrochloride | M6524 | a1A, Aa1D |
| Naphazoline hydrochloride | N5504 | a1A, Aa1D |
| Agmatine sulfate salt min. 97 % powder | A7127 | a2 |
| p-Aminoclonidine hydrochloride solid | A0779 | a2 |
| B-HT 920 dihydrochloride solid | B-162 | a2 |
| B-HT 933 dihydrochloride solid | B-161 | a2 |
| Clonidine hydrochloride solid | C7897 | a2 |
| Guanabenz acetate salt solid | G-110 | a2 |
| Guanfacine hydrochloride >98 % (HPLC) | G1043 | a2 |
| Rilmenidine hemifumarate salt solid | R-134 | a2 |
| UK 14,304 | U-104 | a2 |
| Xylazine min. 99 % | X1126 | a2 |
| Xylazine hydrochloride min. 99 % | X1251 | a2 |
| Moxonidine hydrochloride min. 98 % | M1559 | a2A |
| Oxymetazoline hydrochloride solid | O2378 | a2A (partial) |
| Bambuterol hydrochloride >98 %
(HPLC) powder | B8684 | b |
| (±)-CGP-12177 hydrochloride >98
% solid | C-125 | b |
| Isoetharine mesylate salt | I3639 | b |
| (±)-Isoproterenol hydrochloride | I5627 | b |
| (-)-Isoproterenol (+)-bitartrate
salt | I2760 | b |
| (-)-Isoproterenol hydrochloride
| I6504 | b |
| (±)-Isoproterenol hemisulfate salt | I5752 | b |
| Tulobuterol hydrochloride | T6050 | b |
| R(-)-Denopamine >98 %
(HPLC) | D7815 | b1 |
| Dobutamine hydrochloride min. 98 % | D0676 | b1 |
| Xamoterol hemifumarate salt >98 % (HPLC) | X3253 | b1 (Partial) |
| Clenbuterol hydrochloride min. 95 % | C5423 | b2 |
| Fenoterol hydrobromide | F1016 | b2 |
| Formoterol fumarate >98 % (HPLC) | F9552 | b2 |
| Metaproterenol hemisulfate salt | M2398 | b2 |
| Procaterol hydrochloride | P9180 | b2 |
| Ritodrine hydrochloride | R0758 | b2 |
| Salbutamol | S8260 | b2 |
| Salbutamol hemisulfate salt | S5013 | b2 |
| Salmeterol >98 % (HPLC) solid | S2692 | b2 |
| Terbutaline hemisulfate salt | T2528 | b2 |
| BRL 37344 sodium salt powder | B-169 | b3 |
| CL 316,243 min. 98 % | C5976 | b3 |
| (-)-Epinephrine | E4250 | Endogenous a, b |
| (-)-Epinephrine (+)-bitartrate salt
solid | E4375 | Endogenous a, b |
| (±)-Epinephrine min. 98 % | E1635 | Endogenous a, b |
| (±)-Epinephrine hydrochloride | E4642 | Endogenous a, b |
| (-)-Norepinephrine (+)-bitartrate
salt hydrate min. 99 % solid | A9512 | Endogenous a, b1 |
| (-)-Norepinephrine min. 98
% crystalline | A7257 | Endogenous a, b1 |
| (±)-Norepinephrine (+)-bitartrate salt | A0937 | a, b1 |
| (+)-Isoproterenol (+)-bitartrate salt | I8005 | Less active
enantiomer |
| Antagonists | | |
| Phenoxybenzamine hydrochloride min. 97
% powder | B-019 | a |
| Phentolamine hydrochloride min. 99 %
(TLC) powder | P7547 | a |
| Phentolamine methanesulfonate salt min. 99 %
(TLC) powder | P7561 | a |
| Spiroxatrine powder | S-103 | a |
| Terazosin hydrochloride min. 98 %
(TLC) powder | T4680 | a |
| R(+)-Atenolol solid | A-142 | a1 |
| Benoxathian hydrochloride solid | B-016 | a1 |
| Doxazosin mesylate >98 % (HPLC) | D9815 | a1 |
| Fiduxosin hydrochloride >98 % (HPLC) | F4303 | a1 |
| Moxisylyte hydrochloride >99 % (TLC) powder | M5154 | a1 |
| Naftopidil dihydrochloride solid | N-158 | a1 |
| Urapidil hydrochloride solid | U-100 | a1 |
| Prazosin hydrochloride min. 99 %
(TLC) solid | P7791 | a1 (peripheral) |
| Prazobind solid | P-124 | a1 alkylating agent |
| 5-Methylurapidil solid | U-101 | a1A |
| (±)-Niguldipine hydrochloride solid | N-134 | a1A |
| S(+)-Niguldipine hydrochloride solid | N-135 | a1A |
| WB-4101 hydrochloride >99 % (TLC) solid | D0411 | a1A |
| L-765,314 min. 98 % | L3040 | a1B |
| Spiperone solid | S7395 | a1B |
| Chloroethylclonidine dihydrochloride solid | B-003 | a1B alkylating agent |
| A-315456 >98 % (HPLC) solid | A6351 | a1D |
| BMY 7378 dihydrochloride >98 % solid | B-134 | a1D |
| Efaroxan hydrochloride | E3263 | a2 |
| Idazoxan hydrochloride | I6138 | a2 |
| MK-912 hydrochloride salt >98 % (HPLC) | M7065 | a2 |
| Rauwolscine hydrochloride solid | R-104 | a2 |
| RX 821002 solid | R9525 | a2 |
| SKF 86466 solid | S1563 | a2 |
| Tolazoline hydrochloride min. 99 %
(titration) powder | T6886 | a2 |
| Yohimbine hydrochloride min. 98 %
(TLC) powder | Y3125 | a2 |
| BRL 44408 maleate >99 % (HPLC) solid | B4559 | a2A |
| Imiloxan hydrochloride >98 % (HPLC) | I9531 | a2B |
| Labetalol hydrochloride >99 % (TLC) powder | L1011 | b |
| (S)-(-)-Propranolol hydrochloride
min. 99 % (TLC) powder | P8688 | b |
| (±)-Propranolol hydrochloride min. 99 %
(TLC) powder | P0884 | b |
| (±)-Sotalol hydrochloride min. 98 %
(TLC) powder | S0278 | b |
| Atenolol min. 98 % (TLC) powder | A7655 | b1 |
| S(-)-Atenolol solid | A-143 | b1 |
| Betaxolol hydrochloride >98 % (HPLC) | B5683 | b1 |
| CGP-20712A methanesulfonate salt solid | C-231 | b1 |
| Cyclazosin hydrochloride solid | C-247 | b1B |
| (±)-Metoprolol (+)-tartrate salt min. 99 %
(titration) powder | M5391 | b1 |
| Timolol maleate salt min. 98 % (TLC) powder | T6394 | b1 |
| Alprenolol hydrochloride >99 % (TLC) powder | A8676 | b1, b2 |
| Pindolol min. 98 % (TLC) powder | P0778 | b1, b2 |
| S()-Pindolol solid | P-152 | b1, b2 |
| ICI 118,551 hydrochloride min. 99 %
(TLC) powder | I-127 | b2 |
| Bromoacetyl alprenolol menthane powder | B-015 | b2 > b1 alkylating agent |
| S(-)-Cyanopindolol hemifumarate
salt solid | C-238 | b3 |
| SR 59230A oxalate salt solid | S8688 | b3 |
| (R)-(+)-Propranolol hydrochloride min. 99 %
(TLC) powder | P0689 | Less active
enantiomer |
| Enzyme inhibitors | | |
| Benserazide hydrochloride min. 98 %
(TLC) solid | B7283 | L-Amino acid
decarboxylase |
| S-(-)-Carbidopa >98
% solid | C-126 | L-Amino acid
decarboxylase |
| S(-)-Carbidopa: HBC complex
| C-164 | L-Amino acid
decarboxylase |
| (±)-3-(3,4-Dihydroxyphenyl)-2-methyl-DL-alanine | M7277 | L-Amino acid
decarboxylase |
| 3-Hydroxybenzylhydrazine dihydrochloride 95-98 %
(TLC) | H9382 | L-Amino acid
decarboxylase |
| Ro 41-0960 solid | R-108 | Catechol-O-methyltransferase |
| Fusaric acid from Gibberella fujikuroi | F6513 | Dopamine b-hydroxylase |
| 3-Phenylpropargylamine hydrochloride solid | P-106 | Dopamine b-hydroxylase |
| 1-Phenyl-3-(2-thiazolyl)-2-thiourea | P4015 | Dopamine b-hydroxylase |
| R-(-)-Deprenyl hydrochloride
solid | M-003 | Monoamine
oxidase |
| Hydralazine hydrochloride | H1753 | Monoamine
oxidase |
| Hydroxylamine hydrochloride min. 99 %
(titration) | H9876 | Monoamine
oxidase |
| Hydroxylamine sulfate salt min. 99 %
(titration) | H0127 | Monoamine
oxidase |
| Iproniazid phosphate salt | I7627 | Monoamine
oxidase |
| N-Methyl-N-propargyl-3-(2,4-dichlorophenoxy)propylamine
hydrochloride | M3778 | Monoamine
oxidase |
| Nialamide min. 95 % (HPLC) | N1392 | Monoamine
oxidase |
| Pargyline hydrochloride | P8013 | Monoamine
oxidase |
| Phenelzine sulfate salt | P6777 | Monoamine
oxidase |
| trans-2-Phenylcyclopropylamine hemisulfate salt
| P1761 | Monoamine
oxidase |
| trans-2-Phenylcyclopropylamine hydrochloride | P8511 | Monoamine
oxidase |
| Quinacrine dihydrochloride min. 90 % | Q3251 | Monoamine
oxidase |
| Ro 41-1049 hydrochloride solid | R-107 | Monoamine
oxidase |
| Ro 16-6491 hydrochloride solid | R-106 | Monoamine
oxidase |
| (±)-2,3-Dichloro-a-methylbenzylamine hydrochloride solid | D-103 | Phenylethanolamine
N-methyltransferase |
| 3-Iodo-L-tyrosine | I8250 | Tyrosine
hydroxylase |
| a-Methyl-L-tyrosine
| M8131 | Tyrosine
hydroxylase |
| a-Methyl-DL-tyrosine
methyl ester hydrochloride min. 98 % (TLC) | M3281 | Tyrosine
hydroxylase |
| Enzymes | | |
| Catechol-O-methyl Transferase porcine liver Lyophilized
powder min. 1,000 units/mg protein | C1897 | |
| Phenylethanolamine N-Methyltransferase from bovine adrenal
medulla Lyophilized powder | P8924 | |
| Metabolites | < |