|
All Sigma antibodies are carefully tested to provide high quality performance. The antibodies to HDAC, developed against many of the histone modifying enzymes, are valuable tools for scientists working on chromatin remodeling.
Regulation of gene expression is mediated by several mechanisms including DNA methylation, ATP-dependent chromatin remodeling, and posttranslational modifications of histones. One of the major modifications of histones consists of the dynamic acetylation and deacetylation of ε-amino groups of lysine residues present in the tail of core histones. The enzymes responsible for this reversible acetylation/deacetylation process are histone acetyltransferases (HATs) and histone deacetylases (HDACs), respectively. While HATs act as transcriptional coactivators, HDACs are part of transcriptional corepressor complexes.
These antibodies can be used in a variety of applications such as immunoblotting. Please see the product datasheets for additional information and details. Visit our Antibody Explorer for a complete listing of all Sigma antibodies.
Histone Deacetylases
| Product Name |
Host |
Clone |
Product # |
| Monoclonal Anti-Histone Deacetylase 1 (HDAC1) |
Mouse |
HDAC1-21 |
H6287 |
| Anti-Histone Deacetylase 1 (HDAC1) |
Rabbit |
|
H3284 |
| Monoclonal Anti-Histone Deacetylase 2 (HDAC2) |
Mouse |
HDAC2-62 |
H2663 |
| Anti-Histone Deacetylase 2 (HDAC2) |
Rabbit |
|
H3159 |
| Monoclonal Anti-Histone Deacetylase 3 (HDAC3) |
Mouse |
HDAC3-83 |
H6537 |
| Anti-Histone Deacetylase 3 (HDAC3) |
Rabbit |
|
H3034 |
| Monoclonal Anti-Histone Deacetylase 4 (HDAC4) |
Mouse |
HDAC4-144 |
H0163 |
| Anti-Histone Deacetylase 4 (HDAC4) (ML-19) |
Rabbit |
|
H9411 |
| Anti-Histone Deacetylase 4 (HDAC4) (DM-15) |
Rabbit |
|
H9536 |
| Anti-Histone Deacetylase 6 (HDAC6) |
Rabbit |
|
H2287 |
| Anti-Histone Deacetylase 7 (HDAC7) (YS-19) |
Rabbit |
|
H2537 |
| Anti-Histone Deacetylase 7 (HDAC7) (KG-17) |
Rabbit |
|
H2662 |
| Monoclonal Anti-Histone Deacetylase 8 (HDAC8) |
Mouse |
HDAC8-48 |
H6412 |
| Anti-Histone Deacetylase 10 (HDAC10) |
Rabbit |
|
H3413 |
| Monoclonal Anti-Histone Deacetylase 11 (HDAC11) |
Mouse |
HDAC11-31 |
H2913 |