Available Exclusively from Sigma-RBI
CINALUKAST
Prod. No. C6239
Subtype selective, orally active, cysteinyl leukotriene (CysLT1) receptor antagonist
Cysteinyl leukotriene (CysLT) receptors mediate a range of pro-inflammatory effects, such as constriction of airways and vascular smooth muscle, increased permeability of endothelial membranes leading to plasma exudation and edema, and enhanced secretion of viscous mucus. CysLT receptors have therefore been implicated in a range of inflammatory diseases, notably asthma.1-3 There are at least two subtypes of CysLT leukotriene receptors, referred to as CysLT1 and CysLT2, and although there is some evidence for further subdivision, this remains to be confirmed.4 Sigma-RBI is pleased to be the first company to offer cinalukast (Ro 24-5913; Prod. No. C6239), a selective CysLT1 leukotriene receptor antagonist.
Cinalukast:
- In vitro, competes with [3H]-LTD4 binding to CysLT1 leukotriene receptors in guinea-pig lung membranes, displaying an IC50 value of 6.4 nM.4
- In vivo, dose-dependently inhibits bronchoconstriction induced by both the non-subtype selective CysLT agonist LTC4 (Prod. No. L4886) and the CysLT1 selective agonist LTD4 (Prod. No. L5011) by i.v. (ID50= 0.13 mg/kg), oral (ID50= 0.12 mg/kg) and aerosol (IC50= 0.008%) routes of administration.4
- Inhibits 100% of both LTC4- and LTD4- induced bronchoconstriction at a dose of 1 mg/kg.4
- In a single oral dose (10 mg/kg), produces a long-lasting inhibition of bronchoconstriction, with pharmacological effects being observed within 1 hour and maintained for up to 36 hrs.4
| Physical Description |
|
Off-white solid |
| Solubility |
|
Soluble in DMSO at 10 mg/ml. Insoluble in water. |
| Molecular Formula |
|
C23H28N2O3S |
| Formula Weight |
|
412.55 |
| Sizes |
|
5 mg; 25mg |
|
|
References:
- Figueroa, D.J., et al., Clin. Exp. Allergy, 33, 1380-1388 (2003).
- Kawano, T., J. Allergy Clin. Immunol., 112, 369-374 (2003).
- Hui, Y. and Funk, C.D., Biochem. Pharmacol., 64, 1549-1157 (2002).
- O'Donnell, M., et al., J. Pharmacol. Exp. Ther., 259, 751-758 (1991).
back to top
back to Cell Signaling
|