Product Highlights
Anti-Glutamate NR1
Antibodies to HDAC
BACE1 Assay Kit
Fluorescent Probes
Isradipine
MRS 2395
Monoclonal Anti g Parvin
Monoclonal Anti-D1
Receptor Preparations
SB-431542
XK469
nNOS Inhibitors
Previous Highlights
New Literature
Ion Channels
Key Resources
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 Citalopram: Selective Serotonin Reuptake Inhibitor
Cell Signaling & Neuroscience
Now Available from Sigma-RBI
Prod. No. C 7861
Citalopram (Prod. No. C 7861) belongs to the class of antidepressant drugs known as selective serotonin reuptake inhibitors (SSRIs). SSRIs enhance serotonergic neurotransmission by selectively inhibiting the reuptake of serotonin (Prod. No. H 9523) by serotonergic neurons. This increases the concentration of serotonin in the synaptic cleft and facilitates the activation of pre- and post-synaptic serotonergic receptors. The serotonin transporter (SERT) mediates this reuptake and is the molecular target of SSRIs.1-3
Sigma-RBI is pleased to introduce Citalopram hydrobromide as a selective serotonin reuptake inhibitor:
  • Does not show adverse anticholinergic or cardiovascular effects.
  • Blocks both HERG (humanERG1) potassium channels and L-type calcium channels. 4
  • Inhibits cardiac sodium and calcium channels. 5
References
  1. Fuller, R.W., Prog. Drug Res., 45, 167-204 (1995).
  2. Mortensen, O.V., et al., J. Neurochem., 79, 237-247 (2001).
  3. Horschitz, S., et al., Neuroreport, 12, 2181-2184 (2001).
  4. Witchel, H.J., et al., FEBS Lett., 512, 59-66 (2002).
  5. Pacher, P., et al., Curr. Med. Chem., 6, 469-480 (1999).
Citalopram is featured in Celltransmissions, Volume 18, No. 2. Click here to preview the product highlight.