Product Highlights
Anti-Glutamate NR1
Antibodies to HDAC
BACE1 Assay Kit
Fluorescent Probes
Isradipine
MRS 2395
Monoclonal Anti g Parvin
Monoclonal Anti-D1
Receptor Preparations
SB-431542
XK469
nNOS Inhibitors
Previous Highlights
New Literature
Ion Channels
Key Resources
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 Dephostatins: Selective, Potent and Stable Tyrosine Phosphatase Inhibitors
Cell Signaling & Neuroscience
Now Available from Sigma-RBI
Dephostatin (Prod. No. D 8065)
Ethyl-3,4-dephostatin (Prod. No. E 1904)
Methyl-3,4-dephostatin (Prod. No. M 9440)
Protein-tyrosine phosphatases (PTPases) catalyze the reversible phosphorylation of tyrosine residues. The PTPases comprise two classes, transmembrane receptor PTPases, which contain linked cytoplasmic catalytic domains, and intracellular PTPases, which include SRC homology 2 (SH2) domains.1,2 Sigma-RBI is pleased to offer dephostatin (Prod. No. D 8065), a selective inhibitor of CD45 and SHP1-associated PTPases, and its two stable analogs, Ethyl-3,4-dephostatin (Prod. No. E 1904) and Methyl-3,4-dephostatin (Prod. No. M 9440).
  • Dephostatin inhibits PTPase prepared from a human neoplastic T-cell line with an IC50 = 7.7 µM.3
  • Ethyl-3,4-dephostatin and Methyl-3,4-dephostatin strongly inhibit PTP-1B and SHPTP, with IC50 values 0.58 and 0.96 µg/ml, respectively.1
  • Ethyl-3,4-dephostatin enhances the phosphorylation of other proteins, such as c-Cbl, phospholipase Cg (PLCg), phosphatidylinositol 3-kinase (PI3K) and insulin receptor 3 substrate (IRS-3).4
  • Methyl-3,4-dephostatin inhibits CD45-associated PTPases while Ethyl-3,4-dephostatin does not exhibit inhibitory activity.
References:
  1. Watanabe, T., et al., J. Chem. Soc. Chem. Commun., 4, 437-438 (1993).
  2. Kaplan, R., et al., Proc. Nat. Acad. Sci. USA, 87, 7000-7004 (1990).
  3. Imoto, M., et al., J. Antibiot. (Tokyo), 46, 1342-1346 (1993).
  4. Suzuki, T., et al., J. Biol. Chem., 276, 27511-27518 (2001).