Product Highlights
Anti-Glutamate NR1
Antibodies to HDAC
BACE1 Assay Kit
Fluorescent Probes
Isradipine
MRS 2395
Monoclonal Anti g Parvin
Monoclonal Anti-D1
Receptor Preparations
SB-431542
XK469
nNOS Inhibitors
Previous Highlights
New Literature
Ion Channels
Key Resources
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 Isradipine
Cell Signaling & Neuroscience
Potent, selective L-type voltage gated calcium channel blocker; antihypertensive
Prod. No. I6658
Voltage-gated calcium channels are a major route of calcium translocation across the plasma membrane of excitable cells. Six types of calcium channels have been identified: T, L, N, P, Q, and R. T-type channels need only small depolarizations to be activated (low-voltage activated, LVA), while the other types require larger depolarizations (high-voltage activated, HVA). Compounds such as verapamil, isradipine, nefedipine, dilantizem and 1,4-dihydropine analogs of nefedipine interact with the L-type calcium channel and are widely used as antihypertensives and in the treatment of certain vascular disorders.
Features & Benefits
Isradipine has been widely used in the study of hypertension, ischemia, and depression. It belongs to the class of compounds known as 1,4 dihydropyridines, and appears to be selective for the L-type calcium channel subtype CaV1.2 (1).
  • Isradipine (2.5 mg/kg) significantly reduced the volume of ischemic brain damage in the cerebral hemisphere (25%; P = 0.0001), cerebral cortex (18%; P = 0.0034), and caudate nucleus (33%; P = 0.0002) in spontaneously hypertensive rats when assessed at 24 h post-MCA occlusion (2).
  • Isradipine, as well as several other DHP drugs (nicardipine, nitrendipine, felodipine, nimodipine) displayed antidepressant-like properties in the tail suspension test in mice (3).
  • Isradipine-induced acceleration of the CaV1.2 current decay may reflect enhanced fast voltage-dependent inactivation rather than open channel block in a calcium channel construct Lh, corresponding to rabbit cardiac α1C-a cDNA (1).
Additional L-type calcium channel blockers offered by Sigma-RBI  
Product Name Product # 
(R)-(+)-Bay K8644 Manufactured and sold with the permission of Bayer AG B-132
Calcicludine 2 C2836
Dilantizem D2521
Felodipine F9677
FS-2 F3176
Nicardipine N7510
Nifedipine N7634
Nimodipine N-149
Nitrendipine N-144
S-Petasin P4243
Phloretin P7912
(+)-Verapamil hydrochloride V4629
References
  1. Berjukow, S and Hering, S. Voltage-dependent acceleration of Cav1.2 channel current decay by (+)- and (-)- isradipine. Br. J. Pharmacol., 133, 959-966, 2001.
  2. Campbell, C.A., et al., Effects of isradipine, an L-type calcium channel blocker on permanent and transient focal cerebral ischemia in spontaneously hypertensive rats. Exp. Neurol., 148, 45-50, 1997.
  3. Cohen, C., et al., Assessment of the antidepressant-like effects of L-type voltage-dependent channel modulators. Behav. Pharmacol., 8, 629-638, 1997.
For more information about calcium channel modulators, please visit The Sigma-RBI Handbook of Receptor Classification and Signal Transduction.