Product Highlights
Anti-Glutamate NR1
Antibodies to HDAC
BACE1 Assay Kit
Fluorescent Probes
Isradipine
MRS 2395
Monoclonal Anti g Parvin
Monoclonal Anti-D1
Receptor Preparations
SB-431542
XK469
nNOS Inhibitors
Previous Highlights
New Literature
Ion Channels
Key Resources
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 L-685,458
Cell Signaling & Neuroscience
L-685,458
Potent, selective cell-permeable g-secretase inhibitor
L1790
Alzheimer's disease (AD) accounts for the majority of the dementia diagnosed after the age of 60, with over 12 million sufferers worldwide. The disease is characterized by the progressive loss of cognitive function and results in end-stage patients that are bedridden and dependent on custodial care.
The pathogenesis of AD is believed to result from the progressive accumulation in the brain of b-amyloid (Ab), a 4 kDa protein. Ab originates from the proteolytic cleavage of amyloid precursor protein (APP) by two proteases, b-secretase and g-secretase (Figure 1). Cleavage by b-secretase at the amino terminus of APP results in a soluble b-APP and a 12 kDa membrane-bound carboxyl terminal fragment (CTF). The latter in turn becomes the substrate for g-secretase to yield a soluble Ab peptide, Amyloid b protein fragment (1-40) (Ab (1-40)), or the slightly longer, insoluble peptide Amyloid b protein fragment (1-42) (Ab (1-42)). APP can also be cleaved by a third enzyme, a-secretase, leading to the production of soluble a-APP peptide and a 10 kDa CTF. Although Ab (1-42) and Ab (1-40) account for approximately 10% and 90%, respectively, of the total Ab secreted from cells, Ab (1-42) constitutes the major component of the nonfibrillar extracellular plaques that precede the development of the dense, fibrillar neuritic plaques characteristic of AD [1]. Because inhibition of g-secretase blocks the production of Ab, the identification of compounds that block the activity of this enzyme has become a major focus of AD research.
L-685,458 is a novel, potent and selective cell-permeable g-secretase inhibitor [2-5].
  • Exhibits over 100-fold greater selectivity for g-secretase than for a panel of other proteases [2].
  • Potently inhibits g-secretase and thus the production of Ab total (IC50 17 nM), Ab (1-40), (IC50 48 nM) and Ab (1-42) (IC50 67 nM) in human neuroblastoma SHSY5Y cells overexpressing spbA4CTF, a truncated form of human APP [2].
Manufactured and sold under non-exclusive license from Merck & Co. Inc.
Related secretase and b-amyloid products:
  Secretase Inhibitors
Secretase
Secretase Kit
Secretase Antibodies
Other Secretase Products
b-Amyloid inhibitors
b-Amyloid
b-Amyloid Probes
b-Amyloid Kits
b-Amyloid Antibodies
Secretase Inhibitors  
Product Name Product # 
b-Secretase Inhibitor S4562
g-Secretase Inhibitor S2188
MG132 C2211
Glu-Val-Asn-[(2R,4S,5S)-5-amino-4-hydroxy-2,7-dimethyloctanoyl]-Ala-Glu-Phe G8291
Secretase back to top
b-secretase (BACE1) S5067
Secretase Kit back to top
b-Secretase (BACE1) Activity Detection Kit (Fluorescent) CS0010
Secretase Antibodies back to top
Anti-BACE-1 (EE-17) Rabbit, IgG fraction of antiserum B0681
Anti-BACE-1 (LK-16) Rabbit, IgG fraction of antiserum B0806
Other Secretase Products back to top
7- Methoxycoumarin-4-acetyl-Glu-Val-Lys-Met-Asp-Ala-Glu-Phe-(2,4-dinitrophenyl)-Lys-OH trifluoroacetate salt (FRET peptide for detecting b-secretase) M4689
7-Methoxycoumarin-4-acetyl-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-(2,4-dinitrophenyl)-Lys trifluoroacetate salt (FRET peptide for detecting a-secretase) M4564
Mca-[Asn670, Leu671]-Amyloid b/A4 Precursor Protein 770 fragment 667-675-(Dnp)Lys amide (Peptide containing the b-secretase site of Swedish mutation of APP; fluorogenic b-secretase substrate) M1063
b-Amyloid inhibitors back to top
Cloquinol C8133
Dabcyl-[Asn670,Leu671]-Amyloid b/A4 Protein Precursor770 Fragment (661-675)-Edans A4972
Flufenamic acid F9005
Pepstatin A P5318
b-Amyloid back to top
Ab (1-40) A1075
Ab (1-42) A9810
b-Amyloid Probes back to top
BTA-1 B9934
Chrysamine G C1115
b-Amyloid Kits back to top
b-Amyloid 1-40 ELISA, human BE0100
b-Amyloid 1-42 ELISA, human BE0200
b-Amyloid Antibodies back to top
Monoclonal Anti-b Amyloid [17-24] Human (Clone 4G8) Mouse, purified IgG2b A1349
Monoclonal Anti-b Amyloid [1-17] (Human Clone 6E10) Mouse, purified IgG1 A1474
Anti-Amyloid Peptide b Cleavage Site 42 Rabbit, Affinity isolated antibody, Buffered aqueous solution A1976
Anti-Amyloid Peptide b, Cleavage Site 43 Rabbit, Affinity isolated antibody, Buffered aqueous solution A2101
Monoclonal Anti-Amyloid b-Protein (Clone BAM-10) Mouse, ascites fluid, isotype IgG1 A5213
Anti-Amyloid b-Protein (1-40) Rabbit, whole antiserum A8326
Anti-Amyloid Precursor Protein, C-Terminal Rabbit, IgG fraction of antiserum A8717
Anti-Amyloid Precursor Protein, KPI Insert Rabbit, IgG fraction of antiserum A8842
Anti-Amyloid Precursor Protein, N-Terminal Rabbit, IgG fraction of antiserum A8967
Anti-ERAB (endoplasmic reticulum Rabbit, IgG fraction of antiserum Rabbit, affinity isolated antibody E1151
back to top
Figure 1. Cleavage of Amyloid Precursor Protein.
Amyloid precursor protein (APP) is sequentially cleaved by b-secretase and then g-secretase to form soluble amyloid precursor protein b (sAPPb) and the amyloid b 42 protein fragment (Ab42). The Ab42 fragments then aggregate and form the extracellular plaques common to Alzheimer's disease.
References
  1. Hardy, J. and Selkoe, D.J., Science, 297, 353-356 (2002).
  2. Shearman, M.S., et al., Biochemistry, 39, 8698-8704 (2000).
  3. Li, Y-M., et al., Proc. Nat. Acad. Sci. USA, 97, 6138-6143 (2000).
  4. Li, Y-M., et al., Nature, 405, 689-694 (2000).
  5. Tian G., et al., J. Biol. Chem., 277, 31499-31505 (2002).
back to top
back to Cell Signaling