Product Highlights
Anti-Glutamate NR1
Antibodies to HDAC
BACE1 Assay Kit
Fluorescent Probes
Isradipine
MRS 2395
Monoclonal Anti g Parvin
Monoclonal Anti-D1
Receptor Preparations
SB-431542
XK469
nNOS Inhibitors
Previous Highlights
New Literature
Ion Channels
Key Resources
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 MRS 1754: A Selective A2B Adenosine Receptor Antagonist
Cell Signaling & Neuroscience
Exclusively from Sigma-RBI
Prod. No. M 6316
Adenosine (Prod. No. A 9251) receptors have been implicated in the etiology of various cardiovascular, inflammatory and neurological diseases.
Four adenosine receptor subtypes have been identified and are referred to as A1, A2A, A2B and A3. A2B adenosine receptors stimulate adenylyl cyclase, increase intracellular calcium release and are involved in the control of vascular tone, cell growth and gene expression, mast cell degranulation and intestinal water secretion [1].
Sigma-RBI is pleased to introduce MRS 1754, a selective, high-affinity A2B adenosine receptor antagonist.
  • Binds to human A2B and A2A adenosine receptors expressed in HEK-293 cells with KD values of
    1.13 nM and >50 nM, respectively [2].
  • Does not bind to membranes expressing rat or human A1 or A3 receptors.
  • Displaces both [3H]-ZM 241385 and [125I]-I-ABOPX (non-selective antagonists) from human A2B adenosine receptors with Ki values of 1.97 nM [2].
Manufactured and sold under exclusive license from Adenosine Therapeutics, LLC.
References
  1. Kim, Y-C., et al., J. Med. Chem., 43, 1165-1172 (2000).
  2. Ji, X-D., et al., Biochem. Pharmacol., 61, 657-663 (2001).
MRS 1754 is featured in Celltransmissions Volume 18, No. 4. Click here to preview the product highlight.
back to Cell Signaling