Product Highlights
Anti-Glutamate NR1
Antibodies to HDAC
BACE1 Assay Kit
Fluorescent Probes
Isradipine
MRS 2395
Monoclonal Anti g Parvin
Monoclonal Anti-D1
Receptor Preparations
SB-431542
XK469
nNOS Inhibitors
Previous Highlights
New Literature
Ion Channels
Key Resources
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 MRS 2395
Cell Signaling & Neuroscience
First P2Y12 Receptor Antagonist Available from Sigma-RBI
2,2-Dimethyl-propionic acid 3-(2-chloro-6-methylaminopurin-9-yl)- 2-(2,2-dimethyl-propionyloxymethyl)-propyl ester
Prod. No. M5942
The surface receptors for extracellular nucleotides, both purine and pyrimidine, are called P2 receptors. P2 receptors are divided into two categories based on molecular structure and signal transduction mechanisms: P2Y (G-protein coupled receptors) and P2X (ligand-gated ion channels) (1). These receptor subtypes are found in the central and peripheral nervous system, the cardiovascular system, the endocrine system, lungs, intestines, muscle, and the immune system (2). Twelve P2Y receptor subtypes have been identified: P2Y1-12. Each P2Y receptor binds to a single heterotrimeric G protein (typically Gq/11), although P2Y12 couples to GI. Endogenous P2Y receptors can activate phopholipases A2, C, and D MEP/MAP kinase, Rho-dependent kinase and tyrosine kinase, as well as coupling both positively and negatively to adenylyl cyclase (1). In the cardiovascular system, ADP that is released from platelets and endothelial cells can induce aggregation of platelets by acting at P2Y1 and P2Y12 receptors; thus, antagonists of these subtypes may display antithrombotic properties (2).
Features & Benefits
MRS 2395 is one of the first identified antagonists of P2Y12 receptors, giving researchers a new tool in studying the function of this receptor subtype as well as its role in various physiological functions.
  • MRS 2395 inhibited ADP-induced aggregation in rat platelets with KI = 3.6 7 µM (2).
  • MRS 2395 was found to antagonize ADP-induced inhibition of cAMP in rat and human platelets in the presence of PGE1, (IC50 = 7 µM and 30 µM respectively), without affecting P2Y1 receptor-induced PLC activity in transfected astrocytoma cells (2).
Related P2 Receptor Antagonists available from Sigma-RBI  
Product #  Product Name Selectivity
D6938 Diinosine pentaphosphate pentasodium salt P2X1/P2X3
M7684 MRS 2159 P2X1
M3808 MRS 2179 P2Y1, competitive
N8652 NF 023 P2X1, potent, selective
P2738 PPNDS P2X1
P-178 PPADS P2
R-115 Reactive Blue 2 P2Y
T4193 TNP-ATP P2X2/3
References
  1. The Sigma-RBI Handbook of Receptor Classification and Signal Transduction, 4th edition, Keith J. Watling, ed., pg. 64 (2001).
  2. Xu, B et al., Acyclic analogues of adensoine bisphosphates as P2Y receptor antagonists: phosphate substitution leads to multiple 4 pathways of inhibition of platelet aggregation, J. Med. Chem., 45, 5694-5709 (2002).
For more information about calcium channel modulators, please visit The Sigma-RBI Handbook of Receptor Classification and Signal Transduction.