Product Highlights
Anti-Glutamate NR1
Antibodies to HDAC
BACE1 Assay Kit
Fluorescent Probes
Isradipine
MRS 2395
Monoclonal Anti g Parvin
Monoclonal Anti-D1
Receptor Preparations
SB-431542
XK469
nNOS Inhibitors
Previous Highlights
New Literature
Ion Channels
Key Resources
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 NSC 95397: Potent, Selective Irreversible Inhibitor of Cdc25 Dual Specificity Phosphatases
Cell Signaling & Neuroscience
Now Available from Sigma-RBI
Prod. No. N 1786
Dual specificity protein phosphatases uniquely dephosphorylate both phosphoserines/threonines and phosphotyrosines on the same protein substrate. They regulate intracellular signaling through mitogen activated and stress activated kinases, oncogenesis and cell cycle progression through G1/S and G2/M checkpoints. The Cdc25 phosphatases have been implicated in cancer and Alzheimer’s disease.1,2 Three Cdc25 homologs exist in humans and are referred to as Cdc25A, Cdc25B, and Cdc25C. They are expressed and activated at different times during the cell cycle. Until recently, potent and selective inhibitors of this enzyme class were not available.
Sigma-RBI is pleased to introduce NSC 95397 as a potent and selective irreversible inhibitor of Cdc25 dual specificity phosphatase:
  • Inhibits in vitro oncogenic, full-length, recombinant human CdC25B with IC50 <500 nM.3
  • Displays mixed inhibition kinetics with in vitro Ki values for Cdc25A, -B, and -C of 32, 96, and 40 nM, respectively.3
  • 125- to 180-fold more selective for Cdc25A than VH1-group of dual specificity phosphatases or protein tyrosine phosphatase PTP1B, respectively.3
  • Significantly inhibits in vitro growth of human and murine carcinoma cells and blocks G2/M cell cycle phase transition.3
Physical Description
Solubility
Molecular Formula
Formula Weight
Sizes
An orange solid
Soluble in DMSO at 16 mg/ml
C14H14O4S2
310.39
5 mg, 25 mg
Structure of N1786
References:
  1. Pestell, K.E., et al., Oncogene, 19, 6607-6612 (2000).
  2. Cangi, M.G., et al., J. Clin. Invest., 106, 753-761 (2000).
  3. Lazo, J.S., et al., Mol. Pharmacol., 61, 720-728 (2002).
back to Cell Signaling
back to Enzyme Explorer