Product Highlights
Anti-Glutamate NR1
Antibodies to HDAC
BACE1 Assay Kit
Fluorescent Probes
Isradipine
MRS 2395
Monoclonal Anti g Parvin
Monoclonal Anti-D1
Receptor Preparations
SB-431542
XK469
nNOS Inhibitors
Previous Highlights
New Literature
Ion Channels
Key Resources
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 Neuronal Nitric Oxide Synthase Inhibitors
Cell Signaling & Neuroscience
Sigma offers two potent peptidomimetic isoform-selective nNOS inhibitors and several related nNOS products.
Nitric oxide (NO) is a potent secondary messenger that is both highly reactive and highly diffusible [1]. It is generated physiologically by a family of enzymes, referred to as NO synthases (NOS, E.C. 1.14.13.39). These enzymes catalyze the oxidation of the L-arginine substrate to L-citrulline and nitric oxide. There are three structurally distinct isoforms of NOS: two are constitutively expressed, neuronal NOS (nNOS, NOS1) and endothelial NOS (eNOS, NOS3), while the third is an inducible form found in macrophages and microglia in the nervous system (iNOS, NOS2). The primary sequence homology is only 50% among isoforms, in contrast to the high sequence conservation observed across species.
NO as a secondary messenger has proven to be a paradox [1]. NO produced by nNOS and eNOS is indispensable in neurotransmission and the regulation of vasodilation. NO produced by iNOS is involved in defense against microbial pathogens. However, the overproduction of NO plays a key role in the pathology of a wide range of disorders including diabetes, inflammatory diseases such as septic shock and arthritis, ischemia-reperfusion injury, stroke, neuropathic pain and various chronic neurodegenerative diseases such as Alzheimer's, Parkinson's and Huntington's.
Given the existence of tissue-specific NOS isoforms and the important physiological roles of NO, the development of isoform-selective NOS inhibitors as therapeutic agents has become an area of active research. Most amino acid-based inhibitors are irreversible and possess minimal selectivity. Nω-Nitro-L-arginine (Prod. No. N5501) is a weak inhibitor of iNOS, displaying an IC50 value of 8.1 µM, and though more potent at nNOS and eNOS, the selectivity is similar, displaying IC50 values of 0.025 µM and 0.09 µM, respectively [2-5]. Similarly, although 7-nitroindazole (Prod. No. N7778) exhibits neuroprotective properties and has been described as a relatively selective nNOS inhibitor, it displays minimally different IC50 values between nNOS and eNOS isoforms (0.71 µM and 0.8 µM, respectively) [6-10]. However, in a series of studies beginning with the screening of a library of conformationally constrained arginine analogs, Huang et al. [11] and Hah et al. [12] identified two novel nNOS inhibitors, specifically L-Nω-Nitroarginine-2,4-L-diaminobutyric amide (L-ArgNO2-L-Dbu-NH2, Prod. No. N3411) and (4S)-N-(4-amino-5-[aminoethyl]aminopentyl)-N'-nitroguanidine (Prod. No. A5727). To date, these compounds have by far the highest selectivity for nNOS, particularly in relation to eNOS (Table 1).
In summary, L-Nω-Nitroarginine-2,4-diaminobutyric amide are potent and highly targeted tools for the study of nNOS functions and its role in neurodegenerative diseases.
Manufactured and sold under license to US patent number 6,274,557.
nNOS Selective Inhibitor Structure Ki (nM) Selectivity
(Ki ratios)
eNOS/nNOS
Selectivity
(Ki ratios)
iNOS/nNOS
L-Nω-Nitroarginine-2,4-L-diamino-butyric amide (Prod. No. N3411)
Structure
130 [11] 1538 [11] 192 [11]
(4S)-N-(4-Amino-5-[aminoethyl]aminopentyl)-N'-nitroguanidine(Prod. No. A5727)
Structure
120 [12] 2617 [12] 325 [12]
NOS Related Products available from Sigma-RBI
Product Name Product # 
2-(Aminoethyl)isothiourea (AET) A5879
Aminoguanidine hemisulfate (also A7259, A8835) A7009
AMT A9834
NG,NG-Dimethyl-L-arginine D0390
NG, NG-Dimethylarginine hydrochloride (ADMA) D4268
Diphenyleneiodonium chloride (DPI) D2926
2-Iminobiotin I4632
L-N6-(1-Iminoethyl)lysine (L-NIL) I8021
L-N5-(1-Iminoethyl)ornithine (L-NIO) I8768
Nω-Methyl-L-arginine (L-NMMA) M7033
non-active L-NMMA M7034
S-Methylisothiourea (SMT) M3127
S-Methyl-L-thiocitrulline M5171
Nω-Nitro-L-arginine (L-NNA) N5501
Nω-Nitro-L-arginine methyl ester (L-NAME) N5751
inactive L-NAME N4770
7-Nitroindazole (7-NI) N7778
1,3-PBIT P8227
1,4-PBIT P8352
TFPI T7188
L-Thiocitrulline 88544
References
  1. Kerwin, J.F. Jr, et al., J. Med. Chem., 38, 4343-4362 (1995).
  2. Reif, D.W. and McCreedy, S.A., Arch. Biochem. Biophys., 320, 170-176 (1995).
  3. Dawson, V.L., et al., Proc. Natl. Acad. Sci. USA, 88, 6368-6371 (1991).
  4. Michel, A.D., et al., Br. J. Pharmacol., 109, 287-288 (1993).
  5. Modin, A., et al., Neuroscience, 62, 189 (1994).
  6. Garthwaite, J., et al., Trends Neurosci., 14, 60-67 (1991).
  7. Moore, P.K., et al., Br. J. Pharmacol., 108, 296-297 (1993).
  8. Wolfe, D.J., et al., Arch. Biochem. Biophys., 314, 360 (1994).
  9. Klatt, P., et al., J. Biol. Chem., 269, 13861-16866 (1994).
  10. Schulz, J.B., et al., J. Neurochem., 64, 936-939 (1995).
  11. Huang H., et al., J. Med. Chem., 43, 2938-2945 (2000).
  12. Hah, J.M. et al., J. Med. Chem., 44, 2667-2670 (2001).
back to top
back to Cell Signaling