Product Highlights
Anti-Glutamate NR1
Antibodies to HDAC
BACE1 Assay Kit
Fluorescent Probes
Isradipine
MRS 2395
Monoclonal Anti g Parvin
Monoclonal Anti-D1
Receptor Preparations
SB-431542
XK469
nNOS Inhibitors
Previous Highlights
New Literature
Ion Channels
Key Resources
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 SB–431542: Selective inhibitor of activin receptor–like kinase (ALK) receptors
Cell Signaling & Neuroscience
Prod. No. S4317
Transforming growth factor β1 (TGF-β1) is a member of a large superfamily of pleiotropic cytokines that are involved in many biological activities, including growth, differentiation, migration, cell survival, and adhesion in diseased and normal states. The members of this superfamily fall into two major branches: TGFβ/Activin/Nodal and BMP/GDF (Bone Morphogenetic Protein/Growth and Differentiation Factor). They have very diverse and often complementary functions. Some are expressed only for short periods during embryonic development and/or only in restricted cell types (e.g. anti–Mullerian hormone, AMH, Inhibin) while others are widespread during embryogenesis and in adult tissues (e.g. TGFβ1 and BMP4). TGF-β1 is a potent regulator in the synthesis of the extracellular matrix (fibrotic factor) and plays a role in wound healing.
TGFβ ligand binding induces receptor complex formation consisting of receptor type II and I, both of which are serine/threonine kinases. The type II receptor phosphorylates and activates the type I receptor within the complex. Seven type I receptors have been identified to date (Activin Receptor-Like Kinases, ALKs 1-7), and five mammalian type II receptors (TβR-II, ActR-II, ActR-IIB, BMPR-II, AMHR-II). The relationship between different TGFβ family ligands and usage of receptor types II and I have been reviewed [1]. Downstream signal transduction takes place when the phosphorylated type I receptor phosphorylates transcription factors, R-Smad or Smad substrates for receptors (Smads 1–8). In general ALK-4, -5, -7, corresponding to the TGFβ/Activin/Nodal branch, phosphorylates Smad-2 and -3, while Smad-1, -5, -8 are substrates for ALK-1, -2, -3, and -6 corresponding to the BMP/GDF branch. These phosphorylated Smads then interact with the co-Smad, Smad 4, at high affinity. These Smad complexes accumulate in the nucleus, are required for the assembly of transcriptional apparatus and directly interact with target genes. [1] Aberrant signaling has been implicated in a number of human diseases: cancer, hereditary hemorrhagic telangiectasia, atherosclerosis, and fibrotic disease of kidney, liver and lung.
SB 431542 inhibits the activity of TGF-β1 activin receptor-like kinases (ALKs). It is a selective and potent inhibitor of the phylogenetically related subset of ALK-4 (activin type I receptor), ALK-5 (TGFβ type I receptor), and ALK-7 (nodal type I receptor). SB 431542 inhibits endogenous activin and TGF-β signaling but is without effect on the more divergent ALK-1, -2, -3, and -6 and hence BMP signaling.[2,3] Phosphorylation of Smad2 by ectopically expressed constitutively active ALK-4, ALK-5, ALK-7 in transfected NIH 3T3 cells is completely abolished by SB 431542 at 10µM.[3] In addition, the compound inhibited ligand dependent activation of wild type ALK-4 and endogenous ALK-5 with IC50 of approximately 0.25µM.[3]
SB 431542 (at 0.3µM) also was shown to inhibit TGFβ1 stimulated proliferation of MG63, a human osteosarcoma cell line expressing ALK-1 that contains another TGFβ type I receptor predominantly present in vascular endothelial cells.[4] This observation remains to be reconciled with earlier results on constitutively active ALK-1 and the phosphorylation of Smad1 [3].
At 1µM SB 431542 stimulates proliferation, differentiation and sheet formation of ESC (embryonic stem cells)-derived endothelial cells and selectively upregulated the expression of claudin-5, an endothelial specific component of tight junctions [5]. SB 413542 may become clinically useful as a regulator of vascular permeability, in modulating bioavailability of drugs, for the production of endothelial cells for cellular therapy and as well as a potential research tool in affecting embryonic vasculogenesis [5].
Sold for research purposes under agreement from GlaxoSmithKline.
References
  1. Massague J., et al., Cell, 103, 295 (2000).
  2. Laping, N.J., et al., Mol. Pharmacol., 62, 58–64 (2002).
  3. Inman, G.J., et al., Mol. Pharmacol., 62, 65–74 (2002).
  4. Matsuyama S., et al., Cancer Res., 63(22), 7791, (2003).
  5. Watabe T., et al.., J Cell Biol., 163(6), 1303-11 (2003).
back to Cell Signaling