Product Highlights
Anti-Glutamate NR1
Antibodies to HDAC
BACE1 Assay Kit
Fluorescent Probes
Isradipine
MRS 2395
Monoclonal Anti g Parvin
Monoclonal Anti-D1
Receptor Preparations
SB-431542
XK469
nNOS Inhibitors
Previous Highlights
New Literature
Ion Channels
Key Resources
Custom Synthesis
Drug Discovery
Epigenetics
Functional Genomics and RNAi
Metabolomics
Molecular Biology
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 SB 202190
Cell Signaling & Neuroscience
Selective, potent and cell permeable inhibitor of p38 MAP kinase.
Prod. No. S 7067
The p38 subfamily of MAP kinases (p38 MAPK) consists of p38a, p38b, p38g, and p38d and regulates gene expression in response to various extracellular stimuli including growth factors, hormones, ligands for G protein coupled receptors, inflammatory cytokines (IL-1, IL-8, TNFa) and stresses (heat, osmotic shock). Because of its critical role in inflammation and stress response, p38 MAPK is of great interest in both basic and therapeutic research.1-3
Sigma-RBI is please to introduce SB 202190, a potent, selective and cell permeable p38 MAPK inhibitor:
  • Binds to the ATP binding site on active p38 MAPK with a Kd value of 38 nM for recombinant p38 and 30-50 nM for the native protein.3
  • Selectively inhibits p38a and b isoforms (IC50 = 50 and 100 nM at SAPK2a/p38 and SAPK2b/p38b2 respectively).4
  • Selectively binds p38 and has no effect on JNK, p42/44MAPK or other multiple related protein kinases at concentrations up to 100 µM.4
  • Induces apoptosis in Jurkat and HeLa cells in vitro with typical apoptotic features including nucleus condensation and DNA fragmentation.5
  • At 1 µmol/l, inhibits p38-MAPK in human mesanglial cells and reduces hexosamine-induced TGFb1 expression at normal (7 mmol/l), as well as high (25 mmol/l) glucose levels, implicating the p38 pathway in the pathophysiology of diabetic glomerulopathy.6
Physical Description:
Solubility:
Molecular Formula:
Formula Weight:
Sizes:
Off-white solid
Soluble in DMSO at 30 mg/ml
C20H14N3OF
331.35
5 mg, 25 mg
SB 202190 Structure
Sold for research purposes under agreement from GlaxoSmithKline
References
  1. Johnson, G.L., et al., Science, 298, 1911-1912 (2002).
  2. Wilson, K.P., et al., Chem. Biol., 4, 423-431 (1997).
  3. Frantz, B., et al., Biochemistry, 37, 13846-13853 (1998).
  4. Davies, S.P., et al., Biochem. J., 351, 95-105 (2000).
  5. Nemoto,S., et al., J. Biol. Chem, 273, 16415-16420 (1998).
  6. Burt, D.J., et al., Diabetologia, 46, 531-537 (2003).
back to Cell Signaling